Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20359406-20359842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDILT
Entrez Gene ID
204474
Gene Name
protein disulfide isomerase like, testis expressed
Gene Aliases
PDIA7
UniGene
Hs.376025
Gene Chromosome Location
Chr.16: 20359170-20404742 on Build GRCh38
Transcripts
NM_174924.1 XM_011545766.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1129037 c.1605G>A p.S535S Chr.16: 20359469-20359469 on Build GRCh38
4415606 c.1586G>A p.G529E Chr.16: 20359488-20359488 on Build GRCh38
317861 c.1508T>A p.L503Q Chr.16: 20359566-20359566 on Build GRCh38
1376376 c.1602G>T p.Q534H Chr.16: 20359472-20359472 on Build GRCh38
4713366 c.1596A>T p.E532D Chr.16: 20359478-20359478 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754027683 Chr.16: 20359740-20359740 on Build GRCh38
rs1001286713 Chr.16: 20359800-20359800 on Build GRCh38
rs376735557 Chr.16: 20359501-20359501 on Build GRCh38
rs758891299 Chr.16: 20359568-20359568 on Build GRCh38
rs761285488 Chr.16: 20359526-20359526 on Build GRCh38
rs767601860 Chr.16: 20359451-20359451 on Build GRCh38
rs746451234 Chr.16: 20359590-20359590 on Build GRCh38
rs776362215 Chr.16: 20359429-20359429 on Build GRCh38
rs768109168 Chr.16: 20359503-20359503 on Build GRCh38
rs544021245 Chr.16: 20359669-20359669 on Build GRCh38
rs757657391 Chr.16: 20359466-20359466 on Build GRCh38
rs767496638 Chr.16: 20359628-20359628 on Build GRCh38
rs754185457 Chr.16: 20359463-20359463 on Build GRCh38
rs149748368 Chr.16: 20359441-20359441 on Build GRCh38
rs921382213 Chr.16: 20359630-20359630 on Build GRCh38
rs528797962 Chr.16: 20359793-20359793 on Build GRCh38
rs959981998 Chr.16: 20359581-20359581 on Build GRCh38
rs9929710 Chr.16: 20359659-20359659 on Build GRCh38
rs757492558 Chr.16: 20359556-20359556 on Build GRCh38
rs1006161155 Chr.16: 20359748-20359748 on Build GRCh38
rs1036704266 Chr.16: 20359687-20359687 on Build GRCh38
rs564742166 Chr.16: 20359767-20359767 on Build GRCh38
rs768140782 Chr.16: 20359594-20359594 on Build GRCh38
rs945830288 Chr.16: 20359432-20359432 on Build GRCh38
rs9652589 Chr.16: 20359494-20359494 on Build GRCh38
rs769626085 Chr.16: 20359504-20359504 on Build GRCh38
rs539799189 Chr.16: 20359552-20359552 on Build GRCh38
rs868443922 Chr.16: 20359531-20359531 on Build GRCh38
rs1050245050 Chr.16: 20359647-20359647 on Build GRCh38
rs9929792 Chr.16: 20359753-20359753 on Build GRCh38
rs374534761 Chr.16: 20359524-20359524 on Build GRCh38
rs766247428 Chr.16: 20359528-20359528 on Build GRCh38
rs753100625 Chr.16: 20359609-20359609 on Build GRCh38
rs942782192 Chr.16: 20359686-20359686 on Build GRCh38
rs148051428 Chr.16: 20359562-20359562 on Build GRCh38
rs9652588 Chr.16: 20359488-20359488 on Build GRCh38
rs992753609 Chr.16: 20359626-20359626 on Build GRCh38
rs772919553 Chr.16: 20359523-20359523 on Build GRCh38
rs750774814 Chr.16: 20359648-20359648 on Build GRCh38
rs778167899 Chr.16: 20359462-20359462 on Build GRCh38
rs139010693 Chr.16: 20359445-20359445 on Build GRCh38
rs143609490 Chr.16: 20359558-20359558 on Build GRCh38
rs780570803 Chr.16: 20359576-20359576 on Build GRCh38
rs114825944 Chr.16: 20359813-20359813 on Build GRCh38
rs141680096 Chr.16: 20359470-20359470 on Build GRCh38
rs377018491 Chr.16: 20359539-20359539 on Build GRCh38
rs374255958 Chr.16: 20359442-20359442 on Build GRCh38
rs772499036 Chr.16: 20359474-20359474 on Build GRCh38
rs922613536 Chr.16: 20359707-20359707 on Build GRCh38
rs980196851 Chr.16: 20359566-20359566 on Build GRCh38
rs766137811 Chr.16: 20359713-20359713 on Build GRCh38
rs755865987 Chr.16: 20359688-20359688 on Build GRCh38
rs780746812 Chr.16: 20359595-20359595 on Build GRCh38
rs1034011356 Chr.16: 20359577-20359577 on Build GRCh38
rs758333097 Chr.16: 20359446-20359446 on Build GRCh38
rs147060285 Chr.16: 20359480-20359480 on Build GRCh38
rs747803544 Chr.16: 20359596-20359596 on Build GRCh38
rs531163281 Chr.16: 20359741-20359741 on Build GRCh38
rs1034068255 Chr.16: 20359809-20359809 on Build GRCh38
rs958485515 Chr.16: 20359810-20359810 on Build GRCh38
rs932735445 Chr.16: 20359640-20359640 on Build GRCh38
rs967224053 Chr.16: 20359653-20359653 on Build GRCh38
rs898946619 Chr.16: 20359769-20359769 on Build GRCh38
rs933953278 Chr.16: 20359733-20359733 on Build GRCh38
rs540790184 Chr.16: 20359499-20359499 on Build GRCh38
rs187070424 Chr.16: 20359770-20359770 on Build GRCh38
rs1021831948 Chr.16: 20359819-20359819 on Build GRCh38
rs1041011820 Chr.16: 20359601-20359601 on Build GRCh38
rs779517495 Chr.16: 20359477-20359477 on Build GRCh38
rs913116255 Chr.16: 20359642-20359642 on Build GRCh38
rs752781445 Chr.16: 20359533-20359533 on Build GRCh38
rs182295392 Chr.16: 20359644-20359644 on Build GRCh38
rs756352839 Chr.16: 20359459-20359459 on Build GRCh38
rs563881981 Chr.16: 20359708-20359708 on Build GRCh38
rs376390632 Chr.16: 20359469-20359469 on Build GRCh38
rs752870974 Chr.16: 20359457-20359457 on Build GRCh38

View Full Product Details