Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7930625-7931134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAPPC1
Entrez Gene ID
58485
Gene Name
trafficking protein particle complex 1
Gene Aliases
BET5, MUM2
UniGene
Hs.24379
Gene Chromosome Location
Chr.17: 7930345-7931999 on Build GRCh38
Transcripts
NM_001166621.1 NM_021210.4 NR_030684.1 NR_030697.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5957590 c.268G>T p.V90L Chr.17: 7931052-7931052 on Build GRCh38
6235305 c.346C>A p.P116T Chr.17: 7930698-7930698 on Build GRCh38
6046297 c.268G>A p.V90M Chr.17: 7931052-7931052 on Build GRCh38
5957589 c.268G>T p.V90L Chr.17: 7931052-7931052 on Build GRCh38
6046298 c.268G>A p.V90M Chr.17: 7931052-7931052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765867659 Chr.17: 7930695-7930695 on Build GRCh38
rs762507520 Chr.17: 7930672-7930672 on Build GRCh38
rs775375603 Chr.17: 7931107-7931107 on Build GRCh38
rs150887254 Chr.17: 7931053-7931053 on Build GRCh38
rs531192432 Chr.17: 7930946-7930946 on Build GRCh38
rs149517464 Chr.17: 7930805-7930805 on Build GRCh38
rs370103498 Chr.17: 7930738-7930738 on Build GRCh38
rs577557281 Chr.17: 7930796-7930796 on Build GRCh38
rs931365021 Chr.17: 7930870-7930870 on Build GRCh38
rs530384123 Chr.17: 7930847-7930847 on Build GRCh38
rs777067198 Chr.17: 7930744-7930744 on Build GRCh38
rs142413849 Chr.17: 7930722-7930722 on Build GRCh38
rs1022185734 Chr.17: 7931031-7931031 on Build GRCh38
rs929349179 Chr.17: 7930692-7930692 on Build GRCh38
rs148237150 Chr.17: 7930953-7930953 on Build GRCh38
rs923581675 Chr.17: 7930782-7930782 on Build GRCh38
rs887135645 Chr.17: 7930934-7930934 on Build GRCh38
rs754497176 Chr.17: 7931106-7931117 on Build GRCh38
rs576484394 Chr.17: 7930707-7930707 on Build GRCh38
rs201092246 Chr.17: 7930756-7930756 on Build GRCh38
rs200473904 Chr.17: 7931080-7931080 on Build GRCh38
rs967844300 Chr.17: 7931040-7931040 on Build GRCh38
rs763835813 Chr.17: 7930999-7930999 on Build GRCh38
rs758570880 Chr.17: 7931011-7931011 on Build GRCh38
rs979135806 Chr.17: 7930771-7930771 on Build GRCh38
rs543964983 Chr.17: 7930726-7930726 on Build GRCh38
rs781760535 Chr.17: 7931024-7931024 on Build GRCh38
rs1030739285 Chr.17: 7931103-7931103 on Build GRCh38
rs773796149 Chr.17: 7931098-7931098 on Build GRCh38
rs749842804 Chr.17: 7930669-7930669 on Build GRCh38
rs148790381 Chr.17: 7930665-7930665 on Build GRCh38
rs1021285055 Chr.17: 7930844-7930844 on Build GRCh38
rs762278370 Chr.17: 7930752-7930752 on Build GRCh38
rs755482202 Chr.17: 7931020-7931020 on Build GRCh38
rs560095556 Chr.17: 7930837-7930837 on Build GRCh38
rs765868796 Chr.17: 7930779-7930779 on Build GRCh38
rs747088319 Chr.17: 7931018-7931018 on Build GRCh38
rs113714495 Chr.17: 7930737-7930737 on Build GRCh38
rs759404889 Chr.17: 7930966-7930966 on Build GRCh38
rs200476704 Chr.17: 7930659-7930659 on Build GRCh38
rs942510786 Chr.17: 7930836-7930836 on Build GRCh38
rs761303350 Chr.17: 7930653-7930653 on Build GRCh38
rs780713914 Chr.17: 7930706-7930706 on Build GRCh38
rs780139319 Chr.17: 7931012-7931012 on Build GRCh38
rs992268697 Chr.17: 7930664-7930664 on Build GRCh38
rs561296623 Chr.17: 7931090-7931090 on Build GRCh38
rs986424001 Chr.17: 7931108-7931108 on Build GRCh38
rs372790067 Chr.17: 7930714-7930714 on Build GRCh38
rs769120006 Chr.17: 7930657-7930657 on Build GRCh38
rs1001142904 Chr.17: 7930848-7930848 on Build GRCh38
rs1051587060 Chr.17: 7931068-7931068 on Build GRCh38
rs921806427 Chr.17: 7931059-7931059 on Build GRCh38
rs1128814 Chr.17: 7930689-7930689 on Build GRCh38
rs752496566 Chr.17: 7930702-7930702 on Build GRCh38
rs768379826 Chr.17: 7930681-7930682 on Build GRCh38
rs959574166 Chr.17: 7930873-7930873 on Build GRCh38
rs1039342250 Chr.17: 7930755-7930755 on Build GRCh38
rs775973899 Chr.17: 7930900-7930900 on Build GRCh38
rs767457581 Chr.17: 7930983-7930983 on Build GRCh38
rs767083574 Chr.17: 7931105-7931105 on Build GRCh38
rs773475992 Chr.17: 7931039-7931039 on Build GRCh38
rs933205953 Chr.17: 7931067-7931067 on Build GRCh38
rs759034968 Chr.17: 7931102-7931102 on Build GRCh38
rs1047123588 Chr.17: 7930928-7930928 on Build GRCh38
rs1039906894 Chr.17: 7930845-7930845 on Build GRCh38
rs755898499 Chr.17: 7930992-7930992 on Build GRCh38
rs747799776 Chr.17: 7930943-7930943 on Build GRCh38
rs751103538 Chr.17: 7930785-7930785 on Build GRCh38
rs182597150 Chr.17: 7930854-7930854 on Build GRCh38
rs112108217 Chr.17: 7931010-7931010 on Build GRCh38
rs751496436 Chr.17: 7930696-7930696 on Build GRCh38
rs373189386 Chr.17: 7930661-7930661 on Build GRCh38
rs770160493 Chr.17: 7931034-7931034 on Build GRCh38
rs747591796 Chr.17: 7930729-7930729 on Build GRCh38
rs371210413 Chr.17: 7930699-7930699 on Build GRCh38
rs571006747 Chr.17: 7930974-7930974 on Build GRCh38
rs143123022 Chr.17: 7930851-7930851 on Build GRCh38
rs1035892248 Chr.17: 7930658-7930658 on Build GRCh38
rs202039662 Chr.17: 7931003-7931003 on Build GRCh38
rs934967111 Chr.17: 7930789-7930789 on Build GRCh38
rs891546624 Chr.17: 7930833-7930833 on Build GRCh38
rs752690883 Chr.17: 7930984-7930984 on Build GRCh38
rs762565885 Chr.17: 7930770-7930770 on Build GRCh38
rs1002822579 Chr.17: 7930937-7930937 on Build GRCh38
rs144394232 Chr.17: 7930651-7930651 on Build GRCh38
rs760998464 Chr.17: 7931026-7931026 on Build GRCh38

View Full Product Details