Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74357100-74357588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD17
Entrez Gene ID
388419
Gene Name
BTB domain containing 17
Gene Aliases
BTBD17A, LGALS3BPL, TANGO10A
UniGene
Hs.211870
Gene Chromosome Location
Chr.17: 74356416-74363655 on Build GRCh38
Transcripts
NM_001080466.1 XM_011524791.2 XM_017024622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6565380 c.718C>T p.R240C Chr.17: 74357376-74357376 on Build GRCh38
4551586 c.532G>A p.G178S Chr.17: 74357562-74357562 on Build GRCh38
5770024 c.597G>C p.V199V Chr.17: 74357497-74357497 on Build GRCh38
4642162 c.865G>C p.A289P Chr.17: 74357229-74357229 on Build GRCh38
5793261 c.966C>T p.G322G Chr.17: 74357128-74357128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770150785 Chr.17: 74357302-74357302 on Build GRCh38
rs757038333 Chr.17: 74357521-74357521 on Build GRCh38
rs979037798 Chr.17: 74357398-74357398 on Build GRCh38
rs368697014 Chr.17: 74357209-74357209 on Build GRCh38
rs752279169 Chr.17: 74357352-74357352 on Build GRCh38
rs1035417086 Chr.17: 74357282-74357282 on Build GRCh38
rs767123529 Chr.17: 74357349-74357349 on Build GRCh38
rs560020969 Chr.17: 74357263-74357263 on Build GRCh38
rs868635314 Chr.17: 74357382-74357382 on Build GRCh38
rs745880632 Chr.17: 74357395-74357395 on Build GRCh38
rs777432466 Chr.17: 74357372-74357372 on Build GRCh38
rs369569004 Chr.17: 74357435-74357435 on Build GRCh38
rs763499874 Chr.17: 74357215-74357215 on Build GRCh38
rs199690597 Chr.17: 74357524-74357524 on Build GRCh38
rs770813450 Chr.17: 74357278-74357278 on Build GRCh38
rs922141420 Chr.17: 74357464-74357464 on Build GRCh38
rs746534227 Chr.17: 74357123-74357123 on Build GRCh38
rs752434742 Chr.17: 74357261-74357261 on Build GRCh38
rs376031460 Chr.17: 74357472-74357472 on Build GRCh38
rs769679223 Chr.17: 74357159-74357159 on Build GRCh38
rs767923924 Chr.17: 74357485-74357485 on Build GRCh38
rs760839363 Chr.17: 74357167-74357167 on Build GRCh38
rs894761078 Chr.17: 74357350-74357350 on Build GRCh38
rs1036345171 Chr.17: 74357179-74357179 on Build GRCh38
rs776710120 Chr.17: 74357290-74357290 on Build GRCh38
rs749589254 Chr.17: 74357557-74357557 on Build GRCh38
rs372419018 Chr.17: 74357242-74357242 on Build GRCh38
rs777658286 Chr.17: 74357161-74357161 on Build GRCh38
rs778114473 Chr.17: 74357551-74357551 on Build GRCh38
rs760198026 Chr.17: 74357507-74357507 on Build GRCh38
rs765761976 Chr.17: 74357201-74357201 on Build GRCh38
rs752160937 Chr.17: 74357342-74357342 on Build GRCh38
rs762150144 Chr.17: 74357292-74357292 on Build GRCh38
rs537000700 Chr.17: 74357166-74357166 on Build GRCh38
rs867823430 Chr.17: 74357376-74357376 on Build GRCh38
rs754997252 Chr.17: 74357430-74357430 on Build GRCh38
rs866094956 Chr.17: 74357419-74357419 on Build GRCh38
rs749954300 Chr.17: 74357228-74357228 on Build GRCh38
rs749291695 Chr.17: 74357162-74357162 on Build GRCh38
rs775024021 Chr.17: 74357494-74357494 on Build GRCh38
rs753529127 Chr.17: 74357518-74357518 on Build GRCh38
rs749651472 Chr.17: 74357465-74357465 on Build GRCh38
rs756315841 Chr.17: 74357547-74357547 on Build GRCh38
rs755695466 Chr.17: 74357358-74357358 on Build GRCh38
rs754543472 Chr.17: 74357255-74357255 on Build GRCh38
rs779765533 Chr.17: 74357122-74357122 on Build GRCh38
rs758286637 Chr.17: 74357536-74357536 on Build GRCh38
rs770103567 Chr.17: 74357451-74357451 on Build GRCh38
rs770866677 Chr.17: 74357165-74357165 on Build GRCh38
rs773647518 Chr.17: 74357314-74357314 on Build GRCh38
rs758937914 Chr.17: 74357345-74357345 on Build GRCh38
rs780932457 Chr.17: 74357136-74357136 on Build GRCh38
rs757091676 Chr.17: 74357388-74357388 on Build GRCh38
rs766947399 Chr.17: 74357337-74357337 on Build GRCh38
rs779891195 Chr.17: 74357539-74357539 on Build GRCh38
rs868137593 Chr.17: 74357480-74357480 on Build GRCh38
rs529825749 Chr.17: 74357535-74357535 on Build GRCh38
rs542481045 Chr.17: 74357500-74357500 on Build GRCh38
rs778787548 Chr.17: 74357389-74357389 on Build GRCh38
rs753580911 Chr.17: 74357378-74357378 on Build GRCh38
rs768818223 Chr.17: 74357286-74357286 on Build GRCh38
rs763713636 Chr.17: 74357515-74357515 on Build GRCh38
rs777603284 Chr.17: 74357267-74357267 on Build GRCh38
rs372146595 Chr.17: 74357471-74357471 on Build GRCh38
rs771409941 Chr.17: 74357565-74357565 on Build GRCh38
rs771462985 Chr.17: 74357466-74357466 on Build GRCh38
rs767000392 Chr.17: 74357229-74357229 on Build GRCh38
rs1038541853 Chr.17: 74357333-74357333 on Build GRCh38
rs191617895 Chr.17: 74357192-74357192 on Build GRCh38
rs377143800 Chr.17: 74357279-74357279 on Build GRCh38
rs780686565 Chr.17: 74357259-74357259 on Build GRCh38
rs748192106 Chr.17: 74357545-74357545 on Build GRCh38
rs763444790 Chr.17: 74357328-74357328 on Build GRCh38
rs768710711 Chr.17: 74357172-74357172 on Build GRCh38
rs748183499 Chr.17: 74357446-74357446 on Build GRCh38
rs977930051 Chr.17: 74357129-74357129 on Build GRCh38
rs535369642 Chr.17: 74357363-74357363 on Build GRCh38
rs749085141 Chr.17: 74357273-74357273 on Build GRCh38
rs773594425 Chr.17: 74357460-74357460 on Build GRCh38
rs200976425 Chr.17: 74357185-74357185 on Build GRCh38
rs912888394 Chr.17: 74357369-74357369 on Build GRCh38
rs971816196 Chr.17: 74357434-74357434 on Build GRCh38
rs9899528 Chr.17: 74357134-74357134 on Build GRCh38
rs747036695 Chr.17: 74357285-74357285 on Build GRCh38

View Full Product Details