Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 49481467-49481971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf32
Entrez Gene ID
497661
Gene Name
chromosome 18 open reading frame 32
Gene Aliases
-
UniGene
Hs.603040
Gene Chromosome Location
Chr.18: 49481178-49487274 on Build GRCh38
Transcripts
NM_001035005.3 NM_001199346.1 NM_001199355.1 NM_001199356.1

Gene Symbol
RPL17-C18orf32
Entrez Gene ID
100526842
Gene Name
RPL17-C18orf32 readthrough
Gene Aliases
-
UniGene
Hs.374588
Gene Chromosome Location
Chr.18: 49481178-49492565 on Build GRCh38
Transcripts
NM_001035005.3 NM_001199346.1 NM_001199355.1 NM_001199356.1

SNP Detail

SNP ID Chromosome Location
rs757498878 Chr.18: 49481663-49481663 on Build GRCh38
rs767713591 Chr.18: 49481576-49481577 on Build GRCh38
rs767356257 Chr.18: 49481495-49481495 on Build GRCh38
rs746310840 Chr.18: 49481857-49481857 on Build GRCh38
rs753318952 Chr.18: 49481548-49481548 on Build GRCh38
rs575237651 Chr.18: 49481687-49481687 on Build GRCh38
rs563130737 Chr.18: 49481643-49481643 on Build GRCh38
rs78252690 Chr.18: 49481825-49481825 on Build GRCh38
rs201090231 Chr.18: 49481577-49481577 on Build GRCh38
rs8087450 Chr.18: 49481533-49481533 on Build GRCh38
rs1049120549 Chr.18: 49481926-49481926 on Build GRCh38
rs1044857651 Chr.18: 49481528-49481528 on Build GRCh38
rs182485698 Chr.18: 49481535-49481535 on Build GRCh38
rs986592849 Chr.18: 49481730-49481730 on Build GRCh38
rs185777205 Chr.18: 49481860-49481860 on Build GRCh38
rs973050931 Chr.18: 49481771-49481771 on Build GRCh38
rs940941402 Chr.18: 49481937-49481937 on Build GRCh38
rs968099672 Chr.18: 49481683-49481683 on Build GRCh38
rs919878841 Chr.18: 49481806-49481806 on Build GRCh38
rs901990901 Chr.18: 49481695-49481695 on Build GRCh38
rs199509158 Chr.18: 49481579-49481579 on Build GRCh38
rs760686998 Chr.18: 49481523-49481523 on Build GRCh38
rs545142602 Chr.18: 49481722-49481722 on Build GRCh38
rs1061816 Chr.18: 49481834-49481834 on Build GRCh38
rs1029127622 Chr.18: 49481741-49481741 on Build GRCh38
rs144242583 Chr.18: 49481577-49481579 on Build GRCh38
rs892000876 Chr.18: 49481539-49481539 on Build GRCh38
rs970937125 Chr.18: 49481866-49481866 on Build GRCh38
rs961236327 Chr.18: 49481745-49481745 on Build GRCh38
rs138987121 Chr.18: 49481603-49481603 on Build GRCh38
rs190248698 Chr.18: 49481513-49481513 on Build GRCh38
rs867674637 Chr.18: 49481734-49481734 on Build GRCh38
rs888064422 Chr.18: 49481635-49481635 on Build GRCh38
rs749925280 Chr.18: 49481880-49481880 on Build GRCh38
rs142236439 Chr.18: 49481700-49481700 on Build GRCh38
rs370088747 Chr.18: 49481749-49481749 on Build GRCh38
rs770400135 Chr.18: 49481925-49481925 on Build GRCh38
rs139800108 Chr.18: 49481875-49481875 on Build GRCh38
rs754552588 Chr.18: 49481549-49481549 on Build GRCh38
rs1000977482 Chr.18: 49481809-49481809 on Build GRCh38
rs560481217 Chr.18: 49481891-49481893 on Build GRCh38
rs778524122 Chr.18: 49481551-49481551 on Build GRCh38
rs200140788 Chr.18: 49481534-49481534 on Build GRCh38
rs146402878 Chr.18: 49481831-49481831 on Build GRCh38
rs1010383197 Chr.18: 49481944-49481944 on Build GRCh38
rs1043677941 Chr.18: 49481945-49481945 on Build GRCh38
rs1044409609 Chr.18: 49481849-49481849 on Build GRCh38
rs555296090 Chr.18: 49481684-49481684 on Build GRCh38
rs750257505 Chr.18: 49481499-49481499 on Build GRCh38
rs956648722 Chr.18: 49481836-49481836 on Build GRCh38
rs909196242 Chr.18: 49481928-49481928 on Build GRCh38
rs577759191 Chr.18: 49481497-49481497 on Build GRCh38
rs1010436882 Chr.18: 49481574-49481574 on Build GRCh38

View Full Product Details