Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 54590969-54591478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNAP
Entrez Gene ID
284254
Gene Name
dynactin associated protein
Gene Aliases
C18orf26
UniGene
Hs.376146
Gene Chromosome Location
Chr.18: 54532713-54599544 on Build GRCh38
Transcripts
NM_001307955.1 NM_173629.2 XM_011525923.2 XM_011525924.2 XM_017025709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
988992 c.8C>T p.A3V Chr.18: 54591212-54591212 on Build GRCh38
227629 c.81G>A p.M27I Chr.18: 54591285-54591285 on Build GRCh38
7218519 c.113T>C p.V38A Chr.18: 54591317-54591317 on Build GRCh38
6083610 c.68G>T p.G23V Chr.18: 54591272-54591272 on Build GRCh38
3891132 c.124G>A p.E42K Chr.18: 54591328-54591328 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968354467 Chr.18: 54591058-54591058 on Build GRCh38
rs777418626 Chr.18: 54591211-54591211 on Build GRCh38
rs918009960 Chr.18: 54591160-54591160 on Build GRCh38
rs934507142 Chr.18: 54591122-54591122 on Build GRCh38
rs867137188 Chr.18: 54591029-54591029 on Build GRCh38
rs772753276 Chr.18: 54591262-54591262 on Build GRCh38
rs530237694 Chr.18: 54591380-54591380 on Build GRCh38
rs766094590 Chr.18: 54591162-54591162 on Build GRCh38
rs760221730 Chr.18: 54591159-54591159 on Build GRCh38
rs752467570 Chr.18: 54591212-54591212 on Build GRCh38
rs776289970 Chr.18: 54591288-54591288 on Build GRCh38
rs199996781 Chr.18: 54590991-54590991 on Build GRCh38
rs975151182 Chr.18: 54591048-54591048 on Build GRCh38
rs1032009350 Chr.18: 54591443-54591443 on Build GRCh38
rs752263666 Chr.18: 54591307-54591307 on Build GRCh38
rs1016858924 Chr.18: 54591096-54591096 on Build GRCh38
rs773416779 Chr.18: 54591266-54591266 on Build GRCh38
rs150110790 Chr.18: 54591236-54591236 on Build GRCh38
rs373641178 Chr.18: 54591352-54591352 on Build GRCh38
rs968385781 Chr.18: 54591420-54591420 on Build GRCh38
rs1049137522 Chr.18: 54591221-54591221 on Build GRCh38
rs965089518 Chr.18: 54591043-54591043 on Build GRCh38
rs949506358 Chr.18: 54591191-54591191 on Build GRCh38
rs752400582 Chr.18: 54591066-54591066 on Build GRCh38
rs779110228 Chr.18: 54591339-54591339 on Build GRCh38
rs573102371 Chr.18: 54591158-54591158 on Build GRCh38
rs914888128 Chr.18: 54591363-54591363 on Build GRCh38
rs978428502 Chr.18: 54591428-54591428 on Build GRCh38
rs911766804 Chr.18: 54591286-54591286 on Build GRCh38
rs749482165 Chr.18: 54591243-54591243 on Build GRCh38
rs962743179 Chr.18: 54591113-54591113 on Build GRCh38
rs955949403 Chr.18: 54591448-54591448 on Build GRCh38
rs747484069 Chr.18: 54591367-54591367 on Build GRCh38
rs377066854 Chr.18: 54591316-54591316 on Build GRCh38
rs935091789 Chr.18: 54590999-54590999 on Build GRCh38
rs753464273 Chr.18: 54591173-54591173 on Build GRCh38
rs35428499 Chr.18: 54591317-54591317 on Build GRCh38
rs1004289497 Chr.18: 54591091-54591091 on Build GRCh38
rs562705168 Chr.18: 54591358-54591358 on Build GRCh38
rs1045127003 Chr.18: 54591197-54591197 on Build GRCh38
rs565086529 Chr.18: 54591248-54591248 on Build GRCh38
rs754435214 Chr.18: 54591193-54591193 on Build GRCh38
rs1045339087 Chr.18: 54591045-54591045 on Build GRCh38
rs118050985 Chr.18: 54591057-54591057 on Build GRCh38
rs1019783305 Chr.18: 54591003-54591003 on Build GRCh38
rs759193155 Chr.18: 54591293-54591293 on Build GRCh38
rs201661625 Chr.18: 54591381-54591381 on Build GRCh38
rs564760470 Chr.18: 54591302-54591302 on Build GRCh38
rs757030914 Chr.18: 54591322-54591322 on Build GRCh38
rs369748790 Chr.18: 54591338-54591338 on Build GRCh38
rs140068793 Chr.18: 54591075-54591075 on Build GRCh38
rs112328465 Chr.18: 54591205-54591205 on Build GRCh38
rs372257198 Chr.18: 54591251-54591251 on Build GRCh38
rs879121541 Chr.18: 54591073-54591073 on Build GRCh38
rs575526371 Chr.18: 54591074-54591074 on Build GRCh38
rs752564137 Chr.18: 54591207-54591207 on Build GRCh38
rs761060818 Chr.18: 54591276-54591276 on Build GRCh38
rs995138196 Chr.18: 54591118-54591118 on Build GRCh38
rs780849946 Chr.18: 54591332-54591332 on Build GRCh38
rs764725166 Chr.18: 54591303-54591303 on Build GRCh38
rs758398341 Chr.18: 54591208-54591208 on Build GRCh38
rs766675393 Chr.18: 54591278-54591278 on Build GRCh38
rs777980055 Chr.18: 54591157-54591158 on Build GRCh38
rs746873933 Chr.18: 54591217-54591217 on Build GRCh38
rs576849928 Chr.18: 54591315-54591315 on Build GRCh38
rs754103944 Chr.18: 54591337-54591337 on Build GRCh38
rs548898517 Chr.18: 54591095-54591095 on Build GRCh38
rs369887288 Chr.18: 54591421-54591421 on Build GRCh38
rs368060476 Chr.18: 54591127-54591127 on Build GRCh38
rs549822086 Chr.18: 54591004-54591004 on Build GRCh38
rs914943704 Chr.18: 54591015-54591015 on Build GRCh38

View Full Product Details