Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7120539-7120991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSR
Entrez Gene ID
3643
Gene Name
insulin receptor
Gene Aliases
CD220, HHF5
UniGene
Hs.465744
Gene Chromosome Location
Chr.19: 7112255-7294405 on Build GRCh38
Transcripts
NM_000208.3 NM_001079817.2 XM_011527988.2 XM_011527989.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4082686 c.3560C>T p.T1187M Chr.19: 7120719-7120719 on Build GRCh38
6971144 c.3602G>A p.R1201Q Chr.19: 7120677-7120677 on Build GRCh38
3541175 c.3610G>A p.A1204T Chr.19: 7120669-7120669 on Build GRCh38
3836123 c.3556G>C p.E1186Q Chr.19: 7120723-7120723 on Build GRCh38
6983864 c.3595C>T p.P1199S Chr.19: 7120684-7120684 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146863879 Chr.19: 7120837-7120837 on Build GRCh38
rs371494268 Chr.19: 7120788-7120788 on Build GRCh38
rs778681780 Chr.19: 7120658-7120658 on Build GRCh38
rs183693669 Chr.19: 7120867-7120867 on Build GRCh38
rs121913150 Chr.19: 7120707-7120707 on Build GRCh38
rs200677311 Chr.19: 7120591-7120591 on Build GRCh38
rs767608313 Chr.19: 7120772-7120772 on Build GRCh38
rs200281940 Chr.19: 7120590-7120590 on Build GRCh38
rs751821207 Chr.19: 7120603-7120603 on Build GRCh38
rs777694266 Chr.19: 7120946-7120946 on Build GRCh38
rs781430127 Chr.19: 7120624-7120624 on Build GRCh38
rs752858033 Chr.19: 7120773-7120773 on Build GRCh38
rs1040810001 Chr.19: 7120812-7120812 on Build GRCh38
rs538094849 Chr.19: 7120962-7120962 on Build GRCh38
rs1008340759 Chr.19: 7120810-7120810 on Build GRCh38
rs779765143 Chr.19: 7120662-7120662 on Build GRCh38
rs768944050 Chr.19: 7120613-7120613 on Build GRCh38
rs369258369 Chr.19: 7120570-7120570 on Build GRCh38
rs201189144 Chr.19: 7120764-7120764 on Build GRCh38
rs199989287 Chr.19: 7120786-7120786 on Build GRCh38
rs763471377 Chr.19: 7120718-7120718 on Build GRCh38
rs758264537 Chr.19: 7120785-7120785 on Build GRCh38
rs750339226 Chr.19: 7120779-7120779 on Build GRCh38
rs771839433 Chr.19: 7120661-7120661 on Build GRCh38
rs747762725 Chr.19: 7120792-7120792 on Build GRCh38
rs761206818 Chr.19: 7120697-7120697 on Build GRCh38
rs769176650 Chr.19: 7120801-7120801 on Build GRCh38
rs766776682 Chr.19: 7120596-7120596 on Build GRCh38
rs994357035 Chr.19: 7120920-7120920 on Build GRCh38
rs541076178 Chr.19: 7120856-7120856 on Build GRCh38
rs770374822 Chr.19: 7120700-7120700 on Build GRCh38
rs959845205 Chr.19: 7120585-7120585 on Build GRCh38
rs1015507626 Chr.19: 7120960-7120960 on Build GRCh38
rs752667089 Chr.19: 7120628-7120628 on Build GRCh38
rs1014712480 Chr.19: 7120915-7120915 on Build GRCh38
rs121913157 Chr.19: 7120739-7120739 on Build GRCh38
rs1015073698 Chr.19: 7120587-7120587 on Build GRCh38
rs1003600682 Chr.19: 7120911-7120911 on Build GRCh38
rs529172045 Chr.19: 7120918-7120918 on Build GRCh38
rs762413781 Chr.19: 7120576-7120576 on Build GRCh38
rs1044722934 Chr.19: 7120865-7120865 on Build GRCh38
rs79750603 Chr.19: 7120637-7120637 on Build GRCh38
rs753279286 Chr.19: 7120611-7120611 on Build GRCh38
rs752446959 Chr.19: 7120866-7120866 on Build GRCh38
rs191551615 Chr.19: 7120861-7120861 on Build GRCh38
rs755198022 Chr.19: 7120619-7120619 on Build GRCh38
rs899581659 Chr.19: 7120858-7120858 on Build GRCh38
rs766867210 Chr.19: 7120719-7120719 on Build GRCh38
rs893727131 Chr.19: 7120842-7120842 on Build GRCh38
rs201905854 Chr.19: 7120796-7120796 on Build GRCh38
rs76799388 Chr.19: 7120752-7120752 on Build GRCh38
rs886054686 Chr.19: 7120708-7120708 on Build GRCh38
rs911204095 Chr.19: 7120708-7120708 on Build GRCh38
rs555633697 Chr.19: 7120705-7120705 on Build GRCh38
rs757264298 Chr.19: 7120632-7120632 on Build GRCh38
rs146546375 Chr.19: 7120664-7120664 on Build GRCh38
rs548918657 Chr.19: 7120926-7120926 on Build GRCh38
rs145399067 Chr.19: 7120753-7120753 on Build GRCh38
rs759749789 Chr.19: 7120600-7120601 on Build GRCh38
rs52800171 Chr.19: 7120620-7120620 on Build GRCh38
rs376750105 Chr.19: 7120776-7120776 on Build GRCh38
rs776228235 Chr.19: 7120688-7120688 on Build GRCh38
rs569996961 Chr.19: 7120916-7120916 on Build GRCh38
rs532999701 Chr.19: 7120863-7120863 on Build GRCh38
rs373534538 Chr.19: 7120789-7120789 on Build GRCh38
rs121913156 Chr.19: 7120677-7120677 on Build GRCh38

View Full Product Details