Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7690032-7690531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCER2
Entrez Gene ID
2208
Gene Name
Fc fragment of IgE receptor II
Gene Aliases
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF
UniGene
Hs.465778
Gene Chromosome Location
Chr.19: 7688757-7702755 on Build GRCh38
Transcripts
NM_001207019.2 NM_001220500.1 NM_002002.4 XM_005272462.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4445982 c.621+3G>A p.? Chr.19: 7690403-7690403 on Build GRCh38
6580999 c.574G>A p.D192N Chr.19: 7690453-7690453 on Build GRCh38
4518537 c.688_689GG>AA p.G230K Chr.19: 7690198-7690199 on Build GRCh38
1751266 c.521G>C p.C174S Chr.19: 7690506-7690506 on Build GRCh38
6662664 c.651C>A p.T217T Chr.19: 7690236-7690236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751787016 Chr.19: 7690506-7690506 on Build GRCh38
rs4996973 Chr.19: 7690273-7690273 on Build GRCh38
rs377148193 Chr.19: 7690074-7690074 on Build GRCh38
rs757378683 Chr.19: 7690316-7690316 on Build GRCh38
rs4996978 Chr.19: 7690056-7690056 on Build GRCh38
rs746104507 Chr.19: 7690357-7690357 on Build GRCh38
rs769204272 Chr.19: 7690370-7690370 on Build GRCh38
rs768462140 Chr.19: 7690500-7690500 on Build GRCh38
rs753914101 Chr.19: 7690311-7690311 on Build GRCh38
rs139487727 Chr.19: 7690475-7690475 on Build GRCh38
rs957194543 Chr.19: 7690395-7690395 on Build GRCh38
rs182569239 Chr.19: 7690497-7690497 on Build GRCh38
rs889849476 Chr.19: 7690080-7690080 on Build GRCh38
rs764749532 Chr.19: 7690283-7690283 on Build GRCh38
rs148383467 Chr.19: 7690355-7690356 on Build GRCh38
rs911429952 Chr.19: 7690495-7690495 on Build GRCh38
rs781488649 Chr.19: 7690447-7690447 on Build GRCh38
rs144440869 Chr.19: 7690451-7690451 on Build GRCh38
rs775906963 Chr.19: 7690236-7690236 on Build GRCh38
rs762589833 Chr.19: 7690290-7690290 on Build GRCh38
rs377564979 Chr.19: 7690411-7690411 on Build GRCh38
rs775997166 Chr.19: 7690167-7690167 on Build GRCh38
rs771142988 Chr.19: 7690432-7690432 on Build GRCh38
rs2229229 Chr.19: 7690188-7690188 on Build GRCh38
rs866995831 Chr.19: 7690203-7690203 on Build GRCh38
rs748919449 Chr.19: 7690465-7690465 on Build GRCh38
rs149647877 Chr.19: 7690348-7690348 on Build GRCh38
rs201422970 Chr.19: 7690120-7690120 on Build GRCh38
rs774327816 Chr.19: 7690404-7690404 on Build GRCh38
rs771903938 Chr.19: 7690484-7690484 on Build GRCh38
rs373313846 Chr.19: 7690379-7690379 on Build GRCh38
rs757460159 Chr.19: 7690222-7690222 on Build GRCh38
rs765054864 Chr.19: 7690191-7690191 on Build GRCh38
rs867413961 Chr.19: 7690444-7690444 on Build GRCh38
rs772683395 Chr.19: 7690398-7690398 on Build GRCh38
rs4996972 Chr.19: 7690327-7690327 on Build GRCh38
rs776049068 Chr.19: 7690279-7690279 on Build GRCh38
rs773302708 Chr.19: 7690361-7690362 on Build GRCh38
rs375077946 Chr.19: 7690415-7690415 on Build GRCh38
rs375114070 Chr.19: 7690367-7690367 on Build GRCh38
rs748218616 Chr.19: 7690274-7690283 on Build GRCh38
rs528496860 Chr.19: 7690368-7690368 on Build GRCh38
rs141227304 Chr.19: 7690405-7690405 on Build GRCh38
rs768137028 Chr.19: 7690269-7690269 on Build GRCh38
rs191142459 Chr.19: 7690372-7690372 on Build GRCh38
rs4996975 Chr.19: 7690085-7690085 on Build GRCh38
rs72558010 Chr.19: 7690328-7690328 on Build GRCh38
rs770845770 Chr.19: 7690403-7690403 on Build GRCh38
rs934015813 Chr.19: 7690373-7690373 on Build GRCh38
rs765369748 Chr.19: 7690418-7690418 on Build GRCh38
rs748703601 Chr.19: 7690450-7690450 on Build GRCh38
rs1038551504 Chr.19: 7690440-7690440 on Build GRCh38
rs770755660 Chr.19: 7690468-7690468 on Build GRCh38
rs571314475 Chr.19: 7690107-7690107 on Build GRCh38
rs1025959340 Chr.19: 7690248-7690248 on Build GRCh38
rs542938135 Chr.19: 7690351-7690351 on Build GRCh38
rs764237783 Chr.19: 7690309-7690309 on Build GRCh38
rs1042429 Chr.19: 7690197-7690197 on Build GRCh38
rs989938926 Chr.19: 7690397-7690397 on Build GRCh38
rs4996976 Chr.19: 7690082-7690082 on Build GRCh38
rs2228138 Chr.19: 7690170-7690170 on Build GRCh38
rs767633435 Chr.19: 7690402-7690403 on Build GRCh38
rs565338813 Chr.19: 7690416-7690416 on Build GRCh38
rs761508625 Chr.19: 7690282-7690282 on Build GRCh38
rs4996977 Chr.19: 7690078-7690078 on Build GRCh38
rs28364072 Chr.19: 7690399-7690399 on Build GRCh38
rs200648991 Chr.19: 7690464-7690464 on Build GRCh38
rs747524495 Chr.19: 7690243-7690243 on Build GRCh38
rs114265483 Chr.19: 7690053-7690053 on Build GRCh38
rs750593910 Chr.19: 7690421-7690421 on Build GRCh38
rs758701728 Chr.19: 7690361-7690361 on Build GRCh38
rs755369130 Chr.19: 7690446-7690446 on Build GRCh38
rs990622247 Chr.19: 7690312-7690312 on Build GRCh38
rs959158304 Chr.19: 7690304-7690304 on Build GRCh38
rs766292864 Chr.19: 7690297-7690297 on Build GRCh38
rs763047133 Chr.19: 7690499-7690499 on Build GRCh38
rs774169912 Chr.19: 7690472-7690472 on Build GRCh38
rs267605763 Chr.19: 7690483-7690483 on Build GRCh38
rs756205377 Chr.19: 7690216-7690216 on Build GRCh38
rs868346681 Chr.19: 7690121-7690121 on Build GRCh38
rs138319893 Chr.19: 7690308-7690308 on Build GRCh38
rs778961760 Chr.19: 7690356-7690356 on Build GRCh38
rs200319365 Chr.19: 7690132-7690132 on Build GRCh38
rs369443024 Chr.19: 7690417-7690417 on Build GRCh38
rs772952733 Chr.19: 7690287-7690287 on Build GRCh38
rs377575240 Chr.19: 7690502-7690502 on Build GRCh38
rs758952802 Chr.19: 7690441-7690441 on Build GRCh38
rs760134645 Chr.19: 7690169-7690169 on Build GRCh38
rs181329074 Chr.19: 7690100-7690100 on Build GRCh38
rs536515427 Chr.19: 7690496-7690496 on Build GRCh38
rs73921522 Chr.19: 7690145-7690145 on Build GRCh38
rs201998118 Chr.19: 7690217-7690217 on Build GRCh38
rs566145810 Chr.19: 7690081-7690081 on Build GRCh38
rs915166798 Chr.19: 7690151-7690151 on Build GRCh38
rs144339823 Chr.19: 7690231-7690231 on Build GRCh38
rs558619258 Chr.19: 7690229-7690229 on Build GRCh38
rs758683190 Chr.19: 7690428-7690428 on Build GRCh38
rs138488307 Chr.19: 7690196-7690196 on Build GRCh38
rs373413003 Chr.19: 7690494-7690494 on Build GRCh38
rs751530180 Chr.19: 7690305-7690305 on Build GRCh38
rs148627411 Chr.19: 7690235-7690235 on Build GRCh38
rs369340108 Chr.19: 7690206-7690206 on Build GRCh38

View Full Product Details