Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11399752-11400241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGL3
Entrez Gene ID
57139
Gene Name
ral guanine nucleotide dissociation stimulator like 3
Gene Aliases
-
UniGene
Hs.375142
Gene Chromosome Location
Chr.19: 11394056-11419342 on Build GRCh38
Transcripts
NM_001035223.3 NM_001161616.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4860283 c.1680G>A p.G560G Chr.19: 11399939-11399939 on Build GRCh38
7261739 c.1581-5C>A p.? Chr.19: 11400113-11400113 on Build GRCh38
7261738 c.1581-5C>A p.? Chr.19: 11400113-11400113 on Build GRCh38
7326898 c.1709C>T p.P570L Chr.19: 11399892-11399892 on Build GRCh38
5646732 c.1592G>A p.R531Q Chr.19: 11400097-11400097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745857402 Chr.19: 11399861-11399862 on Build GRCh38
rs753983006 Chr.19: 11400019-11400019 on Build GRCh38
rs761507920 Chr.19: 11400065-11400065 on Build GRCh38
rs550237910 Chr.19: 11400079-11400079 on Build GRCh38
rs749059511 Chr.19: 11399775-11399775 on Build GRCh38
rs763284945 Chr.19: 11400126-11400126 on Build GRCh38
rs751378810 Chr.19: 11400048-11400048 on Build GRCh38
rs753264735 Chr.19: 11399878-11399878 on Build GRCh38
rs1023826064 Chr.19: 11399946-11399946 on Build GRCh38
rs372106275 Chr.19: 11400202-11400202 on Build GRCh38
rs200388133 Chr.19: 11399916-11399916 on Build GRCh38
rs942818272 Chr.19: 11399829-11399829 on Build GRCh38
rs767951568 Chr.19: 11399977-11399977 on Build GRCh38
rs375198789 Chr.19: 11399845-11399845 on Build GRCh38
rs950558142 Chr.19: 11399914-11399914 on Build GRCh38
rs762256894 Chr.19: 11400104-11400104 on Build GRCh38
rs367981353 Chr.19: 11400193-11400193 on Build GRCh38
rs769639500 Chr.19: 11399958-11399958 on Build GRCh38
rs375599580 Chr.19: 11400020-11400020 on Build GRCh38
rs369125029 Chr.19: 11400194-11400194 on Build GRCh38
rs758507741 Chr.19: 11400044-11400044 on Build GRCh38
rs780955123 Chr.19: 11399846-11399847 on Build GRCh38
rs317912 Chr.19: 11399796-11399796 on Build GRCh38
rs544727410 Chr.19: 11399871-11399871 on Build GRCh38
rs779664424 Chr.19: 11399915-11399915 on Build GRCh38
rs370917602 Chr.19: 11399970-11399970 on Build GRCh38
rs533934433 Chr.19: 11399776-11399776 on Build GRCh38
rs928148950 Chr.19: 11399922-11399922 on Build GRCh38
rs761215818 Chr.19: 11399994-11399994 on Build GRCh38
rs369245791 Chr.19: 11399825-11399825 on Build GRCh38
rs527990270 Chr.19: 11399932-11399932 on Build GRCh38
rs1031960580 Chr.19: 11399792-11399792 on Build GRCh38
rs755024551 Chr.19: 11400050-11400050 on Build GRCh38
rs755968851 Chr.19: 11400154-11400154 on Build GRCh38
rs767710617 Chr.19: 11399874-11399874 on Build GRCh38
rs939607739 Chr.19: 11399930-11399930 on Build GRCh38
rs145606309 Chr.19: 11399890-11399890 on Build GRCh38
rs768549039 Chr.19: 11400016-11400016 on Build GRCh38
rs146472545 Chr.19: 11400066-11400066 on Build GRCh38
rs763104231 Chr.19: 11400015-11400019 on Build GRCh38
rs750439107 Chr.19: 11400116-11400116 on Build GRCh38
rs1023378146 Chr.19: 11400189-11400189 on Build GRCh38
rs775681385 Chr.19: 11399990-11399990 on Build GRCh38
rs185391263 Chr.19: 11400136-11400136 on Build GRCh38
rs900321328 Chr.19: 11399859-11399859 on Build GRCh38
rs374124810 Chr.19: 11400192-11400192 on Build GRCh38
rs776939368 Chr.19: 11400197-11400197 on Build GRCh38
rs747650026 Chr.19: 11399955-11399955 on Build GRCh38
rs773425612 Chr.19: 11400206-11400206 on Build GRCh38
rs148913854 Chr.19: 11400045-11400045 on Build GRCh38
rs746008565 Chr.19: 11399899-11399899 on Build GRCh38
rs769752308 Chr.19: 11399929-11399930 on Build GRCh38
rs777387748 Chr.19: 11400161-11400161 on Build GRCh38
rs373073975 Chr.19: 11399901-11399901 on Build GRCh38
rs994456480 Chr.19: 11400145-11400145 on Build GRCh38
rs973088397 Chr.19: 11399875-11399875 on Build GRCh38
rs532489333 Chr.19: 11400150-11400150 on Build GRCh38
rs149671936 Chr.19: 11399889-11399889 on Build GRCh38
rs748730868 Chr.19: 11400175-11400175 on Build GRCh38
rs750678706 Chr.19: 11400026-11400026 on Build GRCh38
rs544273850 Chr.19: 11399819-11399819 on Build GRCh38
rs775239320 Chr.19: 11399938-11399939 on Build GRCh38
rs145674805 Chr.19: 11400201-11400201 on Build GRCh38
rs770679737 Chr.19: 11400176-11400176 on Build GRCh38
rs1017195297 Chr.19: 11399820-11399820 on Build GRCh38
rs781272925 Chr.19: 11400058-11400058 on Build GRCh38
rs377576323 Chr.19: 11399972-11399972 on Build GRCh38
rs924164404 Chr.19: 11399995-11399995 on Build GRCh38
rs372755592 Chr.19: 11399827-11399827 on Build GRCh38
rs765170967 Chr.19: 11400024-11400024 on Build GRCh38
rs768302919 Chr.19: 11399851-11399851 on Build GRCh38
rs935436336 Chr.19: 11400062-11400062 on Build GRCh38
rs942724338 Chr.19: 11400046-11400046 on Build GRCh38
rs749538460 Chr.19: 11399807-11399807 on Build GRCh38
rs148120187 Chr.19: 11400211-11400211 on Build GRCh38
rs771966871 Chr.19: 11399828-11399828 on Build GRCh38
rs752707539 Chr.19: 11400151-11400151 on Build GRCh38
rs868470326 Chr.19: 11399950-11399950 on Build GRCh38
rs577557917 Chr.19: 11399861-11399861 on Build GRCh38
rs562480010 Chr.19: 11399844-11399844 on Build GRCh38
rs757434618 Chr.19: 11399813-11399813 on Build GRCh38
rs776330286 Chr.19: 11399952-11399952 on Build GRCh38
rs768940364 Chr.19: 11400097-11400097 on Build GRCh38
rs766497598 Chr.19: 11400132-11400132 on Build GRCh38
rs915260141 Chr.19: 11400035-11400035 on Build GRCh38
rs762459338 Chr.19: 11399976-11399976 on Build GRCh38
rs189588847 Chr.19: 11400060-11400060 on Build GRCh38
rs770464221 Chr.19: 11400068-11400068 on Build GRCh38
rs779188410 Chr.19: 11399893-11399893 on Build GRCh38
rs369645825 Chr.19: 11399891-11399891 on Build GRCh38
rs377749544 Chr.19: 11400074-11400074 on Build GRCh38
rs754827235 Chr.19: 11400143-11400143 on Build GRCh38
rs774638166 Chr.19: 11399872-11399872 on Build GRCh38
rs924238069 Chr.19: 11399782-11399782 on Build GRCh38
rs759942872 Chr.19: 11399873-11399873 on Build GRCh38
rs765388303 Chr.19: 11400115-11400115 on Build GRCh38
rs898811123 Chr.19: 11400158-11400158 on Build GRCh38
rs764568386 Chr.19: 11400001-11400001 on Build GRCh38
rs777133565 Chr.19: 11400098-11400098 on Build GRCh38
rs141019468 Chr.19: 11400067-11400067 on Build GRCh38
rs145451138 Chr.19: 11400064-11400064 on Build GRCh38
rs555616320 Chr.19: 11399793-11399793 on Build GRCh38

View Full Product Details