Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36545564-36546109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF529
Entrez Gene ID
57711
Gene Name
zinc finger protein 529
Gene Aliases
-
UniGene
Hs.654960
Gene Chromosome Location
Chr.19: 36534491-36605411 on Build GRCh38
Transcripts
NM_001145649.1 NM_001145650.1 NM_001321351.1 NM_020951.4 XM_006723302.3 XM_006723304.2 XM_011527164.2 XM_011527165.2 XM_011527166.2 XM_011527167.2 XM_011527169.2 XM_011527170.1 XM_017027039.1 XM_017027040.1 XM_017027041.1 XM_017027042.1 XM_017027043.1 XM_017027044.1 XM_017027045.1

SNP Detail

SNP ID Chromosome Location
rs913551689 Chr.19: 36545899-36545899 on Build GRCh38
rs953866333 Chr.19: 36545867-36545867 on Build GRCh38
rs61648978 Chr.19: 36546063-36546063 on Build GRCh38
rs749463364 Chr.19: 36545687-36545688 on Build GRCh38
rs73034119 Chr.19: 36545851-36545851 on Build GRCh38
rs962917079 Chr.19: 36545803-36545803 on Build GRCh38
rs34309889 Chr.19: 36546061-36546064 on Build GRCh38
rs1003150755 Chr.19: 36546058-36546065 on Build GRCh38
rs751157470 Chr.19: 36545896-36545897 on Build GRCh38
rs770008090 Chr.19: 36546046-36546049 on Build GRCh38
rs905478791 Chr.19: 36546053-36546053 on Build GRCh38
rs200517643 Chr.19: 36546079-36546082 on Build GRCh38
rs542149578 Chr.19: 36545691-36545691 on Build GRCh38
rs956265970 Chr.19: 36546064-36546064 on Build GRCh38
rs369074531 Chr.19: 36545995-36545996 on Build GRCh38
rs750460092 Chr.19: 36546048-36546049 on Build GRCh38
rs72240473 Chr.19: 36546046-36546047 on Build GRCh38
rs376780375 Chr.19: 36545783-36545783 on Build GRCh38
rs368851421 Chr.19: 36545870-36545870 on Build GRCh38
rs1053170920 Chr.19: 36545917-36545917 on Build GRCh38
rs749695661 Chr.19: 36546060-36546065 on Build GRCh38
rs1040162480 Chr.19: 36545932-36545932 on Build GRCh38
rs912274733 Chr.19: 36545887-36545887 on Build GRCh38
rs375987417 Chr.19: 36546059-36546060 on Build GRCh38
rs960720411 Chr.19: 36545871-36545871 on Build GRCh38
rs1022983259 Chr.19: 36546046-36546046 on Build GRCh38
rs541142979 Chr.19: 36545878-36545878 on Build GRCh38
rs901908783 Chr.19: 36545599-36545602 on Build GRCh38
rs185102587 Chr.19: 36546021-36546021 on Build GRCh38
rs970315695 Chr.19: 36545718-36545718 on Build GRCh38
rs906100295 Chr.19: 36546058-36546063 on Build GRCh38
rs947067012 Chr.19: 36546007-36546007 on Build GRCh38
rs397859248 Chr.19: 36546080-36546081 on Build GRCh38
rs201960962 Chr.19: 36546081-36546082 on Build GRCh38
rs1004579745 Chr.19: 36545750-36545750 on Build GRCh38
rs201905035 Chr.19: 36546081-36546081 on Build GRCh38
rs10593037 Chr.19: 36546062-36546063 on Build GRCh38
rs137928661 Chr.19: 36545736-36545736 on Build GRCh38
rs896535798 Chr.19: 36546078-36546078 on Build GRCh38
rs1006198507 Chr.19: 36546045-36546046 on Build GRCh38
rs374818611 Chr.19: 36546011-36546011 on Build GRCh38
rs760895852 Chr.19: 36545726-36545727 on Build GRCh38
rs564655994 Chr.19: 36545905-36545905 on Build GRCh38
rs28396441 Chr.19: 36546061-36546061 on Build GRCh38
rs374313902 Chr.19: 36546065-36546065 on Build GRCh38
rs58537943 Chr.19: 36546061-36546062 on Build GRCh38
rs901722477 Chr.19: 36545933-36545933 on Build GRCh38
rs1035858537 Chr.19: 36546062-36546069 on Build GRCh38
rs993618516 Chr.19: 36545607-36545607 on Build GRCh38
rs960819191 Chr.19: 36545739-36545739 on Build GRCh38
rs140442780 Chr.19: 36546060-36546063 on Build GRCh38
rs779806867 Chr.19: 36545776-36545776 on Build GRCh38
rs1006639927 Chr.19: 36545695-36545695 on Build GRCh38
rs905344433 Chr.19: 36545662-36545662 on Build GRCh38
rs943305195 Chr.19: 36545930-36545930 on Build GRCh38
rs921202972 Chr.19: 36545865-36545865 on Build GRCh38
rs981577688 Chr.19: 36545734-36545734 on Build GRCh38
rs910446907 Chr.19: 36545919-36545919 on Build GRCh38
rs558196763 Chr.19: 36545589-36545589 on Build GRCh38
rs200800725 Chr.19: 36546080-36546080 on Build GRCh38
rs1023182844 Chr.19: 36545706-36545706 on Build GRCh38
rs887861369 Chr.19: 36546041-36546049 on Build GRCh38
rs372203861 Chr.19: 36546059-36546059 on Build GRCh38
rs929371282 Chr.19: 36545976-36545976 on Build GRCh38
rs373092651 Chr.19: 36545879-36545879 on Build GRCh38
rs938156767 Chr.19: 36546055-36546055 on Build GRCh38
rs914264192 Chr.19: 36545963-36545963 on Build GRCh38
rs934435668 Chr.19: 36545915-36545915 on Build GRCh38
rs575637944 Chr.19: 36545821-36545821 on Build GRCh38

View Full Product Details