Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37738691-37739194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF573
Entrez Gene ID
126231
Gene Name
zinc finger protein 573
Gene Aliases
-
UniGene
Hs.531262
Gene Chromosome Location
Chr.19: 37738274-37779626 on Build GRCh38
Transcripts
NM_001172689.1 NM_001172690.1 NM_001172691.1 NM_001172692.1 NM_152360.3 XM_011526446.2 XM_017026275.1 XM_017026276.1 XM_017026277.1 XM_017026278.1 XM_017026279.1 XM_017026280.1 XM_017026281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1680969 c.1400C>A p.P467H Chr.19: 37738916-37738916 on Build GRCh38
3201161 c.1348_1349insA p.T450fs*6 Chr.19: 37739141-37739142 on Build GRCh38
4869037 c.1627C>A p.L543I Chr.19: 37738863-37738863 on Build GRCh38
4000705 c.1567A>G p.M523V Chr.19: 37738923-37738923 on Build GRCh38
4841646 c.1285C>G p.Q429E Chr.19: 37739031-37739031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374186327 Chr.19: 37738796-37738796 on Build GRCh38
rs763637323 Chr.19: 37739141-37739142 on Build GRCh38
rs777080821 Chr.19: 37738997-37738997 on Build GRCh38
rs866062483 Chr.19: 37738806-37738806 on Build GRCh38
rs976666294 Chr.19: 37739028-37739028 on Build GRCh38
rs779404892 Chr.19: 37739164-37739164 on Build GRCh38
rs565238008 Chr.19: 37738948-37738948 on Build GRCh38
rs777790984 Chr.19: 37738745-37738745 on Build GRCh38
rs752422037 Chr.19: 37739131-37739131 on Build GRCh38
rs941450673 Chr.19: 37738716-37738716 on Build GRCh38
rs765470608 Chr.19: 37738859-37738859 on Build GRCh38
rs768308982 Chr.19: 37738819-37738819 on Build GRCh38
rs758024794 Chr.19: 37739129-37739129 on Build GRCh38
rs11666245 Chr.19: 37739025-37739025 on Build GRCh38
rs778975091 Chr.19: 37738980-37738980 on Build GRCh38
rs1037462447 Chr.19: 37738742-37738742 on Build GRCh38
rs745521977 Chr.19: 37739155-37739155 on Build GRCh38
rs139662850 Chr.19: 37739142-37739142 on Build GRCh38
rs181127839 Chr.19: 37739115-37739115 on Build GRCh38
rs3095726 Chr.19: 37738923-37738923 on Build GRCh38
rs756220666 Chr.19: 37738977-37738977 on Build GRCh38
rs143695781 Chr.19: 37738986-37738986 on Build GRCh38
rs766709906 Chr.19: 37738945-37738948 on Build GRCh38
rs761927757 Chr.19: 37738966-37738966 on Build GRCh38
rs868786154 Chr.19: 37738768-37738768 on Build GRCh38
rs368821224 Chr.19: 37739111-37739111 on Build GRCh38
rs141819433 Chr.19: 37739066-37739066 on Build GRCh38
rs755955995 Chr.19: 37738913-37738913 on Build GRCh38
rs376659072 Chr.19: 37738812-37738812 on Build GRCh38
rs150282268 Chr.19: 37739086-37739086 on Build GRCh38
rs867008744 Chr.19: 37738894-37738894 on Build GRCh38
rs893583903 Chr.19: 37739092-37739092 on Build GRCh38
rs865831443 Chr.19: 37738748-37738749 on Build GRCh38
rs780905290 Chr.19: 37738788-37738788 on Build GRCh38
rs773384963 Chr.19: 37739114-37739114 on Build GRCh38
rs145613747 Chr.19: 37738758-37738758 on Build GRCh38
rs768094297 Chr.19: 37738765-37738765 on Build GRCh38
rs772456780 Chr.19: 37739113-37739113 on Build GRCh38
rs759767934 Chr.19: 37739129-37739129 on Build GRCh38
rs758836604 Chr.19: 37738886-37738886 on Build GRCh38
rs773855958 Chr.19: 37738832-37738832 on Build GRCh38
rs780055976 Chr.19: 37738978-37738978 on Build GRCh38
rs547267383 Chr.19: 37738749-37738750 on Build GRCh38
rs772940795 Chr.19: 37738934-37738934 on Build GRCh38
rs747633696 Chr.19: 37738931-37738931 on Build GRCh38
rs752387866 Chr.19: 37739049-37739050 on Build GRCh38
rs146360251 Chr.19: 37739124-37739124 on Build GRCh38
rs761619945 Chr.19: 37738829-37738829 on Build GRCh38
rs761028477 Chr.19: 37739043-37739043 on Build GRCh38
rs368362975 Chr.19: 37738797-37738797 on Build GRCh38
rs145815699 Chr.19: 37738907-37738907 on Build GRCh38
rs750051275 Chr.19: 37739161-37739161 on Build GRCh38
rs777399173 Chr.19: 37739075-37739075 on Build GRCh38
rs139015807 Chr.19: 37739091-37739091 on Build GRCh38
rs781442836 Chr.19: 37738905-37738905 on Build GRCh38
rs184252524 Chr.19: 37738981-37738981 on Build GRCh38
rs763271370 Chr.19: 37739024-37739024 on Build GRCh38
rs780638297 Chr.19: 37739147-37739147 on Build GRCh38
rs897126137 Chr.19: 37738787-37738787 on Build GRCh38
rs778635803 Chr.19: 37738925-37738925 on Build GRCh38
rs147912292 Chr.19: 37738961-37738961 on Build GRCh38
rs751676498 Chr.19: 37738962-37738962 on Build GRCh38
rs543224868 Chr.19: 37738891-37738891 on Build GRCh38
rs759924604 Chr.19: 37738853-37738853 on Build GRCh38
rs758496227 Chr.19: 37738722-37738722 on Build GRCh38
rs746688169 Chr.19: 37739000-37739000 on Build GRCh38
rs561805450 Chr.19: 37738953-37738953 on Build GRCh38
rs767443068 Chr.19: 37738973-37738973 on Build GRCh38
rs761059999 Chr.19: 37738912-37738912 on Build GRCh38
rs775899001 Chr.19: 37739020-37739020 on Build GRCh38
rs746496935 Chr.19: 37739081-37739081 on Build GRCh38
rs561491855 Chr.19: 37738902-37738902 on Build GRCh38
rs755701654 Chr.19: 37739159-37739159 on Build GRCh38
rs550543312 Chr.19: 37739146-37739146 on Build GRCh38
rs751020931 Chr.19: 37739142-37739142 on Build GRCh38
rs753234663 Chr.19: 37738877-37738877 on Build GRCh38
rs868036443 Chr.19: 37738841-37738841 on Build GRCh38
rs745669946 Chr.19: 37738794-37738794 on Build GRCh38
rs778813502 Chr.19: 37739062-37739062 on Build GRCh38
rs758212565 Chr.19: 37739072-37739072 on Build GRCh38
rs745471816 Chr.19: 37739090-37739090 on Build GRCh38
rs372863613 Chr.19: 37739027-37739027 on Build GRCh38
rs750684774 Chr.19: 37738974-37738974 on Build GRCh38
rs141383055 Chr.19: 37738832-37738832 on Build GRCh38
rs141664555 Chr.19: 37738979-37738979 on Build GRCh38
rs547540724 Chr.19: 37738957-37738957 on Build GRCh38
rs767636394 Chr.19: 37739031-37739031 on Build GRCh38
rs776903031 Chr.19: 37738839-37738839 on Build GRCh38
rs201545219 Chr.19: 37739026-37739026 on Build GRCh38
rs866678309 Chr.19: 37739058-37739058 on Build GRCh38
rs757411916 Chr.19: 37738766-37738766 on Build GRCh38
rs774509315 Chr.19: 37739098-37739098 on Build GRCh38
rs369461477 Chr.19: 37738814-37738814 on Build GRCh38
rs372094990 Chr.19: 37739050-37739050 on Build GRCh38
rs758426457 Chr.19: 37739136-37739136 on Build GRCh38
rs765137370 Chr.19: 37739130-37739130 on Build GRCh38
rs1008609025 Chr.19: 37738730-37738731 on Build GRCh38
rs751625200 Chr.19: 37738897-37738897 on Build GRCh38
rs773945681 Chr.19: 37738824-37738824 on Build GRCh38
rs931798454 Chr.19: 37739102-37739102 on Build GRCh38
rs774975744 Chr.19: 37739014-37739014 on Build GRCh38
rs779670343 Chr.19: 37738916-37738916 on Build GRCh38
rs756942189 Chr.19: 37739084-37739084 on Build GRCh38
rs189598355 Chr.19: 37739044-37739044 on Build GRCh38
rs878948289 Chr.19: 37738924-37738924 on Build GRCh38
rs760205658 Chr.19: 37738935-37738935 on Build GRCh38
rs750459899 Chr.19: 37739037-37739037 on Build GRCh38
rs770476433 Chr.19: 37739009-37739009 on Build GRCh38
rs766957074 Chr.19: 37739119-37739119 on Build GRCh38
rs532539977 Chr.19: 37738954-37738954 on Build GRCh38

View Full Product Details