Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47183802-47184290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDZK1IP1
Entrez Gene ID
10158
Gene Name
PDZK1 interacting protein 1
Gene Aliases
DD96, MAP17, SPAP
UniGene
Hs.431099
Gene Chromosome Location
Chr.1: 47183589-47190099 on Build GRCh38
Transcripts
NM_005764.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4517333 c.319_320GG>AA p.G107N Chr.1: 47183996-47183997 on Build GRCh38
6349646 c.275C>G p.S92C Chr.1: 47184041-47184041 on Build GRCh38
910400 c.339G>A p.P113P Chr.1: 47183977-47183977 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs146814645 Chr.1: 47184009-47184009 on Build GRCh38
rs753785604 Chr.1: 47184022-47184022 on Build GRCh38
rs912690486 Chr.1: 47183911-47183911 on Build GRCh38
rs200439136 Chr.1: 47184078-47184078 on Build GRCh38
rs878953587 Chr.1: 47184050-47184050 on Build GRCh38
rs1041714922 Chr.1: 47183999-47183999 on Build GRCh38
rs888733869 Chr.1: 47183833-47183834 on Build GRCh38
rs771246961 Chr.1: 47184073-47184073 on Build GRCh38
rs372102575 Chr.1: 47183910-47183910 on Build GRCh38
rs372277620 Chr.1: 47184001-47184001 on Build GRCh38
rs529047330 Chr.1: 47183977-47183977 on Build GRCh38
rs954705702 Chr.1: 47183895-47183895 on Build GRCh38
rs538757747 Chr.1: 47183886-47183886 on Build GRCh38
rs780411412 Chr.1: 47183858-47183858 on Build GRCh38
rs929720312 Chr.1: 47183855-47183855 on Build GRCh38
rs771730125 Chr.1: 47184090-47184090 on Build GRCh38
rs574186049 Chr.1: 47184082-47184082 on Build GRCh38
rs1005747671 Chr.1: 47183834-47183834 on Build GRCh38
rs778019139 Chr.1: 47184071-47184071 on Build GRCh38
rs370399612 Chr.1: 47183924-47183924 on Build GRCh38
rs989404895 Chr.1: 47184188-47184188 on Build GRCh38
rs927943975 Chr.1: 47184238-47184238 on Build GRCh38
rs996342318 Chr.1: 47183870-47183870 on Build GRCh38
rs974432086 Chr.1: 47184091-47184091 on Build GRCh38
rs141341861 Chr.1: 47183988-47183988 on Build GRCh38
rs748471450 Chr.1: 47183965-47183967 on Build GRCh38
rs559430062 Chr.1: 47184030-47184030 on Build GRCh38
rs761516985 Chr.1: 47184006-47184006 on Build GRCh38
rs1030048818 Chr.1: 47183874-47183874 on Build GRCh38
rs754875617 Chr.1: 47184033-47184033 on Build GRCh38
rs141109844 Chr.1: 47184255-47184255 on Build GRCh38
rs1002923180 Chr.1: 47184018-47184018 on Build GRCh38
rs781545171 Chr.1: 47184077-47184077 on Build GRCh38
rs930086942 Chr.1: 47184241-47184241 on Build GRCh38
rs770266089 Chr.1: 47184081-47184081 on Build GRCh38
rs942480855 Chr.1: 47184199-47184199 on Build GRCh38
rs1035797744 Chr.1: 47184036-47184036 on Build GRCh38
rs370702290 Chr.1: 47183978-47183978 on Build GRCh38
rs75551432 Chr.1: 47184127-47184127 on Build GRCh38
rs564051793 Chr.1: 47184200-47184200 on Build GRCh38
rs948043603 Chr.1: 47183989-47183989 on Build GRCh38
rs749446327 Chr.1: 47183900-47183900 on Build GRCh38
rs568268980 Chr.1: 47183970-47183970 on Build GRCh38
rs1015909677 Chr.1: 47183843-47183843 on Build GRCh38
rs912324223 Chr.1: 47184196-47184196 on Build GRCh38
rs539485353 Chr.1: 47184019-47184019 on Build GRCh38
rs768148219 Chr.1: 47184003-47184003 on Build GRCh38
rs752777684 Chr.1: 47184043-47184043 on Build GRCh38
rs778992931 Chr.1: 47184042-47184042 on Build GRCh38
rs553958497 Chr.1: 47184133-47184133 on Build GRCh38
rs201160255 Chr.1: 47184055-47184055 on Build GRCh38
rs189688166 Chr.1: 47184267-47184267 on Build GRCh38
rs995817820 Chr.1: 47184087-47184087 on Build GRCh38
rs1802762 Chr.1: 47183892-47183892 on Build GRCh38
rs973747534 Chr.1: 47183844-47183844 on Build GRCh38
rs1047215478 Chr.1: 47183823-47183823 on Build GRCh38
rs986488922 Chr.1: 47184148-47184148 on Build GRCh38
rs761265769 Chr.1: 47184084-47184084 on Build GRCh38
rs747256229 Chr.1: 47184072-47184072 on Build GRCh38
rs778890014 Chr.1: 47183957-47183957 on Build GRCh38
rs202226248 Chr.1: 47184054-47184054 on Build GRCh38
rs191581058 Chr.1: 47183981-47183981 on Build GRCh38
rs184154071 Chr.1: 47184070-47184070 on Build GRCh38
rs531624744 Chr.1: 47183863-47183863 on Build GRCh38
rs772947838 Chr.1: 47184094-47184094 on Build GRCh38
rs914763171 Chr.1: 47184259-47184259 on Build GRCh38
rs747415599 Chr.1: 47183987-47183987 on Build GRCh38
rs766297053 Chr.1: 47184021-47184021 on Build GRCh38
rs999256260 Chr.1: 47184107-47184107 on Build GRCh38
rs982167074 Chr.1: 47184235-47184235 on Build GRCh38
rs918287776 Chr.1: 47183853-47183853 on Build GRCh38
rs545852052 Chr.1: 47184175-47184175 on Build GRCh38
rs982807939 Chr.1: 47184257-47184257 on Build GRCh38
rs890246968 Chr.1: 47184095-47184095 on Build GRCh38
rs767138184 Chr.1: 47184007-47184007 on Build GRCh38
rs572161795 Chr.1: 47184142-47184142 on Build GRCh38

View Full Product Details