Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 48298403-48298901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA6
Entrez Gene ID
54558
Gene Name
spermatogenesis associated 6
Gene Aliases
HASH, SRF-1, SRF1
UniGene
Hs.538103
Gene Chromosome Location
Chr.1: 48258643-48472208 on Build GRCh38
Transcripts
NM_001286238.1 NM_001286239.1 NM_019073.3 XM_006710699.2 XM_006710700.2 XM_011541606.1 XM_011541607.2 XM_011541608.1 XM_011541609.2 XM_017001510.1 XM_017001511.1 XM_017001512.1 XM_017001513.1 XM_017001514.1 XM_017001515.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6063753 c.1376G>T p.R459L Chr.1: 48298804-48298804 on Build GRCh38
5678374 c.1379C>T p.P460L Chr.1: 48298801-48298801 on Build GRCh38
4931933 c.1411A>T p.R471W Chr.1: 48298769-48298769 on Build GRCh38
372521 c.1305T>A p.R435R Chr.1: 48298875-48298875 on Build GRCh38
3726332 c.1291T>A p.C431S Chr.1: 48298889-48298889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967652300 Chr.1: 48298443-48298443 on Build GRCh38
rs199508980 Chr.1: 48298682-48298682 on Build GRCh38
rs867306075 Chr.1: 48298809-48298809 on Build GRCh38
rs1030861821 Chr.1: 48298580-48298580 on Build GRCh38
rs146976708 Chr.1: 48298840-48298840 on Build GRCh38
rs759341817 Chr.1: 48298734-48298734 on Build GRCh38
rs777123534 Chr.1: 48298791-48298791 on Build GRCh38
rs752637975 Chr.1: 48298739-48298739 on Build GRCh38
rs980438084 Chr.1: 48298459-48298459 on Build GRCh38
rs1056042 Chr.1: 48298747-48298747 on Build GRCh38
rs41289160 Chr.1: 48298780-48298780 on Build GRCh38
rs756003946 Chr.1: 48298854-48298854 on Build GRCh38
rs776274309 Chr.1: 48298732-48298732 on Build GRCh38
rs769080849 Chr.1: 48298876-48298876 on Build GRCh38
rs529359370 Chr.1: 48298567-48298567 on Build GRCh38
rs926363239 Chr.1: 48298475-48298475 on Build GRCh38
rs749751160 Chr.1: 48298775-48298775 on Build GRCh38
rs780417764 Chr.1: 48298774-48298774 on Build GRCh38
rs949042247 Chr.1: 48298456-48298456 on Build GRCh38
rs760209757 Chr.1: 48298721-48298721 on Build GRCh38
rs748734328 Chr.1: 48298746-48298747 on Build GRCh38
rs772806686 Chr.1: 48298703-48298703 on Build GRCh38
rs970811192 Chr.1: 48298651-48298651 on Build GRCh38
rs138130686 Chr.1: 48298847-48298847 on Build GRCh38
rs755545124 Chr.1: 48298776-48298776 on Build GRCh38
rs1048255698 Chr.1: 48298481-48298481 on Build GRCh38
rs527462674 Chr.1: 48298627-48298627 on Build GRCh38
rs11205467 Chr.1: 48298550-48298550 on Build GRCh38
rs147329302 Chr.1: 48298853-48298853 on Build GRCh38
rs150203833 Chr.1: 48298827-48298827 on Build GRCh38
rs141923322 Chr.1: 48298594-48298594 on Build GRCh38
rs144278281 Chr.1: 48298804-48298804 on Build GRCh38
rs140934334 Chr.1: 48298810-48298810 on Build GRCh38
rs776849926 Chr.1: 48298737-48298739 on Build GRCh38
rs757540548 Chr.1: 48298672-48298672 on Build GRCh38
rs574649626 Chr.1: 48298591-48298591 on Build GRCh38
rs749808377 Chr.1: 48298686-48298686 on Build GRCh38
rs893994786 Chr.1: 48298877-48298877 on Build GRCh38
rs374077798 Chr.1: 48298683-48298683 on Build GRCh38
rs374515413 Chr.1: 48298769-48298769 on Build GRCh38
rs367557644 Chr.1: 48298789-48298789 on Build GRCh38
rs781545066 Chr.1: 48298767-48298767 on Build GRCh38
rs754245448 Chr.1: 48298867-48298867 on Build GRCh38
rs781390686 Chr.1: 48298677-48298677 on Build GRCh38
rs1046589263 Chr.1: 48298458-48298458 on Build GRCh38
rs766695219 Chr.1: 48298858-48298858 on Build GRCh38
rs901078469 Chr.1: 48298793-48298793 on Build GRCh38
rs775466906 Chr.1: 48298836-48298836 on Build GRCh38
rs958166261 Chr.1: 48298619-48298619 on Build GRCh38
rs981783905 Chr.1: 48298654-48298654 on Build GRCh38
rs770165896 Chr.1: 48298776-48298777 on Build GRCh38
rs771433294 Chr.1: 48298692-48298692 on Build GRCh38
rs756358463 Chr.1: 48298856-48298856 on Build GRCh38
rs997499557 Chr.1: 48298574-48298574 on Build GRCh38
rs1004140786 Chr.1: 48298526-48298526 on Build GRCh38
rs534688671 Chr.1: 48298607-48298607 on Build GRCh38
rs377754671 Chr.1: 48298737-48298737 on Build GRCh38
rs926864234 Chr.1: 48298472-48298472 on Build GRCh38
rs373440220 Chr.1: 48298761-48298761 on Build GRCh38
rs1036914025 Chr.1: 48298548-48298548 on Build GRCh38
rs762777924 Chr.1: 48298839-48298839 on Build GRCh38
rs746583980 Chr.1: 48298796-48298796 on Build GRCh38
rs1007720261 Chr.1: 48298598-48298598 on Build GRCh38
rs751693618 Chr.1: 48298670-48298670 on Build GRCh38
rs370778856 Chr.1: 48298805-48298805 on Build GRCh38
rs937852545 Chr.1: 48298476-48298476 on Build GRCh38
rs371064726 Chr.1: 48298743-48298743 on Build GRCh38
rs560432471 Chr.1: 48298626-48298626 on Build GRCh38
rs770663268 Chr.1: 48298726-48298726 on Build GRCh38
rs756411278 Chr.1: 48298770-48298770 on Build GRCh38
rs192222178 Chr.1: 48298509-48298509 on Build GRCh38
rs757409063 Chr.1: 48298764-48298764 on Build GRCh38

View Full Product Details