Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52841966-52842478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF468
Entrez Gene ID
90333
Gene Name
zinc finger protein 468
Gene Aliases
-
UniGene
Hs.467223
Gene Chromosome Location
Chr.19: 52838532-52857649 on Build GRCh38
Transcripts
NM_001008801.1 NM_001277120.1 NR_102299.1 NR_102300.1 NR_102301.1 XM_017027443.1 XM_017027444.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3538221 c.262C>T p.H88Y Chr.19: 52842032-52842032 on Build GRCh38
4537928 c.247G>C p.G83R Chr.19: 52842047-52842047 on Build GRCh38
4000889 c.143-10A>T p.? Chr.19: 52842161-52842161 on Build GRCh38
2928085 c.225C>A p.H75Q Chr.19: 52842069-52842069 on Build GRCh38
5094942 c.265G>A p.E89K Chr.19: 52842029-52842029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746107395 Chr.19: 52842100-52842100 on Build GRCh38
rs544795577 Chr.19: 52842218-52842218 on Build GRCh38
rs60738058 Chr.19: 52842428-52842428 on Build GRCh38
rs764633766 Chr.19: 52842153-52842153 on Build GRCh38
rs765034867 Chr.19: 52842064-52842064 on Build GRCh38
rs765153385 Chr.19: 52842124-52842124 on Build GRCh38
rs773114327 Chr.19: 52842202-52842202 on Build GRCh38
rs573937131 Chr.19: 52842392-52842393 on Build GRCh38
rs779264109 Chr.19: 52842167-52842167 on Build GRCh38
rs763927904 Chr.19: 52842036-52842036 on Build GRCh38
rs776809100 Chr.19: 52842199-52842199 on Build GRCh38
rs147901953 Chr.19: 52842002-52842002 on Build GRCh38
rs111934739 Chr.19: 52842325-52842325 on Build GRCh38
rs756701199 Chr.19: 52842083-52842083 on Build GRCh38
rs910010860 Chr.19: 52842321-52842321 on Build GRCh38
rs769792123 Chr.19: 52842201-52842201 on Build GRCh38
rs62117470 Chr.19: 52842262-52842262 on Build GRCh38
rs1032880809 Chr.19: 52842217-52842217 on Build GRCh38
rs753756859 Chr.19: 52842042-52842042 on Build GRCh38
rs751147150 Chr.19: 52842148-52842148 on Build GRCh38
rs56941243 Chr.19: 52842410-52842410 on Build GRCh38
rs953190094 Chr.19: 52842283-52842283 on Build GRCh38
rs576696694 Chr.19: 52842200-52842200 on Build GRCh38
rs932793720 Chr.19: 52842147-52842147 on Build GRCh38
rs534145246 Chr.19: 52842442-52842442 on Build GRCh38
rs941526874 Chr.19: 52842324-52842324 on Build GRCh38
rs762677205 Chr.19: 52842138-52842138 on Build GRCh38
rs373366254 Chr.19: 52842184-52842184 on Build GRCh38
rs765934259 Chr.19: 52842143-52842143 on Build GRCh38
rs776690541 Chr.19: 52842117-52842117 on Build GRCh38
rs772128288 Chr.19: 52842107-52842107 on Build GRCh38
rs60596790 Chr.19: 52842194-52842194 on Build GRCh38
rs562368324 Chr.19: 52842289-52842289 on Build GRCh38
rs57083197 Chr.19: 52842441-52842441 on Build GRCh38
rs761789847 Chr.19: 52842123-52842123 on Build GRCh38
rs984953602 Chr.19: 52842307-52842307 on Build GRCh38
rs753938639 Chr.19: 52842308-52842308 on Build GRCh38
rs186391051 Chr.19: 52842452-52842452 on Build GRCh38
rs763683414 Chr.19: 52842119-52842121 on Build GRCh38
rs796540391 Chr.19: 52842427-52842428 on Build GRCh38
rs564665407 Chr.19: 52842251-52842251 on Build GRCh38
rs376666708 Chr.19: 52842088-52842088 on Build GRCh38
rs749691791 Chr.19: 52842094-52842094 on Build GRCh38
rs61425250 Chr.19: 52842395-52842395 on Build GRCh38
rs775366270 Chr.19: 52842029-52842029 on Build GRCh38
rs768882803 Chr.19: 52842198-52842198 on Build GRCh38
rs1010070014 Chr.19: 52842227-52842228 on Build GRCh38
rs148965254 Chr.19: 52842118-52842118 on Build GRCh38
rs142157651 Chr.19: 52842228-52842228 on Build GRCh38
rs760676482 Chr.19: 52842030-52842030 on Build GRCh38
rs750119524 Chr.19: 52842067-52842067 on Build GRCh38
rs754375946 Chr.19: 52842158-52842158 on Build GRCh38
rs754413796 Chr.19: 52842151-52842151 on Build GRCh38
rs750973583 Chr.19: 52842077-52842077 on Build GRCh38
rs569775000 Chr.19: 52842054-52842054 on Build GRCh38
rs761671324 Chr.19: 52842046-52842046 on Build GRCh38
rs8100996 Chr.19: 52842161-52842161 on Build GRCh38
rs58201419 Chr.19: 52842427-52842427 on Build GRCh38
rs775701078 Chr.19: 52842108-52842108 on Build GRCh38
rs1045772624 Chr.19: 52842354-52842354 on Build GRCh38
rs747059163 Chr.19: 52842109-52842109 on Build GRCh38
rs112374127 Chr.19: 52842329-52842329 on Build GRCh38
rs755632092 Chr.19: 52842243-52842243 on Build GRCh38
rs544085115 Chr.19: 52842285-52842285 on Build GRCh38
rs115710495 Chr.19: 52842195-52842195 on Build GRCh38
rs758717168 Chr.19: 52842190-52842190 on Build GRCh38
rs182009074 Chr.19: 52842327-52842327 on Build GRCh38
rs897133841 Chr.19: 52842169-52842169 on Build GRCh38
rs965655086 Chr.19: 52842242-52842244 on Build GRCh38
rs773070649 Chr.19: 52842131-52842131 on Build GRCh38
rs555245251 Chr.19: 52842110-52842110 on Build GRCh38
rs8100893 Chr.19: 52842068-52842068 on Build GRCh38
rs113914462 Chr.19: 52842319-52842319 on Build GRCh38
rs368832182 Chr.19: 52842196-52842196 on Build GRCh38
rs1028623027 Chr.19: 52842443-52842443 on Build GRCh38
rs779211124 Chr.19: 52842099-52842099 on Build GRCh38
rs60660813 Chr.19: 52842358-52842358 on Build GRCh38
rs60582014 Chr.19: 52842403-52842403 on Build GRCh38
rs547755174 Chr.19: 52842351-52842351 on Build GRCh38
rs551532146 Chr.19: 52842015-52842015 on Build GRCh38
rs1029156856 Chr.19: 52842280-52842280 on Build GRCh38

View Full Product Details