Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54738312-54738815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIR2DL3
Entrez Gene ID
3804
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 3
Gene Aliases
CD158B2, CD158b, GL183, KIR-023GB, KIR-K7b, KIR-K7c, KIR2DS3, KIR2DS5, KIRCL23, NKAT, NKAT2, NKAT2A, NKAT2B, p58
UniGene
Hs.654608
Gene Chromosome Location
Chr.19: 54738509-54753052 on Build GRCh38
Transcripts
NM_015868.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3538984 c.15C>T p.V5V Chr.19: 54738560-54738560 on Build GRCh38
4985814 c.10A>C p.M4L Chr.19: 54738555-54738555 on Build GRCh38
4985813 c.10A>C p.M4L Chr.19: 54738555-54738555 on Build GRCh38
4287124 c.26T>C p.V9A Chr.19: 54738571-54738571 on Build GRCh38
118791 c.33T>A p.V11V Chr.19: 54738578-54738578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111826346 Chr.19: 54738518-54738518 on Build GRCh38
rs753946981 Chr.19: 54738611-54738611 on Build GRCh38
rs10415421 Chr.19: 54738525-54738525 on Build GRCh38
rs761681200 Chr.19: 54738598-54738598 on Build GRCh38
rs772472290 Chr.19: 54738561-54738561 on Build GRCh38
rs755792064 Chr.19: 54738627-54738627 on Build GRCh38
rs373412527 Chr.19: 54738418-54738418 on Build GRCh38
rs758944125 Chr.19: 54738529-54738529 on Build GRCh38
rs375579828 Chr.19: 54738593-54738593 on Build GRCh38
rs3810344 Chr.19: 54738596-54738596 on Build GRCh38
rs368378 Chr.19: 54738792-54738792 on Build GRCh38
rs1024168156 Chr.19: 54738348-54738348 on Build GRCh38
rs529967167 Chr.19: 54738563-54738563 on Build GRCh38
rs753242502 Chr.19: 54738616-54738616 on Build GRCh38
rs368374 Chr.19: 54738789-54738789 on Build GRCh38
rs766313083 Chr.19: 54738603-54738603 on Build GRCh38
rs748497705 Chr.19: 54738560-54738560 on Build GRCh38
rs561174842 Chr.19: 54738557-54738557 on Build GRCh38
rs778220227 Chr.19: 54738621-54738621 on Build GRCh38
rs774491734 Chr.19: 54738597-54738597 on Build GRCh38
rs747170169 Chr.19: 54738478-54738478 on Build GRCh38
rs781358433 Chr.19: 54738513-54738513 on Build GRCh38
rs188621146 Chr.19: 54738683-54738683 on Build GRCh38
rs55862502 Chr.19: 54738782-54738782 on Build GRCh38
rs934224990 Chr.19: 54738647-54738647 on Build GRCh38
rs192994413 Chr.19: 54738748-54738748 on Build GRCh38
rs10414999 Chr.19: 54738370-54738370 on Build GRCh38
rs542226112 Chr.19: 54738496-54738496 on Build GRCh38
rs538558431 Chr.19: 54738396-54738396 on Build GRCh38
rs761771065 Chr.19: 54738544-54738544 on Build GRCh38
rs750709166 Chr.19: 54738550-54738550 on Build GRCh38
rs779272367 Chr.19: 54738520-54738520 on Build GRCh38
rs768863768 Chr.19: 54738519-54738519 on Build GRCh38
rs980435592 Chr.19: 54738402-54738402 on Build GRCh38
rs201216657 Chr.19: 54738599-54738599 on Build GRCh38
rs1042533081 Chr.19: 54738562-54738562 on Build GRCh38
rs56297109 Chr.19: 54738770-54738770 on Build GRCh38
rs565123739 Chr.19: 54738524-54738524 on Build GRCh38
rs55813315 Chr.19: 54738753-54738753 on Build GRCh38
rs796707921 Chr.19: 54738441-54738441 on Build GRCh38
rs751409500 Chr.19: 54738543-54738543 on Build GRCh38
rs762966104 Chr.19: 54738536-54738536 on Build GRCh38
rs542915405 Chr.19: 54738769-54738769 on Build GRCh38
rs555105570 Chr.19: 54738690-54738690 on Build GRCh38
rs3810343 Chr.19: 54738571-54738571 on Build GRCh38
rs767842737 Chr.19: 54738548-54738548 on Build GRCh38
rs370736325 Chr.19: 54738614-54738614 on Build GRCh38
rs574797049 Chr.19: 54738709-54738709 on Build GRCh38
rs796155146 Chr.19: 54738697-54738715 on Build GRCh38
rs558879038 Chr.19: 54738497-54738497 on Build GRCh38
rs550889258 Chr.19: 54738556-54738556 on Build GRCh38
rs938028599 Chr.19: 54738467-54738467 on Build GRCh38
rs56256013 Chr.19: 54738738-54738738 on Build GRCh38
rs569481537 Chr.19: 54738667-54738667 on Build GRCh38
rs769133926 Chr.19: 54738585-54738585 on Build GRCh38
rs374653140 Chr.19: 54738551-54738551 on Build GRCh38
rs936184588 Chr.19: 54738734-54738734 on Build GRCh38
rs755109161 Chr.19: 54738613-54738613 on Build GRCh38
rs199707006 Chr.19: 54738618-54738618 on Build GRCh38
rs766657143 Chr.19: 54738717-54738717 on Build GRCh38
rs200849113 Chr.19: 54738735-54738736 on Build GRCh38
rs377555763 Chr.19: 54738600-54738600 on Build GRCh38
rs56088901 Chr.19: 54738725-54738725 on Build GRCh38
rs972547735 Chr.19: 54738335-54738335 on Build GRCh38
rs763919169 Chr.19: 54738538-54738538 on Build GRCh38
rs772013498 Chr.19: 54738387-54738387 on Build GRCh38
rs745451232 Chr.19: 54738626-54738626 on Build GRCh38
rs111263451 Chr.19: 54738744-54738744 on Build GRCh38
rs756277741 Chr.19: 54738517-54738517 on Build GRCh38
rs371181432 Chr.19: 54738547-54738547 on Build GRCh38
rs573343981 Chr.19: 54738427-54738427 on Build GRCh38
rs554764038 Chr.19: 54738369-54738369 on Build GRCh38
rs72487146 Chr.19: 54738711-54738711 on Build GRCh38
rs552566722 Chr.19: 54738788-54738788 on Build GRCh38
rs750469223 Chr.19: 54738516-54738516 on Build GRCh38
rs56014926 Chr.19: 54738758-54738758 on Build GRCh38
rs757562255 Chr.19: 54738501-54738501 on Build GRCh38
rs878917900 Chr.19: 54738386-54738386 on Build GRCh38
rs575733363 Chr.19: 54738393-54738393 on Build GRCh38
rs377407133 Chr.19: 54738535-54738535 on Build GRCh38
rs544745580 Chr.19: 54738523-54738523 on Build GRCh38

View Full Product Details