Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54803800-54804308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIR2DL4
Entrez Gene ID
3805
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 4
Gene Aliases
CD158D, G9P, KIR, KIR-103AS, KIR-2DL4, KIR103, KIR103AS
UniGene
Hs.661219
Gene Chromosome Location
Chr.19: 54803612-54814517 on Build GRCh38
Transcripts
NM_001080770.1 NM_001080772.1 NM_002255.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3539017 c.42C>T p.F14F Chr.19: 54803898-54803898 on Build GRCh38
713623 c.61G>T p.V21L Chr.19: 54803911-54803911 on Build GRCh38
7172255 c.72C>G p.H24Q Chr.19: 54803922-54803922 on Build GRCh38
3539015 c.48C>T p.F16F Chr.19: 54803898-54803898 on Build GRCh38
6695863 c.45C>A p.F15L Chr.19: 54803895-54803895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754017507 Chr.19: 54803853-54803853 on Build GRCh38
rs1018376638 Chr.19: 54803996-54803996 on Build GRCh38
rs778312810 Chr.19: 54804198-54804198 on Build GRCh38
rs574920795 Chr.19: 54804232-54804232 on Build GRCh38
rs964871144 Chr.19: 54804010-54804010 on Build GRCh38
rs597598 Chr.19: 54803985-54803985 on Build GRCh38
rs757711299 Chr.19: 54803879-54803879 on Build GRCh38
rs763154628 Chr.19: 54803941-54803941 on Build GRCh38
rs1038999335 Chr.19: 54803964-54803964 on Build GRCh38
rs772945256 Chr.19: 54803849-54803849 on Build GRCh38
rs561029274 Chr.19: 54804024-54804024 on Build GRCh38
rs748888663 Chr.19: 54803893-54803893 on Build GRCh38
rs776553253 Chr.19: 54803926-54803926 on Build GRCh38
rs779576225 Chr.19: 54803909-54803928 on Build GRCh38
rs1029649370 Chr.19: 54803828-54803828 on Build GRCh38
rs754703177 Chr.19: 54803897-54803897 on Build GRCh38
rs918796971 Chr.19: 54804133-54804133 on Build GRCh38
rs1010118767 Chr.19: 54804251-54804251 on Build GRCh38
rs759856886 Chr.19: 54803857-54803857 on Build GRCh38
rs560915366 Chr.19: 54803821-54803821 on Build GRCh38
rs764638684 Chr.19: 54803948-54803948 on Build GRCh38
rs604076 Chr.19: 54803871-54803871 on Build GRCh38
rs538996766 Chr.19: 54803956-54803956 on Build GRCh38
rs373153736 Chr.19: 54803922-54803922 on Build GRCh38
rs1057170141 Chr.19: 54803895-54803895 on Build GRCh38
rs955937544 Chr.19: 54804019-54804019 on Build GRCh38
rs772050236 Chr.19: 54803842-54803842 on Build GRCh38
rs375766742 Chr.19: 54803902-54803902 on Build GRCh38
rs988760886 Chr.19: 54804062-54804062 on Build GRCh38
rs1038520440 Chr.19: 54804114-54804114 on Build GRCh38
rs34825495 Chr.19: 54804079-54804080 on Build GRCh38
rs1033282024 Chr.19: 54804014-54804014 on Build GRCh38
rs748328234 Chr.19: 54804017-54804017 on Build GRCh38
rs955865266 Chr.19: 54804246-54804246 on Build GRCh38
rs1001396045 Chr.19: 54804212-54804212 on Build GRCh38
rs978604134 Chr.19: 54804012-54804012 on Build GRCh38
rs1051286348 Chr.19: 54804149-54804149 on Build GRCh38
rs200791941 Chr.19: 54804112-54804112 on Build GRCh38
rs778445513 Chr.19: 54803875-54803875 on Build GRCh38
rs765545265 Chr.19: 54803934-54803934 on Build GRCh38
rs941557102 Chr.19: 54803961-54803961 on Build GRCh38
rs757905731 Chr.19: 54803949-54803949 on Build GRCh38
rs891132736 Chr.19: 54803991-54803991 on Build GRCh38
rs1029618227 Chr.19: 54803990-54803990 on Build GRCh38
rs971024134 Chr.19: 54804280-54804280 on Build GRCh38
rs1024246849 Chr.19: 54804267-54804267 on Build GRCh38
rs1032836887 Chr.19: 54804230-54804230 on Build GRCh38
rs375419633 Chr.19: 54803923-54803923 on Build GRCh38
rs758950743 Chr.19: 54803874-54803874 on Build GRCh38
rs541099983 Chr.19: 54803914-54803914 on Build GRCh38
rs1006904339 Chr.19: 54804159-54804159 on Build GRCh38
rs369033322 Chr.19: 54803917-54803917 on Build GRCh38
rs867450023 Chr.19: 54803838-54803838 on Build GRCh38
rs598452 Chr.19: 54804161-54804161 on Build GRCh38
rs754542587 Chr.19: 54803967-54803967 on Build GRCh38
rs760498163 Chr.19: 54803851-54803851 on Build GRCh38
rs897393598 Chr.19: 54803984-54803984 on Build GRCh38
rs369973702 Chr.19: 54803968-54803968 on Build GRCh38
rs765224033 Chr.19: 54803860-54803860 on Build GRCh38
rs752861823 Chr.19: 54803866-54803866 on Build GRCh38
rs949703570 Chr.19: 54804095-54804095 on Build GRCh38
rs371120439 Chr.19: 54804135-54804135 on Build GRCh38
rs980237565 Chr.19: 54804109-54804109 on Build GRCh38
rs1006887844 Chr.19: 54803995-54803995 on Build GRCh38
rs903918210 Chr.19: 54804200-54804200 on Build GRCh38
rs955440443 Chr.19: 54803843-54803843 on Build GRCh38
rs767743322 Chr.19: 54803951-54803951 on Build GRCh38
rs2295802 Chr.19: 54803852-54803852 on Build GRCh38
rs916886227 Chr.19: 54804075-54804075 on Build GRCh38
rs367868728 Chr.19: 54803876-54803876 on Build GRCh38
rs747682034 Chr.19: 54803907-54803907 on Build GRCh38
rs56133375 Chr.19: 54804045-54804046 on Build GRCh38
rs56186508 Chr.19: 54803892-54803892 on Build GRCh38
rs759645440 Chr.19: 54803929-54803929 on Build GRCh38
rs56136619 Chr.19: 54804111-54804111 on Build GRCh38
rs891100233 Chr.19: 54804156-54804156 on Build GRCh38
rs775661067 Chr.19: 54803939-54803939 on Build GRCh38

View Full Product Details