Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50517349-50517851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAPH2
Entrez Gene ID
29781
Gene Name
non-SMC condensin II complex subunit H2
Gene Aliases
CAPH2
UniGene
Hs.730607
Gene Chromosome Location
Chr.22: 50508216-50524780 on Build GRCh38
Transcripts
NM_001185011.1 NM_014551.4 NM_152299.3 XM_005261912.4 XM_011530685.2 XM_017028793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1035492 c.267-2A>G p.? Chr.22: 50517575-50517575 on Build GRCh38
1035494 c.414G>A p.T138T Chr.22: 50517803-50517803 on Build GRCh38
1239557 c.408T>G p.D136E Chr.22: 50517797-50517797 on Build GRCh38
3065921 c.263A>G p.K88R Chr.22: 50517479-50517479 on Build GRCh38
6872041 c.331C>T p.P111S Chr.22: 50517641-50517641 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745977314 Chr.22: 50517456-50517456 on Build GRCh38
rs758751077 Chr.22: 50517563-50517563 on Build GRCh38
rs371595904 Chr.22: 50517710-50517710 on Build GRCh38
rs113288865 Chr.22: 50517662-50517662 on Build GRCh38
rs2269388 Chr.22: 50517713-50517713 on Build GRCh38
rs201118245 Chr.22: 50517808-50517808 on Build GRCh38
rs773949592 Chr.22: 50517493-50517493 on Build GRCh38
rs201673419 Chr.22: 50517745-50517745 on Build GRCh38
rs368250304 Chr.22: 50517614-50517614 on Build GRCh38
rs765630778 Chr.22: 50517694-50517695 on Build GRCh38
rs749111633 Chr.22: 50517534-50517534 on Build GRCh38
rs376989523 Chr.22: 50517722-50517722 on Build GRCh38
rs377723640 Chr.22: 50517447-50517447 on Build GRCh38
rs372044931 Chr.22: 50517635-50517635 on Build GRCh38
rs752216231 Chr.22: 50517501-50517501 on Build GRCh38
rs770278461 Chr.22: 50517545-50517545 on Build GRCh38
rs369000915 Chr.22: 50517714-50517714 on Build GRCh38
rs926698864 Chr.22: 50517773-50517773 on Build GRCh38
rs756149592 Chr.22: 50517719-50517719 on Build GRCh38
rs201599631 Chr.22: 50517642-50517642 on Build GRCh38
rs199689462 Chr.22: 50517597-50517597 on Build GRCh38
rs756618804 Chr.22: 50517424-50517424 on Build GRCh38
rs780923245 Chr.22: 50517737-50517737 on Build GRCh38
rs779301926 Chr.22: 50517519-50517519 on Build GRCh38
rs905140475 Chr.22: 50517378-50517378 on Build GRCh38
rs772231508 Chr.22: 50517459-50517459 on Build GRCh38
rs770819683 Chr.22: 50517585-50517585 on Build GRCh38
rs759759210 Chr.22: 50517771-50517771 on Build GRCh38
rs917909034 Chr.22: 50517744-50517744 on Build GRCh38
rs11704871 Chr.22: 50517730-50517730 on Build GRCh38
rs745695877 Chr.22: 50517653-50517653 on Build GRCh38
rs914981032 Chr.22: 50517762-50517762 on Build GRCh38
rs748498506 Chr.22: 50517779-50517779 on Build GRCh38
rs200126237 Chr.22: 50517688-50517688 on Build GRCh38
rs141239310 Chr.22: 50517522-50517522 on Build GRCh38
rs753094280 Chr.22: 50517422-50517422 on Build GRCh38
rs764200384 Chr.22: 50517797-50517797 on Build GRCh38
rs746768930 Chr.22: 50517729-50517729 on Build GRCh38
rs200663937 Chr.22: 50517687-50517687 on Build GRCh38
rs750201844 Chr.22: 50517399-50517399 on Build GRCh38
rs765827468 Chr.22: 50517615-50517615 on Build GRCh38
rs776252361 Chr.22: 50517609-50517609 on Build GRCh38
rs984950466 Chr.22: 50517716-50517716 on Build GRCh38
rs373870538 Chr.22: 50517577-50517577 on Build GRCh38
rs765737574 Chr.22: 50517560-50517560 on Build GRCh38
rs1136705 Chr.22: 50517443-50517443 on Build GRCh38
rs780264028 Chr.22: 50517567-50517567 on Build GRCh38
rs750830683 Chr.22: 50517619-50517619 on Build GRCh38
rs569448391 Chr.22: 50517803-50517803 on Build GRCh38
rs766538568 Chr.22: 50517626-50517626 on Build GRCh38
rs768040574 Chr.22: 50517420-50517420 on Build GRCh38
rs138318301 Chr.22: 50517802-50517802 on Build GRCh38
rs769379570 Chr.22: 50517746-50517746 on Build GRCh38
rs770426757 Chr.22: 50517690-50517690 on Build GRCh38
rs762750604 Chr.22: 50517486-50517486 on Build GRCh38
rs771631683 Chr.22: 50517591-50517591 on Build GRCh38
rs755243082 Chr.22: 50517752-50517752 on Build GRCh38
rs771854678 Chr.22: 50517658-50517658 on Build GRCh38
rs749518820 Chr.22: 50517376-50517376 on Build GRCh38
rs142921052 Chr.22: 50517749-50517749 on Build GRCh38
rs369402376 Chr.22: 50517691-50517691 on Build GRCh38
rs62239486 Chr.22: 50517554-50517554 on Build GRCh38
rs762390370 Chr.22: 50517557-50517557 on Build GRCh38
rs771184206 Chr.22: 50517382-50517382 on Build GRCh38
rs756641153 Chr.22: 50517505-50517505 on Build GRCh38
rs769765812 Chr.22: 50517471-50517471 on Build GRCh38
rs750814257 Chr.22: 50517562-50517562 on Build GRCh38
rs760261671 Chr.22: 50517512-50517513 on Build GRCh38
rs779966401 Chr.22: 50517727-50517727 on Build GRCh38
rs755043609 Chr.22: 50517578-50517578 on Build GRCh38
rs779692981 Chr.22: 50517675-50517675 on Build GRCh38
rs372646427 Chr.22: 50517589-50517589 on Build GRCh38
rs775617474 Chr.22: 50517466-50517466 on Build GRCh38
rs761692582 Chr.22: 50517390-50517390 on Build GRCh38
rs147027738 Chr.22: 50517772-50517772 on Build GRCh38
rs1036692820 Chr.22: 50517603-50517603 on Build GRCh38
rs199513142 Chr.22: 50517582-50517582 on Build GRCh38
rs754323391 Chr.22: 50517556-50517556 on Build GRCh38
rs772824821 Chr.22: 50517747-50517747 on Build GRCh38
rs747097527 Chr.22: 50517527-50517527 on Build GRCh38
rs760022279 Chr.22: 50517567-50517568 on Build GRCh38
rs749654643 Chr.22: 50517444-50517444 on Build GRCh38
rs753858953 Chr.22: 50517801-50517801 on Build GRCh38
rs200222785 Chr.22: 50517551-50517551 on Build GRCh38
rs373229326 Chr.22: 50517386-50517386 on Build GRCh38
rs140833290 Chr.22: 50517453-50517453 on Build GRCh38
rs991471228 Chr.22: 50517498-50517498 on Build GRCh38
rs965059019 Chr.22: 50517499-50517499 on Build GRCh38
rs377259008 Chr.22: 50517526-50517526 on Build GRCh38
rs374607010 Chr.22: 50517598-50517598 on Build GRCh38
rs768191519 Chr.22: 50517680-50517680 on Build GRCh38
rs1008858456 Chr.22: 50517590-50517590 on Build GRCh38
rs753038408 Chr.22: 50517765-50517772 on Build GRCh38
rs373827183 Chr.22: 50517721-50517721 on Build GRCh38
rs931755087 Chr.22: 50517764-50517764 on Build GRCh38
rs369720270 Chr.22: 50517735-50517735 on Build GRCh38
rs758663307 Chr.22: 50517523-50517523 on Build GRCh38
rs1053586949 Chr.22: 50517780-50517780 on Build GRCh38
rs150150677 Chr.22: 50517631-50517631 on Build GRCh38
rs776198405 Chr.22: 50517416-50517416 on Build GRCh38
rs764608213 Chr.22: 50517510-50517510 on Build GRCh38
rs759222844 Chr.22: 50517550-50517550 on Build GRCh38
rs1050311517 Chr.22: 50517373-50517373 on Build GRCh38
rs760167692 Chr.22: 50517553-50517553 on Build GRCh38
rs995094234 Chr.22: 50517674-50517674 on Build GRCh38
rs778070413 Chr.22: 50517425-50517425 on Build GRCh38
rs1008724072 Chr.22: 50517389-50517389 on Build GRCh38
rs774879640 Chr.22: 50517552-50517552 on Build GRCh38
rs777135387 Chr.22: 50517549-50517550 on Build GRCh38
rs749838289 Chr.22: 50517748-50517748 on Build GRCh38
rs535638620 Chr.22: 50517495-50517495 on Build GRCh38
rs956591417 Chr.22: 50517448-50517448 on Build GRCh38
rs772938145 Chr.22: 50517473-50517473 on Build GRCh38
rs777591184 Chr.22: 50517579-50517579 on Build GRCh38
rs763342233 Chr.22: 50517623-50517623 on Build GRCh38
rs962283792 Chr.22: 50517724-50517724 on Build GRCh38
rs763248571 Chr.22: 50517707-50517707 on Build GRCh38
rs147758819 Chr.22: 50517404-50517404 on Build GRCh38
rs746714891 Chr.22: 50517678-50517678 on Build GRCh38
rs759067436 Chr.22: 50517494-50517494 on Build GRCh38
rs374588731 Chr.22: 50517531-50517531 on Build GRCh38
rs768262618 Chr.22: 50517607-50517607 on Build GRCh38
rs568442647 Chr.22: 50517829-50517829 on Build GRCh38
rs368713408 Chr.22: 50517394-50517394 on Build GRCh38
rs985430239 Chr.22: 50517533-50517533 on Build GRCh38
rs752766206 Chr.22: 50517640-50517640 on Build GRCh38
rs758238985 Chr.22: 50517723-50517723 on Build GRCh38
rs552455534 Chr.22: 50517383-50517383 on Build GRCh38
rs574846073 Chr.22: 50517638-50517638 on Build GRCh38
rs949439055 Chr.22: 50517564-50517564 on Build GRCh38
rs770660298 Chr.22: 50517541-50517541 on Build GRCh38
rs766001548 Chr.22: 50517406-50517406 on Build GRCh38
rs972091586 Chr.22: 50517743-50517743 on Build GRCh38
rs373230081 Chr.22: 50517784-50517784 on Build GRCh38
rs113818181 Chr.22: 50517664-50517664 on Build GRCh38
rs759634125 Chr.22: 50517384-50517384 on Build GRCh38
rs958808611 Chr.22: 50517689-50517689 on Build GRCh38
rs368611781 Chr.22: 50517588-50517588 on Build GRCh38
rs762848166 Chr.22: 50517542-50517542 on Build GRCh38
rs754416891 Chr.22: 50517419-50517419 on Build GRCh38
rs370859039 Chr.22: 50517528-50517528 on Build GRCh38
rs748077061 Chr.22: 50517469-50517469 on Build GRCh38
rs192993579 Chr.22: 50517517-50517517 on Build GRCh38
rs766023747 Chr.22: 50517492-50517492 on Build GRCh38
rs375327095 Chr.22: 50517387-50517387 on Build GRCh38
rs754160402 Chr.22: 50517511-50517511 on Build GRCh38
rs774108172 Chr.22: 50517549-50517549 on Build GRCh38
rs756154977 Chr.22: 50517708-50517708 on Build GRCh38
rs138641341 Chr.22: 50517637-50517637 on Build GRCh38

View Full Product Details