Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 8881187-8881671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD18
Entrez Gene ID
56852
Gene Name
RAD18, E3 ubiquitin protein ligase
Gene Aliases
RNF73
UniGene
Hs.375684
Gene Chromosome Location
Chr.3: 8877196-8969249 on Build GRCh38
Transcripts
NM_020165.3 XM_017006873.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
397252 c.1393C>G p.L465V Chr.3: 8881452-8881452 on Build GRCh38
76154 c.1466G>C p.R489T Chr.3: 8881379-8881379 on Build GRCh38
1425615 c.1430G>A p.R477H Chr.3: 8881415-8881415 on Build GRCh38
1048845 c.1475G>A p.R492H Chr.3: 8881370-8881370 on Build GRCh38
6469128 c.1427G>A p.R476H Chr.3: 8881418-8881418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556401440 Chr.3: 8881414-8881414 on Build GRCh38
rs1025361943 Chr.3: 8881640-8881640 on Build GRCh38
rs371021843 Chr.3: 8881431-8881431 on Build GRCh38
rs1026277588 Chr.3: 8881598-8881598 on Build GRCh38
rs1034951050 Chr.3: 8881214-8881214 on Build GRCh38
rs1041826904 Chr.3: 8881502-8881502 on Build GRCh38
rs746590678 Chr.3: 8881311-8881311 on Build GRCh38
rs374666316 Chr.3: 8881501-8881501 on Build GRCh38
rs1035710120 Chr.3: 8881589-8881589 on Build GRCh38
rs190987093 Chr.3: 8881395-8881395 on Build GRCh38
rs1001558482 Chr.3: 8881585-8881585 on Build GRCh38
rs901091045 Chr.3: 8881239-8881239 on Build GRCh38
rs984690075 Chr.3: 8881622-8881622 on Build GRCh38
rs760941554 Chr.3: 8881485-8881485 on Build GRCh38
rs776845416 Chr.3: 8881507-8881507 on Build GRCh38
rs775482814 Chr.3: 8881462-8881462 on Build GRCh38
rs769587099 Chr.3: 8881350-8881350 on Build GRCh38
rs139019521 Chr.3: 8881377-8881377 on Build GRCh38
rs113306262 Chr.3: 8881241-8881241 on Build GRCh38
rs895086186 Chr.3: 8881560-8881560 on Build GRCh38
rs764501799 Chr.3: 8881396-8881396 on Build GRCh38
rs45438900 Chr.3: 8881240-8881240 on Build GRCh38
rs547433239 Chr.3: 8881596-8881596 on Build GRCh38
rs372609640 Chr.3: 8881313-8881313 on Build GRCh38
rs771268503 Chr.3: 8881406-8881406 on Build GRCh38
rs893600035 Chr.3: 8881274-8881274 on Build GRCh38
rs149913355 Chr.3: 8881415-8881415 on Build GRCh38
rs375666126 Chr.3: 8881314-8881314 on Build GRCh38
rs959268643 Chr.3: 8881215-8881215 on Build GRCh38
rs1056759514 Chr.3: 8881552-8881552 on Build GRCh38
rs754263589 Chr.3: 8881398-8881398 on Build GRCh38
rs755351061 Chr.3: 8881458-8881459 on Build GRCh38
rs779529673 Chr.3: 8881306-8881306 on Build GRCh38
rs760234514 Chr.3: 8881626-8881626 on Build GRCh38
rs747775964 Chr.3: 8881335-8881335 on Build GRCh38
rs771049896 Chr.3: 8881367-8881367 on Build GRCh38
rs560854411 Chr.3: 8881390-8881390 on Build GRCh38
rs774383366 Chr.3: 8881455-8881455 on Build GRCh38
rs373218575 Chr.3: 8881357-8881357 on Build GRCh38
rs545452321 Chr.3: 8881561-8881561 on Build GRCh38
rs781575602 Chr.3: 8881466-8881467 on Build GRCh38
rs369014549 Chr.3: 8881310-8881310 on Build GRCh38
rs140344625 Chr.3: 8881418-8881418 on Build GRCh38
rs772161303 Chr.3: 8881456-8881456 on Build GRCh38
rs566725589 Chr.3: 8881300-8881300 on Build GRCh38
rs1003390737 Chr.3: 8881531-8881531 on Build GRCh38
rs1021744396 Chr.3: 8881564-8881564 on Build GRCh38
rs182227199 Chr.3: 8881571-8881571 on Build GRCh38
rs1035973506 Chr.3: 8881383-8881383 on Build GRCh38
rs902309329 Chr.3: 8881525-8881525 on Build GRCh38
rs370582611 Chr.3: 8881352-8881352 on Build GRCh38
rs759805996 Chr.3: 8881416-8881416 on Build GRCh38
rs1011667329 Chr.3: 8881299-8881299 on Build GRCh38
rs150411306 Chr.3: 8881370-8881370 on Build GRCh38
rs1016618232 Chr.3: 8881304-8881304 on Build GRCh38
rs901123458 Chr.3: 8881446-8881446 on Build GRCh38
rs757759853 Chr.3: 8881404-8881404 on Build GRCh38
rs140490893 Chr.3: 8881450-8881450 on Build GRCh38
rs960260294 Chr.3: 8881386-8881386 on Build GRCh38
rs952949168 Chr.3: 8881620-8881620 on Build GRCh38
rs946152414 Chr.3: 8881438-8881438 on Build GRCh38
rs45521033 Chr.3: 8881405-8881405 on Build GRCh38
rs141454362 Chr.3: 8881419-8881419 on Build GRCh38
rs150991424 Chr.3: 8881435-8881435 on Build GRCh38
rs759632365 Chr.3: 8881371-8881371 on Build GRCh38
rs45519434 Chr.3: 8881624-8881624 on Build GRCh38

View Full Product Details