Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73481619-73482125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFM
Entrez Gene ID
173
Gene Name
afamin
Gene Aliases
ALB2, ALBA, ALF
UniGene
Hs.168718
Gene Chromosome Location
Chr.4: 73481740-73504001 on Build GRCh38
Transcripts
NM_001133.2 XM_017007842.1 XM_017007843.1 XM_017007844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
396565 c.73C>A p.Q25K Chr.4: 73481848-73481848 on Build GRCh38
3605699 c.56C>T p.S19F Chr.4: 73481831-73481831 on Build GRCh38
4896836 c.69C>T p.P23P Chr.4: 73481844-73481844 on Build GRCh38
3605700 c.82G>A p.D28N Chr.4: 73481857-73481857 on Build GRCh38
158950 c.36C>A p.F12L Chr.4: 73481811-73481811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187320979 Chr.4: 73481760-73481760 on Build GRCh38
rs769430447 Chr.4: 73481838-73481838 on Build GRCh38
rs763039992 Chr.4: 73481846-73481846 on Build GRCh38
rs62619161 Chr.4: 73481761-73481761 on Build GRCh38
rs765368945 Chr.4: 73481881-73481881 on Build GRCh38
rs769414093 Chr.4: 73481902-73481902 on Build GRCh38
rs940134437 Chr.4: 73481681-73481681 on Build GRCh38
rs372725061 Chr.4: 73481728-73481728 on Build GRCh38
rs1041494347 Chr.4: 73481687-73481687 on Build GRCh38
rs973938741 Chr.4: 73482018-73482018 on Build GRCh38
rs1028083500 Chr.4: 73481673-73481673 on Build GRCh38
rs745316212 Chr.4: 73481837-73481837 on Build GRCh38
rs375207234 Chr.4: 73481990-73481990 on Build GRCh38
rs766573879 Chr.4: 73481856-73481856 on Build GRCh38
rs1024461701 Chr.4: 73482093-73482093 on Build GRCh38
rs565892696 Chr.4: 73481967-73481967 on Build GRCh38
rs776445618 Chr.4: 73481823-73481823 on Build GRCh38
rs779335886 Chr.4: 73481793-73481793 on Build GRCh38
rs973987885 Chr.4: 73481781-73481781 on Build GRCh38
rs758982346 Chr.4: 73481885-73481885 on Build GRCh38
rs939445772 Chr.4: 73482101-73482101 on Build GRCh38
rs961051445 Chr.4: 73482009-73482009 on Build GRCh38
rs753880154 Chr.4: 73481791-73481791 on Build GRCh38
rs373320234 Chr.4: 73481985-73481985 on Build GRCh38
rs1015200450 Chr.4: 73481997-73481997 on Build GRCh38
rs980872536 Chr.4: 73481829-73481829 on Build GRCh38
rs376388409 Chr.4: 73481871-73481871 on Build GRCh38
rs140254262 Chr.4: 73481806-73481806 on Build GRCh38
rs145217978 Chr.4: 73481815-73481815 on Build GRCh38
rs1014869266 Chr.4: 73482077-73482077 on Build GRCh38
rs568499671 Chr.4: 73481889-73481889 on Build GRCh38
rs370247366 Chr.4: 73481896-73481896 on Build GRCh38
rs374579053 Chr.4: 73481851-73481851 on Build GRCh38
rs766341746 Chr.4: 73481780-73481780 on Build GRCh38
rs1028448642 Chr.4: 73481976-73481976 on Build GRCh38
rs762766274 Chr.4: 73481755-73481755 on Build GRCh38
rs561835861 Chr.4: 73481814-73481815 on Build GRCh38
rs901178235 Chr.4: 73481690-73481690 on Build GRCh38
rs745580956 Chr.4: 73481899-73481899 on Build GRCh38
rs755072696 Chr.4: 73481792-73481792 on Build GRCh38
rs955178461 Chr.4: 73481721-73481721 on Build GRCh38
rs75652679 Chr.4: 73481855-73481855 on Build GRCh38
rs778247086 Chr.4: 73481810-73481810 on Build GRCh38
rs374692435 Chr.4: 73481718-73481718 on Build GRCh38
rs775561120 Chr.4: 73481843-73481843 on Build GRCh38
rs753179052 Chr.4: 73481883-73481883 on Build GRCh38
rs1000425949 Chr.4: 73481731-73481731 on Build GRCh38
rs970904346 Chr.4: 73482097-73482097 on Build GRCh38
rs997031070 Chr.4: 73481916-73481916 on Build GRCh38
rs746487864 Chr.4: 73481815-73481815 on Build GRCh38
rs531634253 Chr.4: 73481854-73481854 on Build GRCh38
rs747348985 Chr.4: 73481893-73481893 on Build GRCh38
rs746609442 Chr.4: 73481812-73481812 on Build GRCh38
rs561667995 Chr.4: 73481686-73481686 on Build GRCh38
rs948525560 Chr.4: 73482088-73482088 on Build GRCh38
rs74826873 Chr.4: 73481723-73481723 on Build GRCh38
rs3765639 Chr.4: 73482054-73482054 on Build GRCh38
rs986547516 Chr.4: 73481988-73481988 on Build GRCh38
rs748660845 Chr.4: 73481802-73481802 on Build GRCh38
rs780844079 Chr.4: 73481881-73481897 on Build GRCh38
rs987910726 Chr.4: 73481771-73481771 on Build GRCh38
rs539759071 Chr.4: 73482020-73482020 on Build GRCh38
rs781406073 Chr.4: 73481909-73481909 on Build GRCh38
rs553949260 Chr.4: 73481924-73481924 on Build GRCh38
rs748191653 Chr.4: 73481911-73481911 on Build GRCh38
rs922712379 Chr.4: 73481897-73481897 on Build GRCh38
rs1053323642 Chr.4: 73482053-73482053 on Build GRCh38
rs1000456895 Chr.4: 73481934-73481934 on Build GRCh38
rs1048241663 Chr.4: 73481647-73481647 on Build GRCh38
rs754073576 Chr.4: 73481864-73481864 on Build GRCh38
rs975542810 Chr.4: 73481659-73481659 on Build GRCh38
rs886921309 Chr.4: 73481658-73481658 on Build GRCh38

View Full Product Details