Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30647212-30647733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf136
Entrez Gene ID
221545
Gene Name
chromosome 6 open reading frame 136
Gene Aliases
-
UniGene
Hs.591787
Gene Chromosome Location
Chr.6: 30647039-30653210 on Build GRCh38
Transcripts
NM_001109938.2 NM_001161376.1 NM_145029.3 XM_011514384.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3994803 c.97A>G p.R33G Chr.6: 30647328-30647328 on Build GRCh38
6397669 c.254G>A p.C85Y Chr.6: 30647485-30647485 on Build GRCh38
6387542 c.268G>A p.A90T Chr.6: 30647499-30647499 on Build GRCh38
7317183 c.398G>A p.R133Q Chr.6: 30647629-30647629 on Build GRCh38
5732503 c.176G>A p.R59K Chr.6: 30647407-30647407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925765586 Chr.6: 30647453-30647453 on Build GRCh38
rs28360037 Chr.6: 30647467-30647467 on Build GRCh38
rs753431554 Chr.6: 30647710-30647710 on Build GRCh38
rs747516318 Chr.6: 30647374-30647374 on Build GRCh38
rs975077908 Chr.6: 30647480-30647480 on Build GRCh38
rs767778593 Chr.6: 30647308-30647308 on Build GRCh38
rs776329801 Chr.6: 30647709-30647709 on Build GRCh38
rs888985195 Chr.6: 30647418-30647418 on Build GRCh38
rs995231343 Chr.6: 30647532-30647532 on Build GRCh38
rs894495085 Chr.6: 30647565-30647565 on Build GRCh38
rs116816200 Chr.6: 30647507-30647507 on Build GRCh38
rs925576876 Chr.6: 30647310-30647312 on Build GRCh38
rs535688451 Chr.6: 30647429-30647429 on Build GRCh38
rs555516210 Chr.6: 30647433-30647433 on Build GRCh38
rs746283081 Chr.6: 30647292-30647292 on Build GRCh38
rs754746788 Chr.6: 30647267-30647267 on Build GRCh38
rs372110822 Chr.6: 30647393-30647393 on Build GRCh38
rs190816727 Chr.6: 30647591-30647591 on Build GRCh38
rs763976351 Chr.6: 30647694-30647694 on Build GRCh38
rs765219539 Chr.6: 30647268-30647268 on Build GRCh38
rs780504031 Chr.6: 30647356-30647356 on Build GRCh38
rs769163774 Chr.6: 30647364-30647364 on Build GRCh38
rs970224908 Chr.6: 30647492-30647492 on Build GRCh38
rs770269177 Chr.6: 30647293-30647293 on Build GRCh38
rs916120554 Chr.6: 30647323-30647323 on Build GRCh38
rs61606469 Chr.6: 30647485-30647485 on Build GRCh38
rs186287743 Chr.6: 30647580-30647580 on Build GRCh38
rs890590022 Chr.6: 30647506-30647506 on Build GRCh38
rs775316855 Chr.6: 30647417-30647417 on Build GRCh38
rs764140056 Chr.6: 30647421-30647421 on Build GRCh38
rs555692995 Chr.6: 30647642-30647642 on Build GRCh38
rs933129123 Chr.6: 30647397-30647397 on Build GRCh38
rs748320980 Chr.6: 30647375-30647375 on Build GRCh38
rs1041483084 Chr.6: 30647282-30647282 on Build GRCh38
rs888870611 Chr.6: 30647255-30647255 on Build GRCh38
rs916987542 Chr.6: 30647248-30647248 on Build GRCh38
rs762033832 Chr.6: 30647245-30647245 on Build GRCh38
rs780959695 Chr.6: 30647289-30647289 on Build GRCh38
rs1051979589 Chr.6: 30647497-30647497 on Build GRCh38
rs762861913 Chr.6: 30647588-30647588 on Build GRCh38
rs920900355 Chr.6: 30647679-30647679 on Build GRCh38
rs760229333 Chr.6: 30647676-30647676 on Build GRCh38
rs948800287 Chr.6: 30647325-30647325 on Build GRCh38
rs755776732 Chr.6: 30647441-30647441 on Build GRCh38
rs925959262 Chr.6: 30647343-30647343 on Build GRCh38
rs762903321 Chr.6: 30647555-30647555 on Build GRCh38
rs749943087 Chr.6: 30647275-30647275 on Build GRCh38
rs559519679 Chr.6: 30647249-30647249 on Build GRCh38
rs977537787 Chr.6: 30647449-30647449 on Build GRCh38
rs967615543 Chr.6: 30647610-30647610 on Build GRCh38
rs777280036 Chr.6: 30647281-30647281 on Build GRCh38
rs745509405 Chr.6: 30647339-30647339 on Build GRCh38
rs3132594 Chr.6: 30647483-30647483 on Build GRCh38
rs756548635 Chr.6: 30647352-30647352 on Build GRCh38
rs563563696 Chr.6: 30647567-30647567 on Build GRCh38
rs899121199 Chr.6: 30647413-30647414 on Build GRCh38
rs1015016209 Chr.6: 30647407-30647407 on Build GRCh38
rs769601702 Chr.6: 30647416-30647416 on Build GRCh38
rs761770831 Chr.6: 30647303-30647303 on Build GRCh38
rs746664675 Chr.6: 30647283-30647283 on Build GRCh38
rs762622744 Chr.6: 30647420-30647420 on Build GRCh38
rs749732291 Chr.6: 30647296-30647296 on Build GRCh38
rs926043544 Chr.6: 30647508-30647508 on Build GRCh38
rs758309114 Chr.6: 30647320-30647320 on Build GRCh38
rs528191201 Chr.6: 30647313-30647313 on Build GRCh38
rs1034372439 Chr.6: 30647538-30647538 on Build GRCh38
rs757959659 Chr.6: 30647310-30647310 on Build GRCh38
rs1026530312 Chr.6: 30647465-30647465 on Build GRCh38
rs774349849 Chr.6: 30647603-30647603 on Build GRCh38
rs773413897 Chr.6: 30647312-30647312 on Build GRCh38
rs201743957 Chr.6: 30647321-30647321 on Build GRCh38
rs754282301 Chr.6: 30647442-30647442 on Build GRCh38
rs766179823 Chr.6: 30647246-30647246 on Build GRCh38
rs749910816 Chr.6: 30647661-30647661 on Build GRCh38
rs146616682 Chr.6: 30647636-30647636 on Build GRCh38
rs757852832 Chr.6: 30647277-30647277 on Build GRCh38
rs1051593990 Chr.6: 30647399-30647399 on Build GRCh38
rs749778757 Chr.6: 30647362-30647362 on Build GRCh38
rs773509215 Chr.6: 30647520-30647520 on Build GRCh38
rs747264656 Chr.6: 30647521-30647521 on Build GRCh38
rs1015928212 Chr.6: 30647270-30647270 on Build GRCh38
rs1047843311 Chr.6: 30647688-30647688 on Build GRCh38
rs774701606 Chr.6: 30647371-30647371 on Build GRCh38
rs933215098 Chr.6: 30647587-30647587 on Build GRCh38
rs539750251 Chr.6: 30647380-30647380 on Build GRCh38
rs766117132 Chr.6: 30647687-30647687 on Build GRCh38
rs866892600 Chr.6: 30647541-30647541 on Build GRCh38
rs975151664 Chr.6: 30647660-30647660 on Build GRCh38
rs747822653 Chr.6: 30647456-30647457 on Build GRCh38
rs771514519 Chr.6: 30647559-30647560 on Build GRCh38
rs115572115 Chr.6: 30647436-30647436 on Build GRCh38
rs751120241 Chr.6: 30647333-30647333 on Build GRCh38
rs977317274 Chr.6: 30647633-30647633 on Build GRCh38
rs900798451 Chr.6: 30647327-30647327 on Build GRCh38
rs764946486 Chr.6: 30647316-30647316 on Build GRCh38
rs912185613 Chr.6: 30647712-30647712 on Build GRCh38
rs899279402 Chr.6: 30647531-30647531 on Build GRCh38
rs755287603 Chr.6: 30647445-30647445 on Build GRCh38
rs550423101 Chr.6: 30647381-30647381 on Build GRCh38
rs551438712 Chr.6: 30647319-30647319 on Build GRCh38
rs932212946 Chr.6: 30647488-30647488 on Build GRCh38
rs756804188 Chr.6: 30647338-30647338 on Build GRCh38
rs753677862 Chr.6: 30647260-30647260 on Build GRCh38
rs774437722 Chr.6: 30647300-30647300 on Build GRCh38
rs778465489 Chr.6: 30647254-30647254 on Build GRCh38

View Full Product Details