Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30647518-30648024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf136
Entrez Gene ID
221545
Gene Name
chromosome 6 open reading frame 136
Gene Aliases
-
UniGene
Hs.591787
Gene Chromosome Location
Chr.6: 30647039-30653210 on Build GRCh38
Transcripts
NM_001109938.2 NM_001161376.1 NM_145029.3 XM_011514384.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3948966 c.553G>T p.G185C Chr.6: 30647784-30647784 on Build GRCh38
6351267 c.595A>G p.R199G Chr.6: 30647826-30647826 on Build GRCh38
7317183 c.398G>A p.R133Q Chr.6: 30647629-30647629 on Build GRCh38
4551139 c.519G>T p.R173R Chr.6: 30647750-30647750 on Build GRCh38
6426801 c.610A>G p.T204A Chr.6: 30647841-30647841 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996518292 Chr.6: 30647915-30647915 on Build GRCh38
rs563563696 Chr.6: 30647567-30647567 on Build GRCh38
rs753431554 Chr.6: 30647710-30647710 on Build GRCh38
rs988944707 Chr.6: 30647908-30647908 on Build GRCh38
rs143403912 Chr.6: 30647948-30647948 on Build GRCh38
rs549203712 Chr.6: 30647951-30647951 on Build GRCh38
rs761504261 Chr.6: 30647792-30647792 on Build GRCh38
rs920654909 Chr.6: 30647739-30647739 on Build GRCh38
rs776329801 Chr.6: 30647709-30647709 on Build GRCh38
rs117903822 Chr.6: 30647826-30647826 on Build GRCh38
rs56196719 Chr.6: 30647976-30647976 on Build GRCh38
rs894495085 Chr.6: 30647565-30647565 on Build GRCh38
rs551411222 Chr.6: 30647822-30647822 on Build GRCh38
rs755378720 Chr.6: 30647740-30647740 on Build GRCh38
rs930688706 Chr.6: 30647780-30647780 on Build GRCh38
rs779090449 Chr.6: 30647811-30647811 on Build GRCh38
rs528205837 Chr.6: 30647799-30647799 on Build GRCh38
rs1036560474 Chr.6: 30647734-30647734 on Build GRCh38
rs1034372439 Chr.6: 30647538-30647538 on Build GRCh38
rs190816727 Chr.6: 30647591-30647591 on Build GRCh38
rs774349849 Chr.6: 30647603-30647603 on Build GRCh38
rs763976351 Chr.6: 30647694-30647694 on Build GRCh38
rs550293871 Chr.6: 30647911-30647911 on Build GRCh38
rs749910816 Chr.6: 30647661-30647661 on Build GRCh38
rs146616682 Chr.6: 30647636-30647636 on Build GRCh38
rs530493615 Chr.6: 30647905-30647905 on Build GRCh38
rs758865017 Chr.6: 30647836-30647836 on Build GRCh38
rs186287743 Chr.6: 30647580-30647580 on Build GRCh38
rs112155635 Chr.6: 30647945-30647945 on Build GRCh38
rs555692995 Chr.6: 30647642-30647642 on Build GRCh38
rs956316808 Chr.6: 30647904-30647904 on Build GRCh38
rs1047843311 Chr.6: 30647688-30647688 on Build GRCh38
rs940844625 Chr.6: 30647715-30647715 on Build GRCh38
rs933215098 Chr.6: 30647587-30647587 on Build GRCh38
rs766117132 Chr.6: 30647687-30647687 on Build GRCh38
rs866892600 Chr.6: 30647541-30647541 on Build GRCh38
rs975151664 Chr.6: 30647660-30647660 on Build GRCh38
rs1033058471 Chr.6: 30647837-30647837 on Build GRCh38
rs771514519 Chr.6: 30647559-30647560 on Build GRCh38
rs762861913 Chr.6: 30647588-30647588 on Build GRCh38
rs977317274 Chr.6: 30647633-30647633 on Build GRCh38
rs920900355 Chr.6: 30647679-30647679 on Build GRCh38
rs760229333 Chr.6: 30647676-30647676 on Build GRCh38
rs777154560 Chr.6: 30647892-30647893 on Build GRCh38
rs999077186 Chr.6: 30647891-30647891 on Build GRCh38
rs912185613 Chr.6: 30647712-30647712 on Build GRCh38
rs903130740 Chr.6: 30647861-30647861 on Build GRCh38
rs762903321 Chr.6: 30647555-30647555 on Build GRCh38
rs967615543 Chr.6: 30647610-30647610 on Build GRCh38
rs943146332 Chr.6: 30647977-30647977 on Build GRCh38

View Full Product Details