Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30650978-30651493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf136
Entrez Gene ID
221545
Gene Name
chromosome 6 open reading frame 136
Gene Aliases
-
UniGene
Hs.591787
Gene Chromosome Location
Chr.6: 30647039-30653210 on Build GRCh38
Transcripts
NM_001109938.2 NM_001161376.1 NM_145029.3 XM_011514384.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6616410 c.1100G>A p.R367H Chr.6: 30651076-30651076 on Build GRCh38
741466 c.238G>A p.E80K Chr.6: 30651369-30651369 on Build GRCh38
3684192 c.209T>G p.L70W Chr.6: 30651340-30651340 on Build GRCh38
6616407 c.1178G>A p.R393H Chr.6: 30651337-30651337 on Build GRCh38
3777443 c.306C>T p.F102F Chr.6: 30651437-30651437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956460940 Chr.6: 30651359-30651359 on Build GRCh38
rs750964129 Chr.6: 30651470-30651471 on Build GRCh38
rs1003672122 Chr.6: 30651398-30651398 on Build GRCh38
rs983087227 Chr.6: 30651013-30651013 on Build GRCh38
rs773589330 Chr.6: 30651006-30651006 on Build GRCh38
rs1040782014 Chr.6: 30651200-30651200 on Build GRCh38
rs185110317 Chr.6: 30651141-30651141 on Build GRCh38
rs35201959 Chr.6: 30651368-30651368 on Build GRCh38
rs753798518 Chr.6: 30651323-30651323 on Build GRCh38
rs773122395 Chr.6: 30651179-30651179 on Build GRCh38
rs780402499 Chr.6: 30651423-30651423 on Build GRCh38
rs777283810 Chr.6: 30651229-30651229 on Build GRCh38
rs774539217 Chr.6: 30651288-30651288 on Build GRCh38
rs1005435842 Chr.6: 30651174-30651174 on Build GRCh38
rs770525226 Chr.6: 30651274-30651274 on Build GRCh38
rs554320988 Chr.6: 30651002-30651002 on Build GRCh38
rs759230424 Chr.6: 30651457-30651457 on Build GRCh38
rs200976150 Chr.6: 30651115-30651115 on Build GRCh38
rs967690045 Chr.6: 30651433-30651433 on Build GRCh38
rs747351662 Chr.6: 30651020-30651020 on Build GRCh38
rs771419771 Chr.6: 30651221-30651221 on Build GRCh38
rs9262135 Chr.6: 30651129-30651129 on Build GRCh38
rs760527127 Chr.6: 30651297-30651297 on Build GRCh38
rs144380927 Chr.6: 30651445-30651445 on Build GRCh38
rs145374052 Chr.6: 30651449-30651449 on Build GRCh38
rs970890655 Chr.6: 30651030-30651030 on Build GRCh38
rs536061367 Chr.6: 30651135-30651135 on Build GRCh38
rs769784783 Chr.6: 30651235-30651235 on Build GRCh38
rs1047035121 Chr.6: 30651165-30651165 on Build GRCh38
rs970837847 Chr.6: 30651390-30651390 on Build GRCh38
rs372723087 Chr.6: 30651103-30651103 on Build GRCh38
rs150233869 Chr.6: 30651360-30651360 on Build GRCh38
rs764422167 Chr.6: 30651281-30651281 on Build GRCh38
rs145813113 Chr.6: 30651444-30651444 on Build GRCh38
rs762911105 Chr.6: 30651271-30651271 on Build GRCh38
rs994557013 Chr.6: 30651149-30651153 on Build GRCh38
rs373040690 Chr.6: 30651270-30651270 on Build GRCh38
rs770369001 Chr.6: 30651414-30651414 on Build GRCh38
rs141051300 Chr.6: 30651189-30651189 on Build GRCh38
rs888147200 Chr.6: 30651337-30651337 on Build GRCh38
rs751193071 Chr.6: 30651395-30651395 on Build GRCh38
rs376546503 Chr.6: 30651341-30651341 on Build GRCh38
rs11557269 Chr.6: 30651429-30651429 on Build GRCh38
rs746405414 Chr.6: 30651413-30651413 on Build GRCh38
rs771615967 Chr.6: 30651021-30651021 on Build GRCh38
rs765292120 Chr.6: 30651075-30651075 on Build GRCh38
rs377317868 Chr.6: 30651109-30651109 on Build GRCh38
rs372840949 Chr.6: 30651096-30651096 on Build GRCh38
rs777552786 Chr.6: 30651391-30651391 on Build GRCh38
rs200795261 Chr.6: 30651345-30651345 on Build GRCh38
rs753768068 Chr.6: 30651106-30651106 on Build GRCh38
rs765281708 Chr.6: 30651466-30651466 on Build GRCh38
rs747790395 Chr.6: 30651111-30651111 on Build GRCh38
rs369215354 Chr.6: 30651455-30651455 on Build GRCh38
rs757707277 Chr.6: 30651397-30651397 on Build GRCh38
rs796658763 Chr.6: 30651425-30651426 on Build GRCh38
rs767883008 Chr.6: 30651374-30651390 on Build GRCh38
rs901284905 Chr.6: 30651146-30651146 on Build GRCh38
rs781646958 Chr.6: 30651409-30651409 on Build GRCh38
rs554711801 Chr.6: 30651277-30651277 on Build GRCh38
rs766093375 Chr.6: 30651304-30651304 on Build GRCh38
rs1041681239 Chr.6: 30651169-30651169 on Build GRCh38
rs763042784 Chr.6: 30651266-30651266 on Build GRCh38
rs79679868 Chr.6: 30651090-30651090 on Build GRCh38
rs775598877 Chr.6: 30651250-30651250 on Build GRCh38
rs776450954 Chr.6: 30651456-30651456 on Build GRCh38
rs199806173 Chr.6: 30651392-30651392 on Build GRCh38
rs989885801 Chr.6: 30651117-30651117 on Build GRCh38
rs747437491 Chr.6: 30651216-30651216 on Build GRCh38
rs777265395 Chr.6: 30651024-30651024 on Build GRCh38
rs575874165 Chr.6: 30651232-30651232 on Build GRCh38
rs376377195 Chr.6: 30651076-30651076 on Build GRCh38
rs1033848329 Chr.6: 30651402-30651402 on Build GRCh38
rs759758080 Chr.6: 30651066-30651066 on Build GRCh38
rs959713855 Chr.6: 30651442-30651442 on Build GRCh38
rs749442787 Chr.6: 30651432-30651432 on Build GRCh38
rs757836760 Chr.6: 30651371-30651371 on Build GRCh38

View Full Product Details