Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 86643834-86644342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRM3
Entrez Gene ID
2913
Gene Name
glutamate metabotropic receptor 3
Gene Aliases
GLUR3, GPRC1C, MGLUR3, mGlu3
UniGene
Hs.590575
Gene Chromosome Location
Chr.7: 86643914-86864879 on Build GRCh38
Transcripts
NM_000840.2

SNP Detail

SNP ID Chromosome Location
rs971417784 Chr.7: 86643902-86643902 on Build GRCh38
rs1027553933 Chr.7: 86644248-86644248 on Build GRCh38
rs375161182 Chr.7: 86644137-86644137 on Build GRCh38
rs2073549 Chr.7: 86644268-86644268 on Build GRCh38
rs952000223 Chr.7: 86644249-86644249 on Build GRCh38
rs180801475 Chr.7: 86644064-86644064 on Build GRCh38
rs944410382 Chr.7: 86644038-86644038 on Build GRCh38
rs550719496 Chr.7: 86643905-86643905 on Build GRCh38
rs900763969 Chr.7: 86644063-86644063 on Build GRCh38
rs570732360 Chr.7: 86644090-86644090 on Build GRCh38
rs761431139 Chr.7: 86644148-86644148 on Build GRCh38
rs184863976 Chr.7: 86644169-86644169 on Build GRCh38
rs1040011101 Chr.7: 86644057-86644057 on Build GRCh38
rs372048007 Chr.7: 86643893-86643893 on Build GRCh38
rs1055406311 Chr.7: 86644176-86644176 on Build GRCh38
rs986062575 Chr.7: 86643932-86643932 on Build GRCh38
rs116123427 Chr.7: 86644088-86644088 on Build GRCh38
rs575849181 Chr.7: 86644189-86644189 on Build GRCh38
rs1020824097 Chr.7: 86644162-86644162 on Build GRCh38
rs573127772 Chr.7: 86644287-86644287 on Build GRCh38
rs930036252 Chr.7: 86644016-86644017 on Build GRCh38
rs1041019379 Chr.7: 86644006-86644006 on Build GRCh38
rs374298065 Chr.7: 86644072-86644072 on Build GRCh38
rs866666505 Chr.7: 86644123-86644123 on Build GRCh38
rs907286018 Chr.7: 86644175-86644175 on Build GRCh38
rs537901542 Chr.7: 86644164-86644164 on Build GRCh38
rs925419985 Chr.7: 86643906-86643906 on Build GRCh38
rs959289583 Chr.7: 86644226-86644226 on Build GRCh38
rs865825047 Chr.7: 86643986-86643986 on Build GRCh38
rs1024608336 Chr.7: 86644260-86644260 on Build GRCh38
rs959237734 Chr.7: 86643921-86643921 on Build GRCh38
rs574732392 Chr.7: 86644184-86644184 on Build GRCh38
rs1034317005 Chr.7: 86644213-86644213 on Build GRCh38
rs944418562 Chr.7: 86644298-86644298 on Build GRCh38
rs532213574 Chr.7: 86643865-86643865 on Build GRCh38
rs773840099 Chr.7: 86644092-86644092 on Build GRCh38
rs149971121 Chr.7: 86643938-86643938 on Build GRCh38
rs748045932 Chr.7: 86643907-86643907 on Build GRCh38
rs1002877469 Chr.7: 86644188-86644188 on Build GRCh38
rs911457548 Chr.7: 86643937-86643937 on Build GRCh38
rs930427850 Chr.7: 86643967-86643967 on Build GRCh38
rs553776431 Chr.7: 86643872-86643872 on Build GRCh38
rs546529739 Chr.7: 86644275-86644275 on Build GRCh38
rs535890340 Chr.7: 86644245-86644245 on Build GRCh38
rs189234667 Chr.7: 86644273-86644273 on Build GRCh38
rs562300942 Chr.7: 86643916-86643916 on Build GRCh38
rs1020351458 Chr.7: 86643860-86643860 on Build GRCh38
rs184681725 Chr.7: 86643951-86643951 on Build GRCh38
rs377698820 Chr.7: 86643926-86643926 on Build GRCh38
rs189201696 Chr.7: 86643993-86643993 on Build GRCh38
rs1013516408 Chr.7: 86644258-86644258 on Build GRCh38
rs893320153 Chr.7: 86644136-86644136 on Build GRCh38
rs991122240 Chr.7: 86644247-86644247 on Build GRCh38
rs772727267 Chr.7: 86643985-86643985 on Build GRCh38
rs748904658 Chr.7: 86644010-86644010 on Build GRCh38

View Full Product Details