Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 134933584-134934098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALD1
Entrez Gene ID
800
Gene Name
caldesmon 1
Gene Aliases
CDM, H-CAD, HCAD, L-CAD, LCAD, NAG22
UniGene
Hs.490203
Gene Chromosome Location
Chr.7: 134779413-134970729 on Build GRCh38
Transcripts
NM_004342.6 NM_033138.3 NM_033139.3 NM_033140.3 NM_033157.3 XM_017012650.1 XM_017012651.1 XM_017012652.1 XM_017012653.1 XM_017012654.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
347343 c.1167C>A p.N389K Chr.7: 134933936-134933936 on Build GRCh38
5011861 c.861G>C p.K287N Chr.7: 134933630-134933630 on Build GRCh38
5043194 c.1216G>T p.G406W Chr.7: 134933985-134933985 on Build GRCh38
5789227 c.1158G>A p.Q386Q Chr.7: 134933927-134933927 on Build GRCh38
7413685 c.902C>T p.A301V Chr.7: 134933671-134933671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942817766 Chr.7: 134933765-134933765 on Build GRCh38
rs201196898 Chr.7: 134933986-134933986 on Build GRCh38
rs969210249 Chr.7: 134933924-134933924 on Build GRCh38
rs145193752 Chr.7: 134933892-134933892 on Build GRCh38
rs754746969 Chr.7: 134933644-134933644 on Build GRCh38
rs762289688 Chr.7: 134934050-134934050 on Build GRCh38
rs201179218 Chr.7: 134933931-134933931 on Build GRCh38
rs61757650 Chr.7: 134933745-134933745 on Build GRCh38
rs930361640 Chr.7: 134933772-134933772 on Build GRCh38
rs769014173 Chr.7: 134934042-134934042 on Build GRCh38
rs776165758 Chr.7: 134933696-134933696 on Build GRCh38
rs901392081 Chr.7: 134933807-134933807 on Build GRCh38
rs1043254908 Chr.7: 134933750-134933750 on Build GRCh38
rs117593246 Chr.7: 134933720-134933720 on Build GRCh38
rs202174796 Chr.7: 134933895-134933895 on Build GRCh38
rs764898963 Chr.7: 134933886-134933886 on Build GRCh38
rs756787426 Chr.7: 134933999-134933999 on Build GRCh38
rs147935316 Chr.7: 134933806-134933806 on Build GRCh38
rs765090265 Chr.7: 134933966-134933966 on Build GRCh38
rs769398416 Chr.7: 134934021-134934021 on Build GRCh38
rs780829907 Chr.7: 134933909-134933909 on Build GRCh38
rs774921696 Chr.7: 134934046-134934046 on Build GRCh38
rs771697836 Chr.7: 134933657-134933657 on Build GRCh38
rs777704344 Chr.7: 134933985-134933985 on Build GRCh38
rs773207941 Chr.7: 134934059-134934059 on Build GRCh38
rs147336538 Chr.7: 134933646-134933646 on Build GRCh38
rs749743284 Chr.7: 134933691-134933693 on Build GRCh38
rs765589889 Chr.7: 134933717-134933717 on Build GRCh38
rs769104192 Chr.7: 134933949-134933949 on Build GRCh38
rs758018045 Chr.7: 134933984-134933984 on Build GRCh38
rs200864409 Chr.7: 134933991-134933991 on Build GRCh38
rs772171717 Chr.7: 134934058-134934058 on Build GRCh38
rs748165535 Chr.7: 134933653-134933653 on Build GRCh38
rs867395131 Chr.7: 134933664-134933665 on Build GRCh38
rs772894421 Chr.7: 134933660-134933660 on Build GRCh38
rs775880471 Chr.7: 134933932-134933932 on Build GRCh38
rs749597641 Chr.7: 134934026-134934026 on Build GRCh38
rs140903770 Chr.7: 134933740-134933740 on Build GRCh38
rs1045221791 Chr.7: 134933794-134933794 on Build GRCh38
rs780623139 Chr.7: 134934007-134934007 on Build GRCh38
rs187878127 Chr.7: 134933922-134933922 on Build GRCh38
rs376074647 Chr.7: 134934063-134934063 on Build GRCh38
rs781302656 Chr.7: 134933632-134933632 on Build GRCh38
rs773202828 Chr.7: 134933647-134933647 on Build GRCh38
rs201912094 Chr.7: 134934018-134934018 on Build GRCh38
rs772183351 Chr.7: 134933985-134933985 on Build GRCh38
rs529198474 Chr.7: 134933849-134933849 on Build GRCh38
rs750647687 Chr.7: 134933964-134933964 on Build GRCh38
rs142006920 Chr.7: 134933643-134933643 on Build GRCh38
rs982005668 Chr.7: 134933703-134933703 on Build GRCh38
rs752498924 Chr.7: 134933972-134933972 on Build GRCh38
rs200467553 Chr.7: 134933962-134933962 on Build GRCh38
rs1023689150 Chr.7: 134933894-134933894 on Build GRCh38
rs969359460 Chr.7: 134933674-134933674 on Build GRCh38
rs763340502 Chr.7: 134933947-134933947 on Build GRCh38
rs372481805 Chr.7: 134933758-134933758 on Build GRCh38
rs750618653 Chr.7: 134933775-134933775 on Build GRCh38
rs139286937 Chr.7: 134933827-134933827 on Build GRCh38
rs751716254 Chr.7: 134933618-134933618 on Build GRCh38
rs773546022 Chr.7: 134934072-134934072 on Build GRCh38
rs778911869 Chr.7: 134933863-134933863 on Build GRCh38
rs193221622 Chr.7: 134933874-134933874 on Build GRCh38
rs748566152 Chr.7: 134933952-134933952 on Build GRCh38
rs6973420 Chr.7: 134933959-134933959 on Build GRCh38
rs199592221 Chr.7: 134934005-134934005 on Build GRCh38
rs770573328 Chr.7: 134933676-134933676 on Build GRCh38
rs770357937 Chr.7: 134933906-134933906 on Build GRCh38
rs760594441 Chr.7: 134933695-134933695 on Build GRCh38
rs764129164 Chr.7: 134933738-134933738 on Build GRCh38
rs915031406 Chr.7: 134933743-134933743 on Build GRCh38
rs994203795 Chr.7: 134933848-134933848 on Build GRCh38
rs996912966 Chr.7: 134933990-134933990 on Build GRCh38
rs117939016 Chr.7: 134933616-134933616 on Build GRCh38
rs149149797 Chr.7: 134933957-134933957 on Build GRCh38
rs751456111 Chr.7: 134933988-134933988 on Build GRCh38
rs774135873 Chr.7: 134933624-134933624 on Build GRCh38
rs573221724 Chr.7: 134933723-134933725 on Build GRCh38
rs201623086 Chr.7: 134933973-134933973 on Build GRCh38
rs562542239 Chr.7: 134933615-134933615 on Build GRCh38
rs185503672 Chr.7: 134933878-134933878 on Build GRCh38
rs763507772 Chr.7: 134933727-134933727 on Build GRCh38
rs758285908 Chr.7: 134933980-134933980 on Build GRCh38
rs746607454 Chr.7: 134933661-134933661 on Build GRCh38
rs759887507 Chr.7: 134933715-134933715 on Build GRCh38
rs761031070 Chr.7: 134933965-134933965 on Build GRCh38
rs148403238 Chr.7: 134933628-134933628 on Build GRCh38
rs939502622 Chr.7: 134933869-134933869 on Build GRCh38
rs779400483 Chr.7: 134933848-134933849 on Build GRCh38

View Full Product Details