Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38511795-38512293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf86
Entrez Gene ID
389649
Gene Name
chromosome 8 open reading frame 86
Gene Aliases
-
UniGene
Hs.546586
Gene Chromosome Location
Chr.8: 38510834-38528662 on Build GRCh38
Transcripts
NM_001303531.1 NM_207412.2

SNP Detail

SNP ID Chromosome Location
rs892666787 Chr.8: 38512014-38512014 on Build GRCh38
rs1041246526 Chr.8: 38512019-38512019 on Build GRCh38
rs570769396 Chr.8: 38512186-38512186 on Build GRCh38
rs992675072 Chr.8: 38512110-38512110 on Build GRCh38
rs868057281 Chr.8: 38512202-38512202 on Build GRCh38
rs765487113 Chr.8: 38511944-38511944 on Build GRCh38
rs191592923 Chr.8: 38511919-38511919 on Build GRCh38
rs947142609 Chr.8: 38512082-38512082 on Build GRCh38
rs1041507281 Chr.8: 38512127-38512127 on Build GRCh38
rs937197126 Chr.8: 38511836-38511837 on Build GRCh38
rs984828710 Chr.8: 38511954-38511954 on Build GRCh38
rs1026637293 Chr.8: 38512064-38512064 on Build GRCh38
rs996826201 Chr.8: 38512181-38512181 on Build GRCh38
rs183894870 Chr.8: 38512001-38512001 on Build GRCh38
rs34919622 Chr.8: 38511844-38511845 on Build GRCh38
rs948005283 Chr.8: 38511915-38511915 on Build GRCh38
rs969985254 Chr.8: 38512130-38512130 on Build GRCh38
rs574451570 Chr.8: 38511891-38511891 on Build GRCh38
rs902135327 Chr.8: 38512174-38512174 on Build GRCh38
rs576314985 Chr.8: 38511927-38511927 on Build GRCh38
rs185986139 Chr.8: 38511907-38511907 on Build GRCh38
rs141300714 Chr.8: 38512168-38512168 on Build GRCh38
rs535919686 Chr.8: 38511837-38511837 on Build GRCh38
rs563970307 Chr.8: 38512087-38512087 on Build GRCh38
rs532611572 Chr.8: 38512015-38512015 on Build GRCh38
rs1015348369 Chr.8: 38512105-38512105 on Build GRCh38
rs369939830 Chr.8: 38512131-38512131 on Build GRCh38
rs1006315405 Chr.8: 38512241-38512241 on Build GRCh38
rs925337304 Chr.8: 38511966-38511966 on Build GRCh38
rs372802096 Chr.8: 38511832-38511832 on Build GRCh38
rs774511956 Chr.8: 38511870-38511871 on Build GRCh38
rs892923984 Chr.8: 38511839-38511839 on Build GRCh38
rs79687910 Chr.8: 38512053-38512053 on Build GRCh38
rs79297228 Chr.8: 38512199-38512199 on Build GRCh38
rs952165927 Chr.8: 38511951-38511951 on Build GRCh38
rs925308406 Chr.8: 38512145-38512145 on Build GRCh38
rs548545342 Chr.8: 38512071-38512071 on Build GRCh38
rs915840290 Chr.8: 38511871-38511871 on Build GRCh38
rs952196773 Chr.8: 38512218-38512218 on Build GRCh38
rs1007876484 Chr.8: 38511912-38511912 on Build GRCh38
rs187150382 Chr.8: 38512113-38512113 on Build GRCh38
rs1011321504 Chr.8: 38511848-38511848 on Build GRCh38
rs907940122 Chr.8: 38511971-38511971 on Build GRCh38
rs559767380 Chr.8: 38512043-38512043 on Build GRCh38
rs980751932 Chr.8: 38512139-38512139 on Build GRCh38
rs962061997 Chr.8: 38512270-38512270 on Build GRCh38
rs541266306 Chr.8: 38512029-38512029 on Build GRCh38
rs181330764 Chr.8: 38511832-38511832 on Build GRCh38
rs879114547 Chr.8: 38511947-38511947 on Build GRCh38
rs6474384 Chr.8: 38511975-38511975 on Build GRCh38
rs534698094 Chr.8: 38511852-38511852 on Build GRCh38
rs960110837 Chr.8: 38512109-38512109 on Build GRCh38
rs959984252 Chr.8: 38511853-38511853 on Build GRCh38
rs192494628 Chr.8: 38512266-38512266 on Build GRCh38
rs996362352 Chr.8: 38511941-38511942 on Build GRCh38
rs543914417 Chr.8: 38511870-38511870 on Build GRCh38

View Full Product Details