Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 63185956-63186455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YTHDF3
Entrez Gene ID
253943
Gene Name
YTH N6-methyladenosine RNA binding protein 3
Gene Aliases
-
UniGene
Hs.491861
Gene Chromosome Location
Chr.8: 63168553-63212788 on Build GRCh38
Transcripts
NM_001277813.1 NM_001277814.1 NM_001277815.1 NM_001277816.1 NM_001277817.1 NM_001277818.1 NM_152758.5 NR_102434.1 XM_011517509.2 XM_011517510.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2720458 c.408G>A p.G136G Chr.8: 63186422-63186422 on Build GRCh38
2720456 c.324T>A p.P108P Chr.8: 63186338-63186338 on Build GRCh38
2720455 c.312A>T p.V104V Chr.8: 63186326-63186326 on Build GRCh38
5657264 c.307G>C p.G103R Chr.8: 63186321-63186321 on Build GRCh38
2720453 c.294C>T p.H98H Chr.8: 63186308-63186308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755517093 Chr.8: 63186407-63186407 on Build GRCh38
rs539063421 Chr.8: 63186066-63186066 on Build GRCh38
rs769939625 Chr.8: 63186315-63186315 on Build GRCh38
rs773653096 Chr.8: 63186323-63186323 on Build GRCh38
rs763456577 Chr.8: 63186342-63186342 on Build GRCh38
rs766744297 Chr.8: 63186128-63186128 on Build GRCh38
rs780958698 Chr.8: 63186164-63186164 on Build GRCh38
rs765632293 Chr.8: 63186118-63186118 on Build GRCh38
rs200588648 Chr.8: 63186380-63186380 on Build GRCh38
rs768737431 Chr.8: 63186236-63186236 on Build GRCh38
rs200374147 Chr.8: 63186168-63186168 on Build GRCh38
rs774686493 Chr.8: 63186374-63186374 on Build GRCh38
rs189950017 Chr.8: 63186117-63186117 on Build GRCh38
rs761751224 Chr.8: 63186153-63186153 on Build GRCh38
rs377236178 Chr.8: 63186356-63186356 on Build GRCh38
rs778846283 Chr.8: 63186224-63186224 on Build GRCh38
rs199632488 Chr.8: 63186099-63186103 on Build GRCh38
rs745713607 Chr.8: 63186230-63186230 on Build GRCh38
rs898576001 Chr.8: 63186082-63186082 on Build GRCh38
rs376064187 Chr.8: 63186101-63186101 on Build GRCh38
rs750991919 Chr.8: 63186101-63186101 on Build GRCh38
rs942808347 Chr.8: 63186070-63186070 on Build GRCh38
rs752412447 Chr.8: 63186396-63186396 on Build GRCh38
rs750961936 Chr.8: 63186121-63186121 on Build GRCh38
rs139728132 Chr.8: 63186172-63186172 on Build GRCh38
rs761390134 Chr.8: 63186108-63186108 on Build GRCh38
rs182939076 Chr.8: 63186134-63186134 on Build GRCh38
rs755677765 Chr.8: 63186187-63186187 on Build GRCh38
rs749247122 Chr.8: 63186204-63186204 on Build GRCh38
rs1014099388 Chr.8: 63186312-63186312 on Build GRCh38
rs776558864 Chr.8: 63186105-63186105 on Build GRCh38
rs748138153 Chr.8: 63186261-63186261 on Build GRCh38
rs752129585 Chr.8: 63186170-63186170 on Build GRCh38
rs776951090 Chr.8: 63186253-63186253 on Build GRCh38
rs540311577 Chr.8: 63186123-63186123 on Build GRCh38
rs767050181 Chr.8: 63186389-63186389 on Build GRCh38
rs922509910 Chr.8: 63186079-63186079 on Build GRCh38
rs768989915 Chr.8: 63186100-63186100 on Build GRCh38
rs777802604 Chr.8: 63186195-63186195 on Build GRCh38
rs267601964 Chr.8: 63186410-63186410 on Build GRCh38
rs767019758 Chr.8: 63186169-63186170 on Build GRCh38
rs753463627 Chr.8: 63186427-63186427 on Build GRCh38
rs771521146 Chr.8: 63186336-63186336 on Build GRCh38
rs1037783524 Chr.8: 63186337-63186337 on Build GRCh38
rs33994594 Chr.8: 63186172-63186173 on Build GRCh38
rs1038820109 Chr.8: 63186074-63186074 on Build GRCh38
rs757043254 Chr.8: 63186222-63186222 on Build GRCh38
rs557729395 Chr.8: 63186084-63186084 on Build GRCh38
rs763699606 Chr.8: 63186426-63186426 on Build GRCh38

View Full Product Details