Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22474834-22475381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP3CC
Entrez Gene ID
5533
Gene Name
protein phosphatase 3 catalytic subunit gamma
Gene Aliases
CALNA3, CNA3, PP2Bgamma
UniGene
Hs.731683
Gene Chromosome Location
Chr.8: 22440970-22541466 on Build GRCh38
Transcripts
NM_001243974.1 NM_001243975.1 NM_005605.4 XM_017013610.1 XM_017013611.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7146289 c.222G>C p.M74I Chr.8: 22475126-22475126 on Build GRCh38
5725424 c.57C>A p.P19P Chr.8: 22474961-22474961 on Build GRCh38
6431152 c.52G>T p.V18F Chr.8: 22474956-22474956 on Build GRCh38
3735318 c.55C>T p.P19S Chr.8: 22474959-22474959 on Build GRCh38
1623857 c.185A>G p.N62S Chr.8: 22475089-22475089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763694932 Chr.8: 22475133-22475133 on Build GRCh38
rs750914252 Chr.8: 22474985-22474985 on Build GRCh38
rs149167807 Chr.8: 22474887-22474887 on Build GRCh38
rs745800385 Chr.8: 22475038-22475038 on Build GRCh38
rs886758169 Chr.8: 22475184-22475184 on Build GRCh38
rs753384106 Chr.8: 22475025-22475025 on Build GRCh38
rs775717455 Chr.8: 22474943-22474943 on Build GRCh38
rs965525087 Chr.8: 22474864-22474864 on Build GRCh38
rs975542366 Chr.8: 22475330-22475330 on Build GRCh38
rs761936868 Chr.8: 22475071-22475071 on Build GRCh38
rs191180753 Chr.8: 22475006-22475006 on Build GRCh38
rs756620884 Chr.8: 22474942-22474942 on Build GRCh38
rs530288872 Chr.8: 22475024-22475024 on Build GRCh38
rs779165958 Chr.8: 22475169-22475171 on Build GRCh38
rs756788817 Chr.8: 22475026-22475026 on Build GRCh38
rs1046898085 Chr.8: 22475276-22475276 on Build GRCh38
rs774623986 Chr.8: 22475120-22475120 on Build GRCh38
rs763316735 Chr.8: 22474990-22474990 on Build GRCh38
rs766657020 Chr.8: 22474993-22474993 on Build GRCh38
rs747232518 Chr.8: 22475060-22475060 on Build GRCh38
rs749206014 Chr.8: 22474911-22474911 on Build GRCh38
rs146104346 Chr.8: 22474959-22474959 on Build GRCh38
rs975593246 Chr.8: 22474868-22474868 on Build GRCh38
rs550388793 Chr.8: 22475354-22475354 on Build GRCh38
rs138834215 Chr.8: 22474978-22474978 on Build GRCh38
rs200358456 Chr.8: 22475124-22475124 on Build GRCh38
rs577458190 Chr.8: 22474940-22474940 on Build GRCh38
rs763437404 Chr.8: 22475114-22475114 on Build GRCh38
rs201270011 Chr.8: 22475061-22475061 on Build GRCh38
rs770899497 Chr.8: 22474914-22474914 on Build GRCh38
rs1019837738 Chr.8: 22475300-22475300 on Build GRCh38
rs769040257 Chr.8: 22475069-22475069 on Build GRCh38
rs371119915 Chr.8: 22474977-22474977 on Build GRCh38
rs750932634 Chr.8: 22474930-22474932 on Build GRCh38
rs764797394 Chr.8: 22475159-22475159 on Build GRCh38
rs753508772 Chr.8: 22475139-22475139 on Build GRCh38
rs894194265 Chr.8: 22475097-22475097 on Build GRCh38
rs943962345 Chr.8: 22475305-22475305 on Build GRCh38
rs1004313333 Chr.8: 22475267-22475267 on Build GRCh38
rs538908474 Chr.8: 22475331-22475331 on Build GRCh38
rs780373923 Chr.8: 22474942-22474943 on Build GRCh38
rs755776313 Chr.8: 22475020-22475020 on Build GRCh38
rs891005136 Chr.8: 22475293-22475295 on Build GRCh38
rs772512470 Chr.8: 22475052-22475052 on Build GRCh38
rs201924888 Chr.8: 22475095-22475095 on Build GRCh38
rs778646320 Chr.8: 22474922-22474922 on Build GRCh38
rs899489776 Chr.8: 22475316-22475316 on Build GRCh38
rs1028506160 Chr.8: 22475345-22475345 on Build GRCh38
rs771780603 Chr.8: 22474942-22474942 on Build GRCh38
rs548956647 Chr.8: 22475220-22475220 on Build GRCh38
rs776901296 Chr.8: 22474960-22474960 on Build GRCh38
rs766751572 Chr.8: 22475116-22475116 on Build GRCh38
rs773699670 Chr.8: 22475075-22475075 on Build GRCh38
rs929778412 Chr.8: 22475242-22475242 on Build GRCh38
rs771899958 Chr.8: 22475055-22475055 on Build GRCh38
rs770006457 Chr.8: 22475073-22475073 on Build GRCh38
rs201721168 Chr.8: 22474958-22474958 on Build GRCh38
rs372948151 Chr.8: 22475169-22475169 on Build GRCh38
rs745633313 Chr.8: 22474934-22474934 on Build GRCh38
rs754208437 Chr.8: 22474943-22474944 on Build GRCh38
rs756693360 Chr.8: 22475140-22475140 on Build GRCh38
rs965638818 Chr.8: 22475315-22475315 on Build GRCh38
rs571490912 Chr.8: 22475164-22475164 on Build GRCh38
rs777026325 Chr.8: 22475070-22475070 on Build GRCh38
rs146307937 Chr.8: 22475031-22475031 on Build GRCh38
rs924164228 Chr.8: 22474888-22474888 on Build GRCh38
rs560532505 Chr.8: 22475058-22475058 on Build GRCh38

View Full Product Details