Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 5742614-5743112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIC1
Entrez Gene ID
57589
Gene Name
RIC1 homolog, RAB6A GEF complex partner 1
Gene Aliases
CIP150, KIAA1432, bA207C16.1
UniGene
Hs.211520
Gene Chromosome Location
Chr.9: 5629030-5778633 on Build GRCh38
Transcripts
NM_001135920.2 NM_001206557.1 NM_020829.3 XM_005251523.3 XM_011517967.2 XM_017014934.1 XM_017014935.1 XM_017014936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4942514 c.674A>G p.N225S Chr.9: 5742878-5742878 on Build GRCh38
1109389 c.778C>T p.Q260* Chr.9: 5742982-5742982 on Build GRCh38
6479975 c.760T>C p.W254R Chr.9: 5742964-5742964 on Build GRCh38
4790935 c.737G>A p.W246* Chr.9: 5742941-5742941 on Build GRCh38
4917889 c.769T>G p.F257V Chr.9: 5742973-5742973 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370606357 Chr.9: 5743014-5743014 on Build GRCh38
rs879332737 Chr.9: 5742747-5742747 on Build GRCh38
rs892495531 Chr.9: 5742939-5742939 on Build GRCh38
rs35503630 Chr.9: 5742808-5742809 on Build GRCh38
rs535353303 Chr.9: 5742829-5742829 on Build GRCh38
rs371214803 Chr.9: 5742955-5742955 on Build GRCh38
rs75782093 Chr.9: 5742888-5742888 on Build GRCh38
rs981527275 Chr.9: 5742701-5742701 on Build GRCh38
rs774030559 Chr.9: 5743021-5743021 on Build GRCh38
rs746431741 Chr.9: 5742983-5742983 on Build GRCh38
rs201838674 Chr.9: 5742813-5742814 on Build GRCh38
rs927352471 Chr.9: 5742713-5742713 on Build GRCh38
rs575007227 Chr.9: 5742931-5742931 on Build GRCh38
rs376555854 Chr.9: 5742862-5742862 on Build GRCh38
rs200790844 Chr.9: 5742807-5742808 on Build GRCh38
rs79136867 Chr.9: 5742800-5742800 on Build GRCh38
rs141009627 Chr.9: 5742943-5742943 on Build GRCh38
rs887818148 Chr.9: 5742774-5742774 on Build GRCh38
rs750355833 Chr.9: 5743033-5743033 on Build GRCh38
rs1002339493 Chr.9: 5743083-5743083 on Build GRCh38
rs984896567 Chr.9: 5742979-5742979 on Build GRCh38
rs369400985 Chr.9: 5742865-5742865 on Build GRCh38
rs767069100 Chr.9: 5743032-5743032 on Build GRCh38
rs937471687 Chr.9: 5742744-5742744 on Build GRCh38
rs1036343826 Chr.9: 5743065-5743065 on Build GRCh38
rs556003300 Chr.9: 5742809-5742809 on Build GRCh38
rs770342311 Chr.9: 5743005-5743005 on Build GRCh38
rs913371498 Chr.9: 5742685-5742685 on Build GRCh38
rs386414372 Chr.9: 5742811-5742812 on Build GRCh38
rs371652597 Chr.9: 5742834-5742834 on Build GRCh38
rs1036921897 Chr.9: 5742781-5742781 on Build GRCh38
rs775662535 Chr.9: 5743013-5743013 on Build GRCh38
rs763824450 Chr.9: 5742846-5742846 on Build GRCh38
rs373331915 Chr.9: 5742798-5742798 on Build GRCh38
rs897063218 Chr.9: 5743066-5743066 on Build GRCh38
rs377726801 Chr.9: 5742670-5742670 on Build GRCh38
rs376955309 Chr.9: 5742834-5742838 on Build GRCh38
rs771766663 Chr.9: 5743054-5743055 on Build GRCh38
rs753525911 Chr.9: 5743052-5743052 on Build GRCh38
rs892721786 Chr.9: 5742710-5742711 on Build GRCh38
rs537095027 Chr.9: 5742710-5742710 on Build GRCh38
rs147003898 Chr.9: 5743048-5743048 on Build GRCh38
rs1051366505 Chr.9: 5743076-5743076 on Build GRCh38
rs750405313 Chr.9: 5742864-5742864 on Build GRCh38
rs575987421 Chr.9: 5743041-5743041 on Build GRCh38
rs376579729 Chr.9: 5742814-5742814 on Build GRCh38
rs779779119 Chr.9: 5742870-5742870 on Build GRCh38
rs754844177 Chr.9: 5743055-5743055 on Build GRCh38
rs899822547 Chr.9: 5742690-5742690 on Build GRCh38
rs77839915 Chr.9: 5742681-5742681 on Build GRCh38
rs747055659 Chr.9: 5742937-5742937 on Build GRCh38
rs762449314 Chr.9: 5742835-5742835 on Build GRCh38
rs777318476 Chr.9: 5742831-5742831 on Build GRCh38
rs374540954 Chr.9: 5742798-5742799 on Build GRCh38
rs768255159 Chr.9: 5743019-5743019 on Build GRCh38
rs1040692114 Chr.9: 5742680-5742692 on Build GRCh38
rs758491007 Chr.9: 5742920-5742920 on Build GRCh38
rs747824017 Chr.9: 5742851-5742854 on Build GRCh38
rs760462978 Chr.9: 5743038-5743038 on Build GRCh38
rs751018737 Chr.9: 5742847-5742847 on Build GRCh38
rs759957938 Chr.9: 5742832-5742832 on Build GRCh38
rs868259967 Chr.9: 5743082-5743082 on Build GRCh38
rs1010116162 Chr.9: 5742724-5742724 on Build GRCh38
rs756789059 Chr.9: 5742849-5742849 on Build GRCh38
rs1049524076 Chr.9: 5742765-5742765 on Build GRCh38
rs372487231 Chr.9: 5742779-5742779 on Build GRCh38
rs755526732 Chr.9: 5743074-5743074 on Build GRCh38
rs933946286 Chr.9: 5742866-5742866 on Build GRCh38
rs1033417823 Chr.9: 5742805-5742806 on Build GRCh38
rs761285532 Chr.9: 5743022-5743022 on Build GRCh38
rs950212536 Chr.9: 5742694-5742694 on Build GRCh38
rs771526425 Chr.9: 5742823-5742823 on Build GRCh38
rs574420190 Chr.9: 5742811-5742811 on Build GRCh38
rs1048886629 Chr.9: 5742657-5742657 on Build GRCh38
rs13293052 Chr.9: 5742807-5742807 on Build GRCh38
rs991855721 Chr.9: 5742810-5742811 on Build GRCh38
rs747110437 Chr.9: 5742821-5742821 on Build GRCh38
rs202171928 Chr.9: 5742836-5742836 on Build GRCh38
rs375672479 Chr.9: 5742948-5742948 on Build GRCh38
rs1051760857 Chr.9: 5742702-5742702 on Build GRCh38
rs564066951 Chr.9: 5742923-5742923 on Build GRCh38
rs150243054 Chr.9: 5742999-5742999 on Build GRCh38
rs773357149 Chr.9: 5742818-5742818 on Build GRCh38

View Full Product Details