Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 107306288-107306797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD23B
Entrez Gene ID
5887
Gene Name
RAD23 homolog B, nucleotide excision repair protein
Gene Aliases
HHR23B, HR23B, P58
UniGene
Hs.521640
Gene Chromosome Location
Chr.9: 107283236-107332194 on Build GRCh38
Transcripts
NM_001244713.1 NM_001244724.1 NM_002874.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4836899 c.448G>A p.E150K Chr.9: 107306598-107306598 on Build GRCh38
6761859 c.273A>T p.S91S Chr.9: 107306423-107306423 on Build GRCh38
4853535 c.325C>T p.Q109* Chr.9: 107306475-107306475 on Build GRCh38
6761861 c.229-1G>A p.? Chr.9: 107306378-107306378 on Build GRCh38
5063489 c.91C>T p.P31S Chr.9: 107306632-107306632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs56175121 Chr.9: 107306412-107306412 on Build GRCh38
rs777629375 Chr.9: 107306532-107306532 on Build GRCh38
rs749832557 Chr.9: 107306577-107306577 on Build GRCh38
rs759152448 Chr.9: 107306515-107306515 on Build GRCh38
rs768746785 Chr.9: 107306480-107306480 on Build GRCh38
rs765015572 Chr.9: 107306694-107306694 on Build GRCh38
rs780308925 Chr.9: 107306466-107306466 on Build GRCh38
rs767087752 Chr.9: 107306591-107306591 on Build GRCh38
rs980661250 Chr.9: 107306703-107306703 on Build GRCh38
rs774956476 Chr.9: 107306656-107306656 on Build GRCh38
rs927462331 Chr.9: 107306724-107306724 on Build GRCh38
rs753846095 Chr.9: 107306698-107306698 on Build GRCh38
rs556221498 Chr.9: 107306744-107306744 on Build GRCh38
rs917424064 Chr.9: 107306496-107306496 on Build GRCh38
rs772480235 Chr.9: 107306668-107306668 on Build GRCh38
rs372889132 Chr.9: 107306658-107306658 on Build GRCh38
rs765239059 Chr.9: 107306616-107306616 on Build GRCh38
rs575309149 Chr.9: 107306748-107306748 on Build GRCh38
rs768675864 Chr.9: 107306353-107306353 on Build GRCh38
rs7041239 Chr.9: 107306564-107306564 on Build GRCh38
rs1018103264 Chr.9: 107306312-107306312 on Build GRCh38
rs778040734 Chr.9: 107306650-107306650 on Build GRCh38
rs55815226 Chr.9: 107306506-107306506 on Build GRCh38
rs759969461 Chr.9: 107306582-107306582 on Build GRCh38
rs376087685 Chr.9: 107306614-107306614 on Build GRCh38
rs753831088 Chr.9: 107306536-107306536 on Build GRCh38
rs775578733 Chr.9: 107306346-107306346 on Build GRCh38
rs750765314 Chr.9: 107306376-107306376 on Build GRCh38
rs762445938 Chr.9: 107306368-107306368 on Build GRCh38
rs950956301 Chr.9: 107306481-107306481 on Build GRCh38
rs571421275 Chr.9: 107306720-107306720 on Build GRCh38
rs141800980 Chr.9: 107306572-107306572 on Build GRCh38
rs754501431 Chr.9: 107306422-107306422 on Build GRCh38
rs765992287 Chr.9: 107306395-107306395 on Build GRCh38
rs747257394 Chr.9: 107306469-107306469 on Build GRCh38
rs751188425 Chr.9: 107306411-107306411 on Build GRCh38
rs986016973 Chr.9: 107306538-107306538 on Build GRCh38
rs548841376 Chr.9: 107306563-107306563 on Build GRCh38
rs756262610 Chr.9: 107306639-107306639 on Build GRCh38
rs79639067 Chr.9: 107306454-107306454 on Build GRCh38
rs760749331 Chr.9: 107306350-107306350 on Build GRCh38
rs565492473 Chr.9: 107306444-107306444 on Build GRCh38
rs772728674 Chr.9: 107306647-107306648 on Build GRCh38
rs749014621 Chr.9: 107306458-107306458 on Build GRCh38
rs755623960 Chr.9: 107306523-107306523 on Build GRCh38
rs748650072 Chr.9: 107306351-107306352 on Build GRCh38
rs763010277 Chr.9: 107306488-107306488 on Build GRCh38
rs773862253 Chr.9: 107306497-107306497 on Build GRCh38
rs777717481 Chr.9: 107306456-107306456 on Build GRCh38
rs760355490 Chr.9: 107306672-107306672 on Build GRCh38
rs763322693 Chr.9: 107306501-107306501 on Build GRCh38
rs758219436 Chr.9: 107306619-107306619 on Build GRCh38
rs770156480 Chr.9: 107306549-107306549 on Build GRCh38
rs148029511 Chr.9: 107306543-107306543 on Build GRCh38
rs1005958347 Chr.9: 107306733-107306733 on Build GRCh38
rs760163367 Chr.9: 107306601-107306601 on Build GRCh38
rs146029019 Chr.9: 107306603-107306603 on Build GRCh38
rs961937045 Chr.9: 107306499-107306499 on Build GRCh38
rs748443238 Chr.9: 107306635-107306635 on Build GRCh38
rs200509488 Chr.9: 107306542-107306542 on Build GRCh38
rs11573671 Chr.9: 107306685-107306685 on Build GRCh38
rs775135855 Chr.9: 107306595-107306595 on Build GRCh38
rs751220555 Chr.9: 107306514-107306514 on Build GRCh38
rs766993636 Chr.9: 107306519-107306519 on Build GRCh38
rs750142843 Chr.9: 107306617-107306617 on Build GRCh38
rs761416401 Chr.9: 107306689-107306689 on Build GRCh38
rs112046427 Chr.9: 107306485-107306486 on Build GRCh38
rs773845845 Chr.9: 107306568-107306568 on Build GRCh38
rs1005174559 Chr.9: 107306691-107306691 on Build GRCh38
rs771296451 Chr.9: 107306655-107306655 on Build GRCh38
rs780493891 Chr.9: 107306729-107306729 on Build GRCh38
rs775115586 Chr.9: 107306670-107306670 on Build GRCh38
rs1013096503 Chr.9: 107306326-107306326 on Build GRCh38
rs777269357 Chr.9: 107306366-107306366 on Build GRCh38
rs111972101 Chr.9: 107306521-107306521 on Build GRCh38
rs181765396 Chr.9: 107306742-107306742 on Build GRCh38
rs781509755 Chr.9: 107306631-107306631 on Build GRCh38
rs994902514 Chr.9: 107306677-107306677 on Build GRCh38
rs374011602 Chr.9: 107306580-107306580 on Build GRCh38
rs775954863 Chr.9: 107306688-107306688 on Build GRCh38
rs376878974 Chr.9: 107306389-107306389 on Build GRCh38
rs185036337 Chr.9: 107306374-107306374 on Build GRCh38
rs532474641 Chr.9: 107306459-107306459 on Build GRCh38
rs747228381 Chr.9: 107306545-107306545 on Build GRCh38
rs752600989 Chr.9: 107306441-107306441 on Build GRCh38
rs189146540 Chr.9: 107306737-107306737 on Build GRCh38
rs1039142546 Chr.9: 107306632-107306632 on Build GRCh38
rs749783361 Chr.9: 107306652-107306652 on Build GRCh38
rs1805333 Chr.9: 107306486-107306486 on Build GRCh38
rs780635159 Chr.9: 107306437-107306437 on Build GRCh38
rs776897206 Chr.9: 107306485-107306485 on Build GRCh38
rs138257027 Chr.9: 107306610-107306610 on Build GRCh38

View Full Product Details