Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 52796056-52796554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPANXN5
Entrez Gene ID
494197
Gene Name
SPANX family member N5
Gene Aliases
CT11.10, SPANX-N5
UniGene
Hs.535218
Gene Chromosome Location
Chr.X: 52796146-52797348 on Build GRCh38
Transcripts
NM_001009616.3

SNP Detail

SNP ID Chromosome Location
rs782147198 Chr.X: 52796180-52796180 on Build GRCh38
rs2806837 Chr.X: 52796431-52796431 on Build GRCh38
rs147823065 Chr.X: 52796376-52796376 on Build GRCh38
rs782584169 Chr.X: 52796503-52796503 on Build GRCh38
rs999440542 Chr.X: 52796151-52796151 on Build GRCh38
rs184293048 Chr.X: 52796424-52796424 on Build GRCh38
rs927649742 Chr.X: 52796421-52796421 on Build GRCh38
rs144412302 Chr.X: 52796156-52796156 on Build GRCh38
rs146656736 Chr.X: 52796511-52796511 on Build GRCh38
rs376429162 Chr.X: 52796461-52796461 on Build GRCh38
rs782280868 Chr.X: 52796181-52796181 on Build GRCh38
rs781990962 Chr.X: 52796248-52796248 on Build GRCh38
rs180885319 Chr.X: 52796227-52796227 on Build GRCh38
rs985538599 Chr.X: 52796213-52796213 on Build GRCh38
rs781967949 Chr.X: 52796391-52796391 on Build GRCh38
rs974178646 Chr.X: 52796291-52796291 on Build GRCh38
rs782047839 Chr.X: 52796392-52796392 on Build GRCh38
rs955382114 Chr.X: 52796164-52796164 on Build GRCh38
rs2806838 Chr.X: 52796506-52796506 on Build GRCh38
rs782133567 Chr.X: 52796323-52796323 on Build GRCh38
rs781853183 Chr.X: 52796494-52796494 on Build GRCh38
rs190443975 Chr.X: 52796105-52796105 on Build GRCh38
rs781970491 Chr.X: 52796406-52796406 on Build GRCh38
rs368340156 Chr.X: 52796413-52796413 on Build GRCh38
rs113926707 Chr.X: 52796484-52796484 on Build GRCh38
rs781818364 Chr.X: 52796439-52796439 on Build GRCh38
rs894855984 Chr.X: 52796519-52796519 on Build GRCh38
rs369362252 Chr.X: 52796498-52796498 on Build GRCh38
rs781910625 Chr.X: 52796474-52796474 on Build GRCh38
rs782074085 Chr.X: 52796451-52796451 on Build GRCh38
rs782661200 Chr.X: 52796483-52796483 on Build GRCh38
rs201378339 Chr.X: 52796452-52796452 on Build GRCh38
rs981700706 Chr.X: 52796412-52796412 on Build GRCh38
rs902392684 Chr.X: 52796138-52796138 on Build GRCh38
rs782533840 Chr.X: 52796475-52796475 on Build GRCh38

View Full Product Details