Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 55221234-55221767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAGE5
Entrez Gene ID
90737
Gene Name
PAGE family member 5
Gene Aliases
CT16, CT16.1, CT16.2, GAGEE1, PAGE-5
UniGene
Hs.668491
Gene Chromosome Location
Chr.X: 55220347-55224108 on Build GRCh38
Transcripts
NM_001013435.2 NM_130467.4

SNP Detail

SNP ID Chromosome Location
rs369755348 Chr.X: 55221342-55221342 on Build GRCh38
rs780915198 Chr.X: 55221745-55221745 on Build GRCh38
rs756798579 Chr.X: 55221482-55221482 on Build GRCh38
rs780062677 Chr.X: 55221661-55221661 on Build GRCh38
rs763010133 Chr.X: 55221394-55221394 on Build GRCh38
rs766485668 Chr.X: 55221397-55221397 on Build GRCh38
rs763720499 Chr.X: 55221442-55221442 on Build GRCh38
rs374453105 Chr.X: 55221453-55221453 on Build GRCh38
rs745338734 Chr.X: 55221490-55221490 on Build GRCh38
rs917968776 Chr.X: 55221549-55221549 on Build GRCh38
rs191772467 Chr.X: 55221647-55221647 on Build GRCh38
rs988649553 Chr.X: 55221529-55221529 on Build GRCh38
rs905490818 Chr.X: 55221653-55221653 on Build GRCh38
rs771565221 Chr.X: 55221287-55221287 on Build GRCh38
rs749538972 Chr.X: 55221739-55221739 on Build GRCh38
rs759888255 Chr.X: 55221321-55221321 on Build GRCh38
rs747562777 Chr.X: 55221328-55221328 on Build GRCh38
rs377562249 Chr.X: 55221727-55221727 on Build GRCh38
rs755959250 Chr.X: 55221433-55221433 on Build GRCh38
rs775816390 Chr.X: 55221474-55221474 on Build GRCh38
rs751503035 Chr.X: 55221682-55221682 on Build GRCh38
rs751538007 Chr.X: 55221406-55221406 on Build GRCh38
rs1017556955 Chr.X: 55221391-55221391 on Build GRCh38
rs764106215 Chr.X: 55221655-55221655 on Build GRCh38
rs752586951 Chr.X: 55221432-55221432 on Build GRCh38
rs769099894 Chr.X: 55221334-55221334 on Build GRCh38
rs767817133 Chr.X: 55221563-55221564 on Build GRCh38
rs750331128 Chr.X: 55221721-55221721 on Build GRCh38
rs770055066 Chr.X: 55221381-55221381 on Build GRCh38
rs934472087 Chr.X: 55221527-55221527 on Build GRCh38
rs774418290 Chr.X: 55221486-55221487 on Build GRCh38
rs1030115200 Chr.X: 55221689-55221689 on Build GRCh38
rs113248496 Chr.X: 55221614-55221614 on Build GRCh38
rs755521905 Chr.X: 55221492-55221492 on Build GRCh38
rs968621615 Chr.X: 55221470-55221470 on Build GRCh38
rs61745114 Chr.X: 55221384-55221384 on Build GRCh38
rs778506253 Chr.X: 55221483-55221483 on Build GRCh38
rs781651026 Chr.X: 55221504-55221504 on Build GRCh38
rs759965291 Chr.X: 55221418-55221418 on Build GRCh38
rs111692798 Chr.X: 55221392-55221392 on Build GRCh38
rs188012543 Chr.X: 55221693-55221693 on Build GRCh38
rs4826274 Chr.X: 55221657-55221657 on Build GRCh38
rs759570775 Chr.X: 55221409-55221409 on Build GRCh38
rs183237538 Chr.X: 55221659-55221659 on Build GRCh38
rs762136316 Chr.X: 55221347-55221347 on Build GRCh38
rs752962148 Chr.X: 55221564-55221564 on Build GRCh38
rs771012163 Chr.X: 55221744-55221744 on Build GRCh38
rs770143069 Chr.X: 55221722-55221722 on Build GRCh38
rs149003149 Chr.X: 55221264-55221264 on Build GRCh38
rs898009252 Chr.X: 55221675-55221675 on Build GRCh38
rs1045025115 Chr.X: 55221569-55221569 on Build GRCh38

View Full Product Details