Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 149480403-149480912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDS
Entrez Gene ID
3423
Gene Name
iduronate 2-sulfatase
Gene Aliases
MPS2, SIDS
UniGene
Hs.460960
Gene Chromosome Location
Chr.X: 149476990-149505354 on Build GRCh38
Transcripts
NM_000202.7 NM_001166550.3

SNP Detail

SNP ID Chromosome Location
rs782467736 Chr.X: 149480583-149480583 on Build GRCh38
rs781995832 Chr.X: 149480824-149480824 on Build GRCh38
rs5936293 Chr.X: 149480636-149480636 on Build GRCh38
rs782217576 Chr.X: 149480775-149480775 on Build GRCh38
rs1020417824 Chr.X: 149480713-149480713 on Build GRCh38
rs142809960 Chr.X: 149480671-149480671 on Build GRCh38
rs891067160 Chr.X: 149480544-149480544 on Build GRCh38
rs782682910 Chr.X: 149480453-149480453 on Build GRCh38
rs782147463 Chr.X: 149480481-149480481 on Build GRCh38
rs147148694 Chr.X: 149480785-149480785 on Build GRCh38
rs150166269 Chr.X: 149480523-149480523 on Build GRCh38
rs143500137 Chr.X: 149480770-149480770 on Build GRCh38
rs942231917 Chr.X: 149480786-149480786 on Build GRCh38
rs184001799 Chr.X: 149480755-149480755 on Build GRCh38
rs1029272363 Chr.X: 149480532-149480532 on Build GRCh38
rs186890639 Chr.X: 149480859-149480859 on Build GRCh38
rs781858361 Chr.X: 149480641-149480641 on Build GRCh38
rs782382293 Chr.X: 149480745-149480745 on Build GRCh38
rs782189646 Chr.X: 149480556-149480556 on Build GRCh38
rs147395729 Chr.X: 149480722-149480722 on Build GRCh38
rs781839654 Chr.X: 149480543-149480543 on Build GRCh38
rs782458654 Chr.X: 149480606-149480606 on Build GRCh38
rs138033183 Chr.X: 149480723-149480723 on Build GRCh38
rs896907620 Chr.X: 149480486-149480486 on Build GRCh38
rs782032446 Chr.X: 149480430-149480430 on Build GRCh38

View Full Product Details