Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 151408455-151408960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VMA21
Entrez Gene ID
203547
Gene Name
VMA21 vacuolar H+-ATPase homolog (S. cerevisiae)
Gene Aliases
MEAX, XMEA
UniGene
Hs.58633
Gene Chromosome Location
Chr.X: 151396555-151409364 on Build GRCh38
Transcripts
NM_001017980.3 XM_011531125.2

SNP Detail

SNP ID Chromosome Location
rs746290338 Chr.X: 151408900-151408900 on Build GRCh38
rs191626805 Chr.X: 151408517-151408517 on Build GRCh38
rs757363161 Chr.X: 151408703-151408703 on Build GRCh38
rs934689523 Chr.X: 151408489-151408489 on Build GRCh38
rs1018022017 Chr.X: 151408824-151408824 on Build GRCh38
rs926056720 Chr.X: 151408888-151408888 on Build GRCh38
rs1053358656 Chr.X: 151408495-151408495 on Build GRCh38
rs964378269 Chr.X: 151408787-151408787 on Build GRCh38
rs989101489 Chr.X: 151408821-151408821 on Build GRCh38
rs961065078 Chr.X: 151408681-151408681 on Build GRCh38
rs978859353 Chr.X: 151408885-151408885 on Build GRCh38
rs989196966 Chr.X: 151408914-151408914 on Build GRCh38
rs910226108 Chr.X: 151408756-151408756 on Build GRCh38
rs896091269 Chr.X: 151408512-151408512 on Build GRCh38
rs988090893 Chr.X: 151408733-151408733 on Build GRCh38
rs761738015 Chr.X: 151408677-151408677 on Build GRCh38
rs756014647 Chr.X: 151408896-151408896 on Build GRCh38
rs6627162 Chr.X: 151408606-151408606 on Build GRCh38
rs915012919 Chr.X: 151408825-151408825 on Build GRCh38
rs781342453 Chr.X: 151408834-151408834 on Build GRCh38
rs985350664 Chr.X: 151408792-151408792 on Build GRCh38
rs147083260 Chr.X: 151408608-151408608 on Build GRCh38
rs1017135984 Chr.X: 151408647-151408647 on Build GRCh38
rs41314149 Chr.X: 151408567-151408567 on Build GRCh38
rs183880461 Chr.X: 151408630-151408630 on Build GRCh38
rs1026872938 Chr.X: 151408687-151408687 on Build GRCh38
rs1029349056 Chr.X: 151408778-151408778 on Build GRCh38
rs11555859 Chr.X: 151408617-151408617 on Build GRCh38
rs1044556988 Chr.X: 151408524-151408524 on Build GRCh38
rs751861525 Chr.X: 151408688-151408688 on Build GRCh38
rs138457145 Chr.X: 151408715-151408715 on Build GRCh38
rs767618049 Chr.X: 151408684-151408684 on Build GRCh38
rs938259568 Chr.X: 151408539-151408539 on Build GRCh38
rs187756671 Chr.X: 151408653-151408653 on Build GRCh38
rs1005267723 Chr.X: 151408638-151408638 on Build GRCh38
rs917269636 Chr.X: 151408935-151408935 on Build GRCh38

View Full Product Details