Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 151923375-151923882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEA4
Entrez Gene ID
4103
Gene Name
MAGE family member A4
Gene Aliases
CT1.4, MAGE-41, MAGE-X2, MAGE4, MAGE4A, MAGE4B
UniGene
Hs.37107
Gene Chromosome Location
Chr.X: 151912889-151925170 on Build GRCh38
Transcripts
NM_001011548.1 NM_001011549.1 NM_001011550.1 NM_002362.4 XM_005274677.2 XM_005274678.3 XM_005274679.3

SNP Detail

SNP ID Chromosome Location
rs772334971 Chr.X: 151923584-151923584 on Build GRCh38
rs747270872 Chr.X: 151923592-151923592 on Build GRCh38
rs146569098 Chr.X: 151923837-151923837 on Build GRCh38
rs763535308 Chr.X: 151923694-151923694 on Build GRCh38
rs781037021 Chr.X: 151923446-151923446 on Build GRCh38
rs778940909 Chr.X: 151923569-151923569 on Build GRCh38
rs758051896 Chr.X: 151923523-151923523 on Build GRCh38
rs752682980 Chr.X: 151923814-151923814 on Build GRCh38
rs958488475 Chr.X: 151923551-151923551 on Build GRCh38
rs1047246 Chr.X: 151923654-151923654 on Build GRCh38
rs760396181 Chr.X: 151923734-151923734 on Build GRCh38
rs759301904 Chr.X: 151923777-151923777 on Build GRCh38
rs756521337 Chr.X: 151923571-151923571 on Build GRCh38
rs776532944 Chr.X: 151923800-151923800 on Build GRCh38
rs990941021 Chr.X: 151923579-151923579 on Build GRCh38
rs952862253 Chr.X: 151923590-151923590 on Build GRCh38
rs746298946 Chr.X: 151923688-151923688 on Build GRCh38
rs752191271 Chr.X: 151923514-151923514 on Build GRCh38
rs373247834 Chr.X: 151923784-151923784 on Build GRCh38
rs753670573 Chr.X: 151923744-151923744 on Build GRCh38
rs372640194 Chr.X: 151923793-151923795 on Build GRCh38
rs746953197 Chr.X: 151923753-151923753 on Build GRCh38
rs766855228 Chr.X: 151923475-151923475 on Build GRCh38
rs943677602 Chr.X: 151923400-151923400 on Build GRCh38
rs758020535 Chr.X: 151923414-151923414 on Build GRCh38
rs769603439 Chr.X: 151923784-151923786 on Build GRCh38
rs148460606 Chr.X: 151923499-151923499 on Build GRCh38
rs762298297 Chr.X: 151923610-151923610 on Build GRCh38
rs368738694 Chr.X: 151923472-151923472 on Build GRCh38
rs748368449 Chr.X: 151923468-151923468 on Build GRCh38
rs369838724 Chr.X: 151923778-151923778 on Build GRCh38
rs1052502633 Chr.X: 151923478-151923478 on Build GRCh38
rs149323734 Chr.X: 151923761-151923761 on Build GRCh38
rs760092762 Chr.X: 151923596-151923596 on Build GRCh38
rs370000858 Chr.X: 151923830-151923830 on Build GRCh38
rs372125339 Chr.X: 151923622-151923622 on Build GRCh38
rs756085292 Chr.X: 151923567-151923567 on Build GRCh38
rs892489387 Chr.X: 151923487-151923487 on Build GRCh38
rs61743686 Chr.X: 151923755-151923755 on Build GRCh38
rs764802119 Chr.X: 151923500-151923500 on Build GRCh38
rs150592561 Chr.X: 151923751-151923751 on Build GRCh38
rs1047248 Chr.X: 151923748-151923748 on Build GRCh38
rs1013977402 Chr.X: 151923491-151923491 on Build GRCh38
rs767725718 Chr.X: 151923821-151923821 on Build GRCh38
rs1011469187 Chr.X: 151923795-151923795 on Build GRCh38
rs764098953 Chr.X: 151923401-151923401 on Build GRCh38
rs750215141 Chr.X: 151923729-151923729 on Build GRCh38
rs757344806 Chr.X: 151923819-151923819 on Build GRCh38
rs148498181 Chr.X: 151923792-151923792 on Build GRCh38
rs777687166 Chr.X: 151923745-151923745 on Build GRCh38
rs191337488 Chr.X: 151923438-151923438 on Build GRCh38
rs763718576 Chr.X: 151923525-151923525 on Build GRCh38
rs749725686 Chr.X: 151923764-151923764 on Build GRCh38
rs773613926 Chr.X: 151923473-151923473 on Build GRCh38
rs763594807 Chr.X: 151923474-151923474 on Build GRCh38
rs377034330 Chr.X: 151923793-151923793 on Build GRCh38
rs769356179 Chr.X: 151923609-151923610 on Build GRCh38
rs756031183 Chr.X: 151923642-151923642 on Build GRCh38
rs771376925 Chr.X: 151923595-151923595 on Build GRCh38
rs767214466 Chr.X: 151923728-151923728 on Build GRCh38
rs866977936 Chr.X: 151923452-151923452 on Build GRCh38
rs780257307 Chr.X: 151923843-151923843 on Build GRCh38
rs200225562 Chr.X: 151923709-151923709 on Build GRCh38
rs183961725 Chr.X: 151923613-151923613 on Build GRCh38
rs759371757 Chr.X: 151923805-151923805 on Build GRCh38
rs770283475 Chr.X: 151923692-151923692 on Build GRCh38
rs775056875 Chr.X: 151923601-151923601 on Build GRCh38
rs41302158 Chr.X: 151923786-151923786 on Build GRCh38
rs866841383 Chr.X: 151923849-151923849 on Build GRCh38
rs905479734 Chr.X: 151923530-151923530 on Build GRCh38
rs780985978 Chr.X: 151923546-151923546 on Build GRCh38
rs751226776 Chr.X: 151923526-151923526 on Build GRCh38
rs965080641 Chr.X: 151923648-151923648 on Build GRCh38
rs779923857 Chr.X: 151923647-151923647 on Build GRCh38
rs902813744 Chr.X: 151923840-151923840 on Build GRCh38
rs750630461 Chr.X: 151923823-151923823 on Build GRCh38
rs767267374 Chr.X: 151923635-151923635 on Build GRCh38
rs965542291 Chr.X: 151923839-151923839 on Build GRCh38
rs764857129 Chr.X: 151923404-151923404 on Build GRCh38
rs745310597 Chr.X: 151923516-151923518 on Build GRCh38
rs756985292 Chr.X: 151923542-151923542 on Build GRCh38
rs766211889 Chr.X: 151923738-151923738 on Build GRCh38
rs765279987 Chr.X: 151923811-151923811 on Build GRCh38
rs745816060 Chr.X: 151923456-151923456 on Build GRCh38
rs777932203 Chr.X: 151923669-151923669 on Build GRCh38
rs773721297 Chr.X: 151923783-151923783 on Build GRCh38
rs781676215 Chr.X: 151923681-151923681 on Build GRCh38
rs142773415 Chr.X: 151923710-151923710 on Build GRCh38
rs1019930953 Chr.X: 151923467-151923467 on Build GRCh38
rs201754404 Chr.X: 151923797-151923797 on Build GRCh38
rs201648066 Chr.X: 151923662-151923662 on Build GRCh38
rs777222146 Chr.X: 151923490-151923490 on Build GRCh38
rs752353797 Chr.X: 151923409-151923409 on Build GRCh38
rs144129975 Chr.X: 151923614-151923614 on Build GRCh38
rs774445671 Chr.X: 151923463-151923463 on Build GRCh38
rs940697185 Chr.X: 151923696-151923696 on Build GRCh38
rs775799958 Chr.X: 151923693-151923693 on Build GRCh38
rs747163064 Chr.X: 151923675-151923675 on Build GRCh38
rs750267906 Chr.X: 151923636-151923636 on Build GRCh38
rs780727589 Chr.X: 151923711-151923711 on Build GRCh38
rs543332211 Chr.X: 151923415-151923415 on Build GRCh38
rs147367105 Chr.X: 151923719-151923719 on Build GRCh38
rs771323726 Chr.X: 151923680-151923680 on Build GRCh38
rs1011936049 Chr.X: 151923434-151923434 on Build GRCh38
rs773242799 Chr.X: 151923586-151923586 on Build GRCh38
rs764002635 Chr.X: 151923440-151923440 on Build GRCh38
rs772345662 Chr.X: 151923747-151923747 on Build GRCh38
rs761483501 Chr.X: 151923615-151923615 on Build GRCh38

View Full Product Details