Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49014974-49015510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VSTM4
Entrez Gene ID
196740
Gene Name
V-set and transmembrane domain containing 4
Gene Aliases
C10orf72
UniGene
Hs.522928
Gene Chromosome Location
Chr.10: 49014245-49129118 on Build GRCh38
Transcripts
NM_001031746.4 XM_017015828.1

SNP Detail

SNP ID Chromosome Location
rs559991423 Chr.10: 49015119-49015119 on Build GRCh38
rs3920245 Chr.10: 49015411-49015411 on Build GRCh38
rs1017936719 Chr.10: 49015342-49015342 on Build GRCh38
rs1025454300 Chr.10: 49015275-49015275 on Build GRCh38
rs925574641 Chr.10: 49015378-49015378 on Build GRCh38
rs942592344 Chr.10: 49015361-49015361 on Build GRCh38
rs545605854 Chr.10: 49015478-49015478 on Build GRCh38
rs1026906015 Chr.10: 49015312-49015312 on Build GRCh38
rs534798213 Chr.10: 49015222-49015222 on Build GRCh38
rs945507949 Chr.10: 49015208-49015208 on Build GRCh38
rs1045704798 Chr.10: 49015115-49015115 on Build GRCh38
rs936913123 Chr.10: 49015382-49015382 on Build GRCh38
rs985085059 Chr.10: 49015122-49015122 on Build GRCh38
rs186379016 Chr.10: 49015277-49015277 on Build GRCh38
rs17700453 Chr.10: 49015481-49015481 on Build GRCh38
rs1042629409 Chr.10: 49015214-49015214 on Build GRCh38
rs768687050 Chr.10: 49015375-49015375 on Build GRCh38
rs911012459 Chr.10: 49015160-49015160 on Build GRCh38
rs1013651015 Chr.10: 49015247-49015247 on Build GRCh38
rs183934994 Chr.10: 49015445-49015445 on Build GRCh38
rs552771190 Chr.10: 49015226-49015226 on Build GRCh38
rs570313287 Chr.10: 49015149-49015149 on Build GRCh38
rs1026875421 Chr.10: 49015111-49015111 on Build GRCh38
rs769997100 Chr.10: 49015241-49015241 on Build GRCh38
rs1024794792 Chr.10: 49015022-49015022 on Build GRCh38
rs976543058 Chr.10: 49015357-49015357 on Build GRCh38
rs869264297 Chr.10: 49015139-49015139 on Build GRCh38
rs548982900 Chr.10: 49015215-49015215 on Build GRCh38
rs972691905 Chr.10: 49015105-49015105 on Build GRCh38
rs761495787 Chr.10: 49015122-49015123 on Build GRCh38
rs538814659 Chr.10: 49015319-49015319 on Build GRCh38
rs943790556 Chr.10: 49015182-49015182 on Build GRCh38
rs76103698 Chr.10: 49015140-49015140 on Build GRCh38
rs541799253 Chr.10: 49015437-49015437 on Build GRCh38
rs1014701093 Chr.10: 49015056-49015056 on Build GRCh38
rs564963408 Chr.10: 49015452-49015452 on Build GRCh38
rs866230045 Chr.10: 49015144-49015144 on Build GRCh38
rs925159616 Chr.10: 49015386-49015386 on Build GRCh38
rs539214338 Chr.10: 49015197-49015197 on Build GRCh38
rs966908461 Chr.10: 49015367-49015367 on Build GRCh38
rs551826540 Chr.10: 49015148-49015148 on Build GRCh38
rs889687084 Chr.10: 49015352-49015352 on Build GRCh38
rs546675117 Chr.10: 49015107-49015107 on Build GRCh38
rs567283542 Chr.10: 49015217-49015217 on Build GRCh38
rs192162062 Chr.10: 49015280-49015280 on Build GRCh38
rs949382982 Chr.10: 49015412-49015412 on Build GRCh38
rs898633996 Chr.10: 49015279-49015279 on Build GRCh38
rs533568163 Chr.10: 49015145-49015145 on Build GRCh38
rs140462655 Chr.10: 49015368-49015368 on Build GRCh38
rs762994123 Chr.10: 49015373-49015373 on Build GRCh38
rs976681315 Chr.10: 49015184-49015184 on Build GRCh38
rs1048249378 Chr.10: 49015446-49015446 on Build GRCh38
rs397847688 Chr.10: 49015138-49015138 on Build GRCh38
rs772656586 Chr.10: 49015121-49015123 on Build GRCh38
rs557000511 Chr.10: 49015363-49015363 on Build GRCh38
rs188491358 Chr.10: 49015454-49015454 on Build GRCh38
rs897191531 Chr.10: 49015428-49015428 on Build GRCh38
rs961612943 Chr.10: 49015042-49015042 on Build GRCh38
rs397846277 Chr.10: 49015137-49015137 on Build GRCh38
rs923482497 Chr.10: 49015190-49015190 on Build GRCh38
rs540375748 Chr.10: 49015451-49015451 on Build GRCh38
rs895232941 Chr.10: 49015017-49015017 on Build GRCh38
rs1055070452 Chr.10: 49015401-49015401 on Build GRCh38
rs34800687 Chr.10: 49015138-49015139 on Build GRCh38
rs994372403 Chr.10: 49015299-49015299 on Build GRCh38
rs531083748 Chr.10: 49015191-49015191 on Build GRCh38
rs76502729 Chr.10: 49015020-49015020 on Build GRCh38
rs775584017 Chr.10: 49015246-49015246 on Build GRCh38
rs1026343166 Chr.10: 49015305-49015305 on Build GRCh38
rs1046483690 Chr.10: 49015421-49015421 on Build GRCh38
rs575513404 Chr.10: 49015364-49015364 on Build GRCh38
rs71026232 Chr.10: 49015120-49015123 on Build GRCh38
rs985156828 Chr.10: 49015329-49015329 on Build GRCh38
rs766614311 Chr.10: 49015014-49015014 on Build GRCh38

View Full Product Details