Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26255491-26255992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP85
Entrez Gene ID
64793
Gene Name
centrosomal protein 85
Gene Aliases
CCDC21
UniGene
Hs.109051
Gene Chromosome Location
Chr.1: 26234153-26278810 on Build GRCh38
Transcripts
NM_001281517.2 NM_001319944.1 NM_022778.4 XM_017002105.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
907600 c.876G>T p.L292F Chr.1: 26255838-26255838 on Build GRCh38
6627179 c.881G>A p.R294H Chr.1: 26255843-26255843 on Build GRCh38
4030942 c.644G>A p.S215N Chr.1: 26255606-26255606 on Build GRCh38
2235017 c.864G>A p.Q288Q Chr.1: 26255826-26255826 on Build GRCh38
4030943 c.656A>G p.Y219C Chr.1: 26255618-26255618 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779326361 Chr.1: 26255608-26255608 on Build GRCh38
rs752071338 Chr.1: 26255871-26255871 on Build GRCh38
rs776277007 Chr.1: 26255833-26255833 on Build GRCh38
rs573572402 Chr.1: 26255950-26255950 on Build GRCh38
rs759492304 Chr.1: 26255534-26255534 on Build GRCh38
rs375455730 Chr.1: 26255632-26255632 on Build GRCh38
rs773078870 Chr.1: 26255926-26255926 on Build GRCh38
rs781186845 Chr.1: 26255791-26255791 on Build GRCh38
rs774208005 Chr.1: 26255647-26255647 on Build GRCh38
rs200313680 Chr.1: 26255578-26255578 on Build GRCh38
rs768418031 Chr.1: 26255638-26255638 on Build GRCh38
rs763650612 Chr.1: 26255731-26255731 on Build GRCh38
rs780666368 Chr.1: 26255528-26255528 on Build GRCh38
rs763404526 Chr.1: 26255851-26255851 on Build GRCh38
rs753559442 Chr.1: 26255743-26255743 on Build GRCh38
rs201675817 Chr.1: 26255703-26255703 on Build GRCh38
rs756333400 Chr.1: 26255884-26255884 on Build GRCh38
rs745647685 Chr.1: 26255811-26255811 on Build GRCh38
rs778089640 Chr.1: 26255887-26255887 on Build GRCh38
rs748406698 Chr.1: 26255673-26255673 on Build GRCh38
rs201391625 Chr.1: 26255551-26255551 on Build GRCh38
rs749814661 Chr.1: 26255522-26255522 on Build GRCh38
rs746607239 Chr.1: 26255623-26255624 on Build GRCh38
rs556682258 Chr.1: 26255878-26255878 on Build GRCh38
rs148974720 Chr.1: 26255564-26255564 on Build GRCh38
rs530085981 Chr.1: 26255633-26255633 on Build GRCh38
rs1017341669 Chr.1: 26255842-26255842 on Build GRCh38
rs760874252 Chr.1: 26255555-26255555 on Build GRCh38
rs755068709 Chr.1: 26255783-26255783 on Build GRCh38
rs922659413 Chr.1: 26255850-26255850 on Build GRCh38
rs750153502 Chr.1: 26255568-26255568 on Build GRCh38
rs749618618 Chr.1: 26255602-26255602 on Build GRCh38
rs1018015977 Chr.1: 26255591-26255591 on Build GRCh38
rs764919627 Chr.1: 26255843-26255843 on Build GRCh38
rs771503845 Chr.1: 26255527-26255527 on Build GRCh38
rs956344185 Chr.1: 26255935-26255935 on Build GRCh38
rs913480513 Chr.1: 26255959-26255959 on Build GRCh38
rs759508200 Chr.1: 26255648-26255648 on Build GRCh38
rs751114445 Chr.1: 26255579-26255579 on Build GRCh38
rs760634049 Chr.1: 26255822-26255822 on Build GRCh38
rs762699203 Chr.1: 26255634-26255634 on Build GRCh38
rs933530885 Chr.1: 26255639-26255639 on Build GRCh38
rs374815195 Chr.1: 26255744-26255744 on Build GRCh38
rs145588256 Chr.1: 26255584-26255584 on Build GRCh38
rs750786638 Chr.1: 26255767-26255767 on Build GRCh38
rs138144782 Chr.1: 26255849-26255849 on Build GRCh38
rs993261124 Chr.1: 26255761-26255761 on Build GRCh38
rs768286565 Chr.1: 26255825-26255825 on Build GRCh38
rs888312509 Chr.1: 26255823-26255823 on Build GRCh38
rs765586972 Chr.1: 26255752-26255752 on Build GRCh38
rs759453442 Chr.1: 26255706-26255706 on Build GRCh38
rs772159240 Chr.1: 26255545-26255545 on Build GRCh38
rs780165101 Chr.1: 26255679-26255679 on Build GRCh38
rs139000810 Chr.1: 26255684-26255684 on Build GRCh38
rs1889739 Chr.1: 26255662-26255662 on Build GRCh38
rs778626387 Chr.1: 26255797-26255797 on Build GRCh38
rs752018514 Chr.1: 26255780-26255780 on Build GRCh38
rs753502469 Chr.1: 26255664-26255664 on Build GRCh38
rs373280524 Chr.1: 26255649-26255649 on Build GRCh38
rs770641977 Chr.1: 26255697-26255697 on Build GRCh38
rs3795686 Chr.1: 26255600-26255600 on Build GRCh38
rs369818657 Chr.1: 26255645-26255645 on Build GRCh38
rs914100333 Chr.1: 26255964-26255964 on Build GRCh38
rs770046964 Chr.1: 26255796-26255796 on Build GRCh38
rs748318340 Chr.1: 26255792-26255792 on Build GRCh38
rs199665247 Chr.1: 26255671-26255671 on Build GRCh38
rs565099659 Chr.1: 26255585-26255585 on Build GRCh38
rs895701659 Chr.1: 26255536-26255536 on Build GRCh38
rs191422364 Chr.1: 26255573-26255573 on Build GRCh38
rs373753784 Chr.1: 26255769-26255769 on Build GRCh38
rs779280111 Chr.1: 26255672-26255672 on Build GRCh38
rs775635614 Chr.1: 26255616-26255616 on Build GRCh38
rs369014809 Chr.1: 26255614-26255614 on Build GRCh38
rs775182640 Chr.1: 26255722-26255722 on Build GRCh38
rs1005381590 Chr.1: 26255847-26255847 on Build GRCh38
rs758939245 Chr.1: 26255674-26255674 on Build GRCh38
rs752802840 Chr.1: 26255882-26255882 on Build GRCh38
rs150424067 Chr.1: 26255689-26255689 on Build GRCh38
rs1006177224 Chr.1: 26255838-26255838 on Build GRCh38
rs758876939 Chr.1: 26255768-26255768 on Build GRCh38
rs567012962 Chr.1: 26255656-26255656 on Build GRCh38
rs187115380 Chr.1: 26255821-26255821 on Build GRCh38
rs895753830 Chr.1: 26255787-26255787 on Build GRCh38
rs761429564 Chr.1: 26255840-26255840 on Build GRCh38
rs749423189 Chr.1: 26255902-26255902 on Build GRCh38
rs755883407 Chr.1: 26255655-26255655 on Build GRCh38
rs1057029712 Chr.1: 26255705-26255705 on Build GRCh38
rs199807594 Chr.1: 26255604-26255604 on Build GRCh38
rs561246833 Chr.1: 26255619-26255619 on Build GRCh38
rs766678709 Chr.1: 26255862-26255862 on Build GRCh38
rs963797343 Chr.1: 26255854-26255854 on Build GRCh38
rs771955434 Chr.1: 26255819-26255819 on Build GRCh38
rs149507810 Chr.1: 26255712-26255712 on Build GRCh38
rs760294988 Chr.1: 26255730-26255730 on Build GRCh38
rs746363032 Chr.1: 26255612-26255612 on Build GRCh38
rs535832298 Chr.1: 26255965-26255965 on Build GRCh38
rs755407610 Chr.1: 26255876-26255876 on Build GRCh38
rs755756505 Chr.1: 26255592-26255592 on Build GRCh38
rs756867294 Chr.1: 26255751-26255751 on Build GRCh38

View Full Product Details