Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40016407-40016913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF3
Entrez Gene ID
1440
Gene Name
colony stimulating factor 3
Gene Aliases
C17orf33, CSF3OS, GCSF
UniGene
Hs.2233
Gene Chromosome Location
Chr.17: 40015361-40017813 on Build GRCh38
Transcripts
NM_000759.3 NM_001178147.1 NM_172219.2 NM_172220.2 NR_033662.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6573523 c.292-5C>G p.? Chr.17: 40016480-40016480 on Build GRCh38
3819472 c.318C>G p.S106R Chr.17: 40016511-40016511 on Build GRCh38
6636125 c.365C>A p.S122Y Chr.17: 40016558-40016558 on Build GRCh38
6190937 c.520G>A p.A174T Chr.17: 40016855-40016855 on Build GRCh38
4443651 c.376G>T p.G126C Chr.17: 40016569-40016569 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367994501 Chr.17: 40016720-40016720 on Build GRCh38
rs764728755 Chr.17: 40016646-40016646 on Build GRCh38
rs375506229 Chr.17: 40016605-40016605 on Build GRCh38
rs2227338 Chr.17: 40016689-40016689 on Build GRCh38
rs768714791 Chr.17: 40016604-40016604 on Build GRCh38
rs767498933 Chr.17: 40016871-40016871 on Build GRCh38
rs772749039 Chr.17: 40016874-40016874 on Build GRCh38
rs761136068 Chr.17: 40016620-40016620 on Build GRCh38
rs762571703 Chr.17: 40016655-40016655 on Build GRCh38
rs375245553 Chr.17: 40016598-40016598 on Build GRCh38
rs374065088 Chr.17: 40016441-40016441 on Build GRCh38
rs774542221 Chr.17: 40016547-40016547 on Build GRCh38
rs9282660 Chr.17: 40016848-40016848 on Build GRCh38
rs33934943 Chr.17: 40016721-40016722 on Build GRCh38
rs1009218442 Chr.17: 40016887-40016887 on Build GRCh38
rs2227330 Chr.17: 40016855-40016855 on Build GRCh38
rs752375006 Chr.17: 40016566-40016566 on Build GRCh38
rs765324783 Chr.17: 40016886-40016886 on Build GRCh38
rs772085839 Chr.17: 40016435-40016435 on Build GRCh38
rs749503197 Chr.17: 40016533-40016533 on Build GRCh38
rs777777864 Chr.17: 40016500-40016500 on Build GRCh38
rs752567650 Chr.17: 40016490-40016490 on Build GRCh38
rs145136406 Chr.17: 40016561-40016561 on Build GRCh38
rs763688260 Chr.17: 40016859-40016859 on Build GRCh38
rs745980343 Chr.17: 40016440-40016440 on Build GRCh38
rs531033567 Chr.17: 40016881-40016881 on Build GRCh38
rs138252418 Chr.17: 40016550-40016550 on Build GRCh38
rs1038032378 Chr.17: 40016780-40016780 on Build GRCh38
rs763576148 Chr.17: 40016699-40016699 on Build GRCh38
rs748383311 Chr.17: 40016613-40016613 on Build GRCh38
rs369553702 Chr.17: 40016821-40016821 on Build GRCh38
rs758218759 Chr.17: 40016493-40016493 on Build GRCh38
rs754670741 Chr.17: 40016489-40016489 on Build GRCh38
rs949117496 Chr.17: 40016761-40016761 on Build GRCh38
rs187929921 Chr.17: 40016471-40016471 on Build GRCh38
rs9282661 Chr.17: 40016787-40016787 on Build GRCh38
rs763335706 Chr.17: 40016448-40016448 on Build GRCh38
rs371912793 Chr.17: 40016587-40016587 on Build GRCh38
rs190847104 Chr.17: 40016874-40016874 on Build GRCh38
rs780254653 Chr.17: 40016601-40016601 on Build GRCh38
rs139012113 Chr.17: 40016562-40016562 on Build GRCh38
rs368124550 Chr.17: 40016508-40016508 on Build GRCh38
rs759057587 Chr.17: 40016643-40016643 on Build GRCh38
rs930250534 Chr.17: 40016706-40016706 on Build GRCh38
rs25645 Chr.17: 40016890-40016890 on Build GRCh38
rs762262454 Chr.17: 40016549-40016549 on Build GRCh38
rs918093535 Chr.17: 40016750-40016750 on Build GRCh38
rs200609044 Chr.17: 40016637-40016637 on Build GRCh38
rs563103855 Chr.17: 40016720-40016721 on Build GRCh38
rs368443474 Chr.17: 40016563-40016563 on Build GRCh38
rs558514549 Chr.17: 40016716-40016716 on Build GRCh38
rs934115679 Chr.17: 40016431-40016431 on Build GRCh38
rs774882119 Chr.17: 40016514-40016514 on Build GRCh38
rs752391702 Chr.17: 40016648-40016648 on Build GRCh38
rs763928631 Chr.17: 40016660-40016660 on Build GRCh38
rs753472086 Chr.17: 40016473-40016473 on Build GRCh38
rs1047211521 Chr.17: 40016729-40016729 on Build GRCh38
rs776070400 Chr.17: 40016642-40016642 on Build GRCh38
rs778915010 Chr.17: 40016763-40016763 on Build GRCh38
rs987840995 Chr.17: 40016686-40016686 on Build GRCh38
rs757308934 Chr.17: 40016599-40016599 on Build GRCh38
rs144408123 Chr.17: 40016870-40016870 on Build GRCh38
rs778070313 Chr.17: 40016788-40016788 on Build GRCh38
rs1024291938 Chr.17: 40016576-40016576 on Build GRCh38
rs746215629 Chr.17: 40016783-40016783 on Build GRCh38
rs776532349 Chr.17: 40016436-40016436 on Build GRCh38
rs142286142 Chr.17: 40016567-40016567 on Build GRCh38
rs781678207 Chr.17: 40016749-40016749 on Build GRCh38
rs141665234 Chr.17: 40016826-40016826 on Build GRCh38
rs774832032 Chr.17: 40016458-40016458 on Build GRCh38
rs748376299 Chr.17: 40016772-40016772 on Build GRCh38
rs769990271 Chr.17: 40016512-40016512 on Build GRCh38
rs370045874 Chr.17: 40016663-40016663 on Build GRCh38
rs1045209841 Chr.17: 40016847-40016847 on Build GRCh38
rs150434691 Chr.17: 40016523-40016523 on Build GRCh38
rs1018373215 Chr.17: 40016857-40016857 on Build GRCh38
rs1014098323 Chr.17: 40016827-40016827 on Build GRCh38
rs755623838 Chr.17: 40016470-40016470 on Build GRCh38
rs374689597 Chr.17: 40016662-40016662 on Build GRCh38
rs776139338 Chr.17: 40016881-40016882 on Build GRCh38
rs5820317 Chr.17: 40016721-40016721 on Build GRCh38
rs764540509 Chr.17: 40016452-40016452 on Build GRCh38
rs910499547 Chr.17: 40016499-40016499 on Build GRCh38
rs202204611 Chr.17: 40016790-40016790 on Build GRCh38
rs869226700 Chr.17: 40016720-40016721 on Build GRCh38
rs772466210 Chr.17: 40016618-40016618 on Build GRCh38
rs753914662 Chr.17: 40016748-40016748 on Build GRCh38
rs777497317 Chr.17: 40016586-40016586 on Build GRCh38
rs769044135 Chr.17: 40016544-40016544 on Build GRCh38
rs775861491 Chr.17: 40016443-40016443 on Build GRCh38
rs749855586 Chr.17: 40016467-40016467 on Build GRCh38
rs2227329 Chr.17: 40016804-40016804 on Build GRCh38
rs148409381 Chr.17: 40016806-40016806 on Build GRCh38
rs746429744 Chr.17: 40016836-40016836 on Build GRCh38
rs920164778 Chr.17: 40016703-40016703 on Build GRCh38
rs755078233 Chr.17: 40016761-40016761 on Build GRCh38
rs749415034 Chr.17: 40016602-40016602 on Build GRCh38
rs985954147 Chr.17: 40016742-40016742 on Build GRCh38
rs954532967 Chr.17: 40016731-40016731 on Build GRCh38
rs768103264 Chr.17: 40016462-40016462 on Build GRCh38
rs773611963 Chr.17: 40016619-40016619 on Build GRCh38
rs987955582 Chr.17: 40016572-40016572 on Build GRCh38
rs762199800 Chr.17: 40016459-40016459 on Build GRCh38
rs774912212 Chr.17: 40016854-40016854 on Build GRCh38
rs143679996 Chr.17: 40016552-40016552 on Build GRCh38

View Full Product Details