Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41365088-41365593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT33B
Entrez Gene ID
3884
Gene Name
keratin 33B
Gene Aliases
HA3II, Ha-3II, K33B, KRTHA3A, KRTHA3B, hHa3-II
UniGene
Hs.32950
Gene Chromosome Location
Chr.17: 41363494-41369800 on Build GRCh38
Transcripts
NM_002279.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3795596 c.652G>A p.A218T Chr.17: 41365490-41365490 on Build GRCh38
6386462 c.792G>T p.Q264H Chr.17: 41365259-41365259 on Build GRCh38
7348141 c.655G>T p.V219L Chr.17: 41365487-41365487 on Build GRCh38
5067376 c.810G>A p.A270A Chr.17: 41365241-41365241 on Build GRCh38
6375678 c.809C>T p.A270V Chr.17: 41365242-41365242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749937008 Chr.17: 41365140-41365140 on Build GRCh38
rs757559638 Chr.17: 41365449-41365449 on Build GRCh38
rs143499346 Chr.17: 41365499-41365499 on Build GRCh38
rs776086937 Chr.17: 41365323-41365323 on Build GRCh38
rs574721377 Chr.17: 41365293-41365293 on Build GRCh38
rs546179706 Chr.17: 41365331-41365331 on Build GRCh38
rs751598347 Chr.17: 41365341-41365341 on Build GRCh38
rs202102547 Chr.17: 41365316-41365316 on Build GRCh38
rs754832278 Chr.17: 41365158-41365158 on Build GRCh38
rs767642130 Chr.17: 41365491-41365491 on Build GRCh38
rs377003814 Chr.17: 41365211-41365211 on Build GRCh38
rs769343695 Chr.17: 41365396-41365396 on Build GRCh38
rs772410804 Chr.17: 41365308-41365308 on Build GRCh38
rs373505728 Chr.17: 41365406-41365406 on Build GRCh38
rs747499855 Chr.17: 41365317-41365317 on Build GRCh38
rs148300082 Chr.17: 41365400-41365400 on Build GRCh38
rs779445556 Chr.17: 41365202-41365202 on Build GRCh38
rs541403283 Chr.17: 41365258-41365258 on Build GRCh38
rs777726481 Chr.17: 41365206-41365206 on Build GRCh38
rs754475517 Chr.17: 41365509-41365509 on Build GRCh38
rs79296577 Chr.17: 41365242-41365242 on Build GRCh38
rs774321951 Chr.17: 41365401-41365401 on Build GRCh38
rs377476050 Chr.17: 41365467-41365467 on Build GRCh38
rs775641878 Chr.17: 41365244-41365268 on Build GRCh38
rs958775673 Chr.17: 41365542-41365542 on Build GRCh38
rs141441300 Chr.17: 41365424-41365424 on Build GRCh38
rs768448141 Chr.17: 41365461-41365461 on Build GRCh38
rs774604733 Chr.17: 41365551-41365551 on Build GRCh38
rs773026347 Chr.17: 41365525-41365525 on Build GRCh38
rs760179357 Chr.17: 41365241-41365241 on Build GRCh38
rs554731588 Chr.17: 41365204-41365204 on Build GRCh38
rs367670650 Chr.17: 41365145-41365145 on Build GRCh38
rs755189451 Chr.17: 41365297-41365297 on Build GRCh38
rs891076059 Chr.17: 41365379-41365379 on Build GRCh38
rs1014739796 Chr.17: 41365488-41365488 on Build GRCh38
rs752727290 Chr.17: 41365166-41365166 on Build GRCh38
rs747784843 Chr.17: 41365519-41365519 on Build GRCh38
rs753417138 Chr.17: 41365154-41365154 on Build GRCh38
rs529947864 Chr.17: 41365464-41365464 on Build GRCh38
rs763024692 Chr.17: 41365548-41365548 on Build GRCh38
rs371331851 Chr.17: 41365376-41365376 on Build GRCh38
rs773077331 Chr.17: 41365473-41365473 on Build GRCh38
rs537065960 Chr.17: 41365217-41365217 on Build GRCh38
rs769237835 Chr.17: 41365455-41365455 on Build GRCh38
rs370317934 Chr.17: 41365538-41365538 on Build GRCh38
rs200967808 Chr.17: 41365359-41365359 on Build GRCh38
rs61741664 Chr.17: 41365500-41365500 on Build GRCh38
rs776954416 Chr.17: 41365349-41365349 on Build GRCh38
rs750935787 Chr.17: 41365495-41365495 on Build GRCh38
rs374338902 Chr.17: 41365492-41365492 on Build GRCh38
rs565920839 Chr.17: 41365202-41365202 on Build GRCh38
rs748507327 Chr.17: 41365224-41365224 on Build GRCh38
rs765341322 Chr.17: 41365423-41365423 on Build GRCh38
rs765271023 Chr.17: 41365475-41365475 on Build GRCh38
rs991432095 Chr.17: 41365543-41365543 on Build GRCh38
rs770474244 Chr.17: 41365319-41365319 on Build GRCh38
rs775148923 Chr.17: 41365336-41365336 on Build GRCh38
rs929622875 Chr.17: 41365234-41365234 on Build GRCh38
rs114488848 Chr.17: 41365216-41365216 on Build GRCh38
rs764818986 Chr.17: 41365334-41365334 on Build GRCh38
rs760616191 Chr.17: 41365274-41365275 on Build GRCh38
rs769114866 Chr.17: 41365218-41365218 on Build GRCh38
rs758235238 Chr.17: 41365199-41365199 on Build GRCh38
rs532425696 Chr.17: 41365375-41365375 on Build GRCh38
rs767525647 Chr.17: 41365342-41365342 on Build GRCh38
rs769138346 Chr.17: 41365133-41365133 on Build GRCh38
rs745382068 Chr.17: 41365395-41365395 on Build GRCh38
rs751941992 Chr.17: 41365425-41365425 on Build GRCh38
rs745967336 Chr.17: 41365208-41365208 on Build GRCh38
rs771250644 Chr.17: 41365169-41365174 on Build GRCh38
rs868781611 Chr.17: 41365415-41365415 on Build GRCh38
rs759598244 Chr.17: 41365418-41365418 on Build GRCh38
rs374912971 Chr.17: 41365134-41365134 on Build GRCh38
rs752466716 Chr.17: 41365256-41365256 on Build GRCh38
rs779587635 Chr.17: 41365458-41365458 on Build GRCh38
rs771457438 Chr.17: 41365387-41365387 on Build GRCh38
rs772109572 Chr.17: 41365217-41365218 on Build GRCh38
rs751713129 Chr.17: 41365490-41365490 on Build GRCh38
rs780657565 Chr.17: 41365151-41365188 on Build GRCh38
rs748479432 Chr.17: 41365299-41365299 on Build GRCh38
rs776320928 Chr.17: 41365248-41365248 on Build GRCh38
rs1010579669 Chr.17: 41365115-41365115 on Build GRCh38
rs748846721 Chr.17: 41365507-41365507 on Build GRCh38
rs61741663 Chr.17: 41365474-41365474 on Build GRCh38
rs755031342 Chr.17: 41365354-41365354 on Build GRCh38
rs749681369 Chr.17: 41365167-41365168 on Build GRCh38
rs1020237021 Chr.17: 41365121-41365121 on Build GRCh38
rs547758340 Chr.17: 41365410-41365410 on Build GRCh38
rs765118403 Chr.17: 41365250-41365250 on Build GRCh38
rs374090545 Chr.17: 41365358-41365358 on Build GRCh38
rs947551673 Chr.17: 41365113-41365113 on Build GRCh38
rs558957351 Chr.17: 41365222-41365222 on Build GRCh38
rs138903249 Chr.17: 41365249-41365249 on Build GRCh38
rs770873170 Chr.17: 41365231-41365231 on Build GRCh38
rs768946992 Chr.17: 41365348-41365349 on Build GRCh38
rs781666493 Chr.17: 41365452-41365452 on Build GRCh38
rs181987869 Chr.17: 41365137-41365137 on Build GRCh38
rs559565368 Chr.17: 41365453-41365453 on Build GRCh38
rs906018761 Chr.17: 41365310-41365310 on Build GRCh38
rs750322005 Chr.17: 41365417-41365417 on Build GRCh38
rs772037496 Chr.17: 41365520-41365520 on Build GRCh38
rs767097018 Chr.17: 41365139-41365139 on Build GRCh38
rs1018105241 Chr.17: 41365186-41365186 on Build GRCh38
rs536347994 Chr.17: 41365203-41365203 on Build GRCh38
rs767846416 Chr.17: 41365493-41365493 on Build GRCh38
rs763943969 Chr.17: 41365263-41365263 on Build GRCh38
rs748708569 Chr.17: 41365459-41365459 on Build GRCh38
rs144208752 Chr.17: 41365298-41365298 on Build GRCh38
rs778778969 Chr.17: 41365512-41365512 on Build GRCh38
rs772482801 Chr.17: 41365228-41365228 on Build GRCh38
rs376169969 Chr.17: 41365340-41365340 on Build GRCh38
rs746738328 Chr.17: 41365537-41365537 on Build GRCh38
rs746359618 Chr.17: 41365204-41365205 on Build GRCh38
rs201040375 Chr.17: 41365487-41365487 on Build GRCh38
rs760132625 Chr.17: 41365143-41365143 on Build GRCh38
rs780491524 Chr.17: 41365212-41365212 on Build GRCh38
rs773713985 Chr.17: 41365232-41365232 on Build GRCh38
rs753995601 Chr.17: 41365353-41365353 on Build GRCh38
rs541906440 Chr.17: 41365294-41365294 on Build GRCh38
rs951202482 Chr.17: 41365428-41365428 on Build GRCh38
rs1040011424 Chr.17: 41365146-41365146 on Build GRCh38
rs367985598 Chr.17: 41365368-41365368 on Build GRCh38
rs778556896 Chr.17: 41365165-41365165 on Build GRCh38
rs781628070 Chr.17: 41365391-41365391 on Build GRCh38
rs199886059 Chr.17: 41365451-41365451 on Build GRCh38
rs200459142 Chr.17: 41365476-41365476 on Build GRCh38
rs764153782 Chr.17: 41365550-41365550 on Build GRCh38
rs553149537 Chr.17: 41365265-41365265 on Build GRCh38

View Full Product Details