Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 56494654-56495198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLPP3
Entrez Gene ID
8613
Gene Name
phospholipid phosphatase 3
Gene Aliases
Dri42, LPP3, PAP2B, PPAP2B, VCIP
UniGene
Hs.405156
Gene Chromosome Location
Chr.1: 56494747-56579584 on Build GRCh38
Transcripts
NM_003713.4

SNP Detail

SNP ID Chromosome Location
rs764816599 Chr.1: 56495048-56495049 on Build GRCh38
rs1050155119 Chr.1: 56494904-56494904 on Build GRCh38
rs538783643 Chr.1: 56494926-56494926 on Build GRCh38
rs555658179 Chr.1: 56494940-56494941 on Build GRCh38
rs763953607 Chr.1: 56495045-56495045 on Build GRCh38
rs969851979 Chr.1: 56495057-56495057 on Build GRCh38
rs918270629 Chr.1: 56494940-56494940 on Build GRCh38
rs994389191 Chr.1: 56495010-56495010 on Build GRCh38
rs972702168 Chr.1: 56494859-56494859 on Build GRCh38
rs999965715 Chr.1: 56494713-56494713 on Build GRCh38
rs17114018 Chr.1: 56495042-56495042 on Build GRCh38
rs1052753912 Chr.1: 56495050-56495050 on Build GRCh38
rs889191103 Chr.1: 56495129-56495129 on Build GRCh38
rs901718885 Chr.1: 56494827-56494827 on Build GRCh38
rs17114017 Chr.1: 56494688-56494688 on Build GRCh38
rs181045708 Chr.1: 56494993-56494993 on Build GRCh38
rs189953915 Chr.1: 56495062-56495062 on Build GRCh38
rs186289233 Chr.1: 56495020-56495020 on Build GRCh38
rs760748686 Chr.1: 56494755-56494755 on Build GRCh38
rs991909307 Chr.1: 56494965-56494965 on Build GRCh38
rs962302146 Chr.1: 56494833-56494833 on Build GRCh38
rs1043701175 Chr.1: 56494689-56494689 on Build GRCh38
rs775559078 Chr.1: 56494749-56494749 on Build GRCh38
rs997571884 Chr.1: 56494771-56494771 on Build GRCh38
rs376629600 Chr.1: 56494963-56494963 on Build GRCh38
rs916681969 Chr.1: 56495048-56495048 on Build GRCh38
rs916329405 Chr.1: 56494991-56494991 on Build GRCh38
rs992279913 Chr.1: 56494683-56494683 on Build GRCh38
rs1016962934 Chr.1: 56494832-56494832 on Build GRCh38
rs1052227264 Chr.1: 56494809-56494809 on Build GRCh38
rs555521724 Chr.1: 56494972-56494972 on Build GRCh38
rs1029074344 Chr.1: 56495105-56495105 on Build GRCh38
rs748827243 Chr.1: 56495169-56495169 on Build GRCh38
rs984921914 Chr.1: 56494829-56494829 on Build GRCh38
rs566561829 Chr.1: 56494921-56494921 on Build GRCh38
rs183172799 Chr.1: 56494773-56494773 on Build GRCh38
rs938965016 Chr.1: 56494957-56494957 on Build GRCh38
rs925239546 Chr.1: 56494722-56494722 on Build GRCh38
rs894986907 Chr.1: 56494685-56494685 on Build GRCh38
rs138145826 Chr.1: 56495069-56495069 on Build GRCh38
rs573673447 Chr.1: 56495152-56495152 on Build GRCh38
rs888850648 Chr.1: 56494924-56494924 on Build GRCh38
rs962995897 Chr.1: 56495008-56495008 on Build GRCh38
rs997434429 Chr.1: 56495063-56495063 on Build GRCh38
rs761804973 Chr.1: 56494972-56494974 on Build GRCh38
rs183599204 Chr.1: 56494690-56494690 on Build GRCh38
rs925240038 Chr.1: 56495095-56495095 on Build GRCh38
rs903795368 Chr.1: 56495044-56495044 on Build GRCh38
rs953550534 Chr.1: 56494774-56494774 on Build GRCh38
rs1052339984 Chr.1: 56495110-56495110 on Build GRCh38

View Full Product Details