Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 56536869-56537381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLPP3
Entrez Gene ID
8613
Gene Name
phospholipid phosphatase 3
Gene Aliases
Dri42, LPP3, PAP2B, PPAP2B, VCIP
UniGene
Hs.405156
Gene Chromosome Location
Chr.1: 56494747-56579584 on Build GRCh38
Transcripts
NM_003713.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5352878 c.256G>A p.A86T Chr.1: 56536996-56536996 on Build GRCh38
911058 c.294C>T p.L98L Chr.1: 56536958-56536958 on Build GRCh38
911059 c.181C>T p.P61S Chr.1: 56537071-56537071 on Build GRCh38
1172160 c.177C>G p.I59M Chr.1: 56537075-56537075 on Build GRCh38
3689772 c.297G>A p.A99A Chr.1: 56536955-56536955 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779042090 Chr.1: 56536934-56536934 on Build GRCh38
rs1019300877 Chr.1: 56537318-56537318 on Build GRCh38
rs202156325 Chr.1: 56537090-56537090 on Build GRCh38
rs952554394 Chr.1: 56537209-56537209 on Build GRCh38
rs984458994 Chr.1: 56537233-56537233 on Build GRCh38
rs373930737 Chr.1: 56537163-56537163 on Build GRCh38
rs763134504 Chr.1: 56537144-56537144 on Build GRCh38
rs761786683 Chr.1: 56537138-56537138 on Build GRCh38
rs546276456 Chr.1: 56537188-56537188 on Build GRCh38
rs552146004 Chr.1: 56537346-56537346 on Build GRCh38
rs975156918 Chr.1: 56537172-56537172 on Build GRCh38
rs938100306 Chr.1: 56537296-56537296 on Build GRCh38
rs748400164 Chr.1: 56536981-56536981 on Build GRCh38
rs189629519 Chr.1: 56537312-56537312 on Build GRCh38
rs1042686283 Chr.1: 56537310-56537310 on Build GRCh38
rs1055174222 Chr.1: 56537298-56537298 on Build GRCh38
rs1011044421 Chr.1: 56537307-56537307 on Build GRCh38
rs761031542 Chr.1: 56537151-56537151 on Build GRCh38
rs1136164 Chr.1: 56536955-56536955 on Build GRCh38
rs778173202 Chr.1: 56537111-56537111 on Build GRCh38
rs566232383 Chr.1: 56537309-56537309 on Build GRCh38
rs558272436 Chr.1: 56537189-56537189 on Build GRCh38
rs779476654 Chr.1: 56536979-56536979 on Build GRCh38
rs773051075 Chr.1: 56537143-56537143 on Build GRCh38
rs998558022 Chr.1: 56537317-56537317 on Build GRCh38
rs113672032 Chr.1: 56537299-56537299 on Build GRCh38
rs762846625 Chr.1: 56536943-56536943 on Build GRCh38
rs776212231 Chr.1: 56536927-56536927 on Build GRCh38
rs761474304 Chr.1: 56537041-56537041 on Build GRCh38
rs758742108 Chr.1: 56537108-56537108 on Build GRCh38
rs751344686 Chr.1: 56536947-56536947 on Build GRCh38
rs751372035 Chr.1: 56537030-56537030 on Build GRCh38
rs375912953 Chr.1: 56537095-56537095 on Build GRCh38
rs755552931 Chr.1: 56537096-56537096 on Build GRCh38
rs990611186 Chr.1: 56537165-56537165 on Build GRCh38
rs747539075 Chr.1: 56537235-56537235 on Build GRCh38
rs769334962 Chr.1: 56536999-56536999 on Build GRCh38
rs138524119 Chr.1: 56536997-56536997 on Build GRCh38
rs190738006 Chr.1: 56536956-56536956 on Build GRCh38
rs141659216 Chr.1: 56536973-56536973 on Build GRCh38
rs186446143 Chr.1: 56537128-56537128 on Build GRCh38
rs753968503 Chr.1: 56537154-56537154 on Build GRCh38
rs755259167 Chr.1: 56536924-56536924 on Build GRCh38
rs908453523 Chr.1: 56537234-56537234 on Build GRCh38
rs756098207 Chr.1: 56536967-56536967 on Build GRCh38
rs745573789 Chr.1: 56537120-56537120 on Build GRCh38
rs372686213 Chr.1: 56537278-56537278 on Build GRCh38
rs774976569 Chr.1: 56537006-56537006 on Build GRCh38
rs937822303 Chr.1: 56537152-56537153 on Build GRCh38
rs762937062 Chr.1: 56537019-56537019 on Build GRCh38
rs748732333 Chr.1: 56537129-56537129 on Build GRCh38
rs753188621 Chr.1: 56537162-56537162 on Build GRCh38
rs982368598 Chr.1: 56536998-56536998 on Build GRCh38
rs977069098 Chr.1: 56537069-56537069 on Build GRCh38
rs765388648 Chr.1: 56537156-56537156 on Build GRCh38
rs756324540 Chr.1: 56537084-56537084 on Build GRCh38
rs767802099 Chr.1: 56537064-56537064 on Build GRCh38
rs934899704 Chr.1: 56537319-56537319 on Build GRCh38
rs71639728 Chr.1: 56536901-56536901 on Build GRCh38
rs564827708 Chr.1: 56536982-56536982 on Build GRCh38
rs748570742 Chr.1: 56537105-56537105 on Build GRCh38
rs199699327 Chr.1: 56536994-56536994 on Build GRCh38
rs763944383 Chr.1: 56536945-56536945 on Build GRCh38
rs765247011 Chr.1: 56536935-56536935 on Build GRCh38
rs774105766 Chr.1: 56537137-56537137 on Build GRCh38
rs112576866 Chr.1: 56536898-56536898 on Build GRCh38
rs368859845 Chr.1: 56536925-56536925 on Build GRCh38
rs755063128 Chr.1: 56537155-56537155 on Build GRCh38
rs143533558 Chr.1: 56536972-56536972 on Build GRCh38
rs765299795 Chr.1: 56536938-56536939 on Build GRCh38
rs77470038 Chr.1: 56537356-56537356 on Build GRCh38
rs11583161 Chr.1: 56536941-56536941 on Build GRCh38
rs780501647 Chr.1: 56537038-56537038 on Build GRCh38
rs760439756 Chr.1: 56536916-56536916 on Build GRCh38
rs779524153 Chr.1: 56537102-56537102 on Build GRCh38
rs766647732 Chr.1: 56537153-56537153 on Build GRCh38
rs940048012 Chr.1: 56537238-56537238 on Build GRCh38
rs201459952 Chr.1: 56537093-56537093 on Build GRCh38
rs997018927 Chr.1: 56537355-56537355 on Build GRCh38
rs571956925 Chr.1: 56537349-56537349 on Build GRCh38
rs763962988 Chr.1: 56537020-56537020 on Build GRCh38
rs750660535 Chr.1: 56537081-56537081 on Build GRCh38
rs371900222 Chr.1: 56537143-56537144 on Build GRCh38
rs780258727 Chr.1: 56536971-56536971 on Build GRCh38
rs768609512 Chr.1: 56537134-56537134 on Build GRCh38
rs769423161 Chr.1: 56537124-56537124 on Build GRCh38
rs575809374 Chr.1: 56537154-56537154 on Build GRCh38
rs750545999 Chr.1: 56536958-56536958 on Build GRCh38
rs749718381 Chr.1: 56537061-56537061 on Build GRCh38
rs767254995 Chr.1: 56537140-56537140 on Build GRCh38
rs867537988 Chr.1: 56536988-56536988 on Build GRCh38
rs1046378829 Chr.1: 56537277-56537277 on Build GRCh38

View Full Product Details