Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 131502819-131503320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POMT1
Entrez Gene ID
10585
Gene Name
protein O-mannosyltransferase 1
Gene Aliases
LGMD2K, MDDGA1, MDDGB1, MDDGC1, RT
UniGene
Hs.522449
Gene Chromosome Location
Chr.9: 131502895-131523806 on Build GRCh38
Transcripts
NM_001077365.1 NM_001077366.1 NM_001136113.1 NM_001136114.1 NM_007171.3 XM_005272156.1 XM_005272158.1 XM_005272159.1 XM_005272162.2 XM_006716932.1 XM_011518140.1 XM_011518141.1 XM_011518142.1 XM_011518143.1 XM_011518145.1 XM_017014203.1 XM_017014204.1 XM_017014205.1

SNP Detail

SNP ID Chromosome Location
rs545232287 Chr.9: 131503253-131503253 on Build GRCh38
rs1026761122 Chr.9: 131503201-131503223 on Build GRCh38
rs1038249427 Chr.9: 131503254-131503254 on Build GRCh38
rs1051274577 Chr.9: 131502844-131502844 on Build GRCh38
rs1014741489 Chr.9: 131503043-131503043 on Build GRCh38
rs77198558 Chr.9: 131502856-131502856 on Build GRCh38
rs946338764 Chr.9: 131503012-131503012 on Build GRCh38
rs1014877936 Chr.9: 131502889-131502889 on Build GRCh38
rs959260180 Chr.9: 131502879-131502879 on Build GRCh38
rs569784340 Chr.9: 131503033-131503033 on Build GRCh38
rs774409088 Chr.9: 131502863-131502863 on Build GRCh38
rs951875526 Chr.9: 131502873-131502873 on Build GRCh38
rs1016825908 Chr.9: 131503040-131503040 on Build GRCh38
rs1054608592 Chr.9: 131502986-131502986 on Build GRCh38
rs112668089 Chr.9: 131503228-131503228 on Build GRCh38
rs547473594 Chr.9: 131502854-131502854 on Build GRCh38
rs914826203 Chr.9: 131503155-131503155 on Build GRCh38
rs962673262 Chr.9: 131502908-131502908 on Build GRCh38
rs767018413 Chr.9: 131502900-131502900 on Build GRCh38
rs761681721 Chr.9: 131502864-131502864 on Build GRCh38
rs918691395 Chr.9: 131502837-131502837 on Build GRCh38
rs1042498134 Chr.9: 131502860-131502860 on Build GRCh38
rs1034892250 Chr.9: 131502867-131502867 on Build GRCh38
rs147928419 Chr.9: 131502855-131502855 on Build GRCh38
rs886063535 Chr.9: 131502908-131502908 on Build GRCh38
rs527286034 Chr.9: 131503298-131503299 on Build GRCh38
rs1012099495 Chr.9: 131503034-131503034 on Build GRCh38
rs1025673121 Chr.9: 131503079-131503079 on Build GRCh38
rs939031360 Chr.9: 131503221-131503221 on Build GRCh38
rs936874138 Chr.9: 131502949-131502949 on Build GRCh38
rs988289184 Chr.9: 131503175-131503175 on Build GRCh38
rs535874278 Chr.9: 131502850-131502850 on Build GRCh38
rs911414733 Chr.9: 131502964-131502964 on Build GRCh38
rs921514117 Chr.9: 131503224-131503224 on Build GRCh38
rs964148135 Chr.9: 131503136-131503136 on Build GRCh38
rs923652344 Chr.9: 131503217-131503217 on Build GRCh38
rs1013472268 Chr.9: 131503110-131503110 on Build GRCh38
rs1050226535 Chr.9: 131503024-131503024 on Build GRCh38
rs994122166 Chr.9: 131503292-131503292 on Build GRCh38
rs753399075 Chr.9: 131503122-131503122 on Build GRCh38
rs569882668 Chr.9: 131502987-131502987 on Build GRCh38
rs760343191 Chr.9: 131502956-131502956 on Build GRCh38
rs972765704 Chr.9: 131502909-131502909 on Build GRCh38
rs1012192928 Chr.9: 131502890-131502890 on Build GRCh38
rs962660797 Chr.9: 131503042-131503042 on Build GRCh38
rs1038659227 Chr.9: 131502852-131502852 on Build GRCh38
rs368285841 Chr.9: 131503247-131503247 on Build GRCh38
rs911467205 Chr.9: 131502847-131502847 on Build GRCh38
rs549553345 Chr.9: 131502838-131502838 on Build GRCh38
rs1025396862 Chr.9: 131503120-131503120 on Build GRCh38
rs556267289 Chr.9: 131502936-131502936 on Build GRCh38
rs978146051 Chr.9: 131503182-131503182 on Build GRCh38
rs990281628 Chr.9: 131503152-131503152 on Build GRCh38
rs992178442 Chr.9: 131502905-131502905 on Build GRCh38
rs987362341 Chr.9: 131502953-131502953 on Build GRCh38
rs866051458 Chr.9: 131502977-131502977 on Build GRCh38
rs370627095 Chr.9: 131502848-131502848 on Build GRCh38
rs942890836 Chr.9: 131502976-131502976 on Build GRCh38
rs911321063 Chr.9: 131503134-131503134 on Build GRCh38
rs946355969 Chr.9: 131503162-131503162 on Build GRCh38
rs1003320463 Chr.9: 131503022-131503022 on Build GRCh38
rs994571945 Chr.9: 131503050-131503050 on Build GRCh38
rs976991632 Chr.9: 131503218-131503218 on Build GRCh38
rs1050494901 Chr.9: 131503243-131503243 on Build GRCh38
rs917895569 Chr.9: 131502916-131502916 on Build GRCh38
rs914180612 Chr.9: 131502969-131502969 on Build GRCh38
rs1006057334 Chr.9: 131502886-131502886 on Build GRCh38
rs973204721 Chr.9: 131502836-131502836 on Build GRCh38
rs914045602 Chr.9: 131503185-131503185 on Build GRCh38
rs898398709 Chr.9: 131503255-131503255 on Build GRCh38
rs1042095844 Chr.9: 131503015-131503015 on Build GRCh38
rs535602204 Chr.9: 131503089-131503089 on Build GRCh38
rs550740786 Chr.9: 131503021-131503021 on Build GRCh38
rs112631832 Chr.9: 131503109-131503109 on Build GRCh38
rs1003786907 Chr.9: 131502866-131502866 on Build GRCh38
rs189805189 Chr.9: 131503030-131503030 on Build GRCh38
rs996332506 Chr.9: 131503023-131503023 on Build GRCh38
rs918627535 Chr.9: 131502927-131502927 on Build GRCh38
rs886063536 Chr.9: 131502909-131502909 on Build GRCh38
rs1026000781 Chr.9: 131503300-131503300 on Build GRCh38
rs568045531 Chr.9: 131502843-131502843 on Build GRCh38
rs563898078 Chr.9: 131503268-131503268 on Build GRCh38
rs931701713 Chr.9: 131503240-131503240 on Build GRCh38

View Full Product Details