Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132896536-132897035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSC1
Entrez Gene ID
7248
Gene Name
tuberous sclerosis 1
Gene Aliases
LAM, TSC
UniGene
Hs.370854
Gene Chromosome Location
Chr.9: 132891348-132945269 on Build GRCh38
Transcripts
NM_000368.4 NM_001162426.1 NM_001162427.1 XM_005272211.1 XM_006717271.1 XM_011518979.2 XM_017015096.1 XM_017015097.1 XM_017015098.1 XM_017015099.1 XM_017015100.1 XM_017015101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6941009 c.3025G>A p.E1009K Chr.9: 132896705-132896705 on Build GRCh38
6962091 c.3154C>T p.P1052S Chr.9: 132896576-132896576 on Build GRCh38
264229 c.3184C>T p.R1062W Chr.9: 132896546-132896546 on Build GRCh38
6942526 c.3062C>T p.P1021L Chr.9: 132896668-132896668 on Build GRCh38
6353943 c.3016G>C p.G1006R Chr.9: 132896714-132896714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958224622 Chr.9: 132897011-132897011 on Build GRCh38
rs182885884 Chr.9: 132896894-132896894 on Build GRCh38
rs138445573 Chr.9: 132896663-132896663 on Build GRCh38
rs764979889 Chr.9: 132896696-132896696 on Build GRCh38
rs587778725 Chr.9: 132896559-132896562 on Build GRCh38
rs906256524 Chr.9: 132896779-132896779 on Build GRCh38
rs768443391 Chr.9: 132896618-132896618 on Build GRCh38
rs786203900 Chr.9: 132896682-132896682 on Build GRCh38
rs764503795 Chr.9: 132896798-132896798 on Build GRCh38
rs2234980 Chr.9: 132896601-132896603 on Build GRCh38
rs1046316073 Chr.9: 132896935-132896935 on Build GRCh38
rs148931779 Chr.9: 132896605-132896605 on Build GRCh38
rs202121327 Chr.9: 132896725-132896725 on Build GRCh38
rs747162992 Chr.9: 132896597-132896597 on Build GRCh38
rs796053461 Chr.9: 132896650-132896650 on Build GRCh38
rs552217429 Chr.9: 132896730-132896730 on Build GRCh38
rs796053449 Chr.9: 132896698-132896698 on Build GRCh38
rs397514866 Chr.9: 132896772-132896772 on Build GRCh38
rs759531937 Chr.9: 132896601-132896609 on Build GRCh38
rs199919348 Chr.9: 132896748-132896748 on Build GRCh38
rs777361506 Chr.9: 132896580-132896580 on Build GRCh38
rs7853849 Chr.9: 132896797-132896797 on Build GRCh38
rs1029761165 Chr.9: 132896983-132896983 on Build GRCh38
rs118203740 Chr.9: 132896808-132896808 on Build GRCh38
rs762149493 Chr.9: 132896779-132896781 on Build GRCh38
rs200205551 Chr.9: 132896803-132896803 on Build GRCh38
rs201192125 Chr.9: 132896601-132896601 on Build GRCh38
rs765033133 Chr.9: 132896787-132896787 on Build GRCh38
rs112066743 Chr.9: 132896558-132896558 on Build GRCh38
rs771810884 Chr.9: 132896743-132896743 on Build GRCh38
rs923543734 Chr.9: 132896736-132896736 on Build GRCh38
rs985885203 Chr.9: 132896999-132896999 on Build GRCh38
rs201192408 Chr.9: 132896788-132896788 on Build GRCh38
rs753374839 Chr.9: 132896607-132896607 on Build GRCh38
rs144314195 Chr.9: 132896746-132896746 on Build GRCh38
rs118203743 Chr.9: 132896600-132896601 on Build GRCh38
rs190854121 Chr.9: 132896842-132896842 on Build GRCh38
rs587778726 Chr.9: 132896590-132896590 on Build GRCh38
rs952963572 Chr.9: 132896621-132896621 on Build GRCh38
rs998317480 Chr.9: 132896972-132896972 on Build GRCh38
rs961534301 Chr.9: 132896858-132896858 on Build GRCh38
rs587778727 Chr.9: 132896561-132896561 on Build GRCh38
rs533565295 Chr.9: 132896654-132896654 on Build GRCh38
rs375394001 Chr.9: 132896651-132896651 on Build GRCh38
rs759106092 Chr.9: 132896770-132896770 on Build GRCh38
rs142954164 Chr.9: 132896706-132896706 on Build GRCh38
rs747142754 Chr.9: 132896944-132896944 on Build GRCh38
rs775387289 Chr.9: 132896706-132896707 on Build GRCh38
rs796053450 Chr.9: 132896644-132896644 on Build GRCh38
rs769722545 Chr.9: 132896601-132896606 on Build GRCh38
rs545896754 Chr.9: 132896857-132896857 on Build GRCh38
rs1034398735 Chr.9: 132896629-132896629 on Build GRCh38
rs1036192689 Chr.9: 132896851-132896851 on Build GRCh38
rs141008354 Chr.9: 132896989-132896989 on Build GRCh38
rs757370776 Chr.9: 132896719-132896719 on Build GRCh38
rs371879917 Chr.9: 132896720-132896720 on Build GRCh38
rs772043928 Chr.9: 132896639-132896639 on Build GRCh38
rs780224196 Chr.9: 132896735-132896735 on Build GRCh38
rs397514831 Chr.9: 132896583-132896583 on Build GRCh38
rs778413037 Chr.9: 132896571-132896571 on Build GRCh38
rs751970451 Chr.9: 132896718-132896718 on Build GRCh38
rs574783200 Chr.9: 132896804-132896804 on Build GRCh38
rs118203741 Chr.9: 132896700-132896700 on Build GRCh38
rs922638686 Chr.9: 132896680-132896680 on Build GRCh38
rs774196458 Chr.9: 132896623-132896623 on Build GRCh38
rs397514812 Chr.9: 132896601-132896603 on Build GRCh38
rs765753157 Chr.9: 132896655-132896655 on Build GRCh38
rs913722609 Chr.9: 132896764-132896764 on Build GRCh38
rs796053462 Chr.9: 132896600-132896600 on Build GRCh38
rs892228756 Chr.9: 132896589-132896589 on Build GRCh38
rs764738792 Chr.9: 132896711-132896711 on Build GRCh38
rs759047948 Chr.9: 132896685-132896685 on Build GRCh38
rs879079312 Chr.9: 132896913-132896913 on Build GRCh38
rs1005786246 Chr.9: 132896832-132896832 on Build GRCh38
rs368652971 Chr.9: 132896768-132896768 on Build GRCh38
rs538228068 Chr.9: 132897005-132897005 on Build GRCh38
rs140303368 Chr.9: 132896892-132896893 on Build GRCh38
rs554053175 Chr.9: 132896809-132896809 on Build GRCh38
rs1020905743 Chr.9: 132896684-132896684 on Build GRCh38
rs756384645 Chr.9: 132896732-132896732 on Build GRCh38
rs910475358 Chr.9: 132896712-132896712 on Build GRCh38
rs368729348 Chr.9: 132896900-132896902 on Build GRCh38
rs118203742 Chr.9: 132896627-132896627 on Build GRCh38
rs78868727 Chr.9: 132896750-132896750 on Build GRCh38
rs566430298 Chr.9: 132896653-132896653 on Build GRCh38
rs1015742388 Chr.9: 132896631-132896631 on Build GRCh38
rs778147394 Chr.9: 132896866-132896866 on Build GRCh38

View Full Product Details