Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153982934-153983442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM187
Entrez Gene ID
8269
Gene Name
transmembrane protein 187
Gene Aliases
CXorf12, DXS9878E, ITBA1
UniGene
Hs.23119
Gene Chromosome Location
Chr.X: 153972540-153983195 on Build GRCh38
Transcripts
NM_003492.2 XM_011531199.2

SNP Detail

SNP ID Chromosome Location
rs782503780 Chr.X: 153983169-153983169 on Build GRCh38
rs886867856 Chr.X: 153983153-153983153 on Build GRCh38
rs112136625 Chr.X: 153983356-153983356 on Build GRCh38
rs192974779 Chr.X: 153983067-153983067 on Build GRCh38
rs934633512 Chr.X: 153982997-153982997 on Build GRCh38
rs367685521 Chr.X: 153983135-153983135 on Build GRCh38
rs914549034 Chr.X: 153983403-153983403 on Build GRCh38
rs1045938 Chr.X: 153982959-153982959 on Build GRCh38
rs185209090 Chr.X: 153983404-153983404 on Build GRCh38
rs782330769 Chr.X: 153983284-153983284 on Build GRCh38
rs1003846302 Chr.X: 153983192-153983192 on Build GRCh38
rs782679380 Chr.X: 153983294-153983294 on Build GRCh38
rs1016611193 Chr.X: 153983197-153983197 on Build GRCh38
rs2239471 Chr.X: 153983048-153983048 on Build GRCh38
rs924664848 Chr.X: 153983329-153983329 on Build GRCh38
rs928920593 Chr.X: 153983417-153983417 on Build GRCh38
rs781834766 Chr.X: 153983006-153983006 on Build GRCh38
rs781889069 Chr.X: 153982989-153982989 on Build GRCh38
rs1034741867 Chr.X: 153983144-153983144 on Build GRCh38
rs986609904 Chr.X: 153983320-153983320 on Build GRCh38
rs1045025546 Chr.X: 153983414-153983414 on Build GRCh38
rs782274595 Chr.X: 153983332-153983332 on Build GRCh38
rs782556935 Chr.X: 153983109-153983109 on Build GRCh38
rs782119718 Chr.X: 153982988-153982988 on Build GRCh38
rs782701992 Chr.X: 153983065-153983065 on Build GRCh38
rs782213418 Chr.X: 153983274-153983274 on Build GRCh38
rs986830870 Chr.X: 153982977-153982977 on Build GRCh38
rs782554456 Chr.X: 153983038-153983038 on Build GRCh38
rs781871985 Chr.X: 153983148-153983148 on Build GRCh38
rs782483135 Chr.X: 153983287-153983287 on Build GRCh38
rs782775548 Chr.X: 153983000-153983000 on Build GRCh38
rs148305449 Chr.X: 153983217-153983217 on Build GRCh38
rs975724329 Chr.X: 153983257-153983257 on Build GRCh38

View Full Product Details