Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 2976405-2976898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFY
Entrez Gene ID
7544
Gene Name
zinc finger protein, Y-linked
Gene Aliases
ZNF911
UniGene
Hs.522845
Gene Chromosome Location
Chr.Y: 2934402-2982508 on Build GRCh38
Transcripts
NM_001145275.1 NM_001145276.1 NM_003411.3 XM_005262570.2 XM_011531473.2 XM_011531474.2 XM_011531475.2 XM_017030075.1 XM_017030076.1 XM_017030077.1

SNP Detail

SNP ID Chromosome Location
rs769544474 Chr.Y: 2976641-2976643 on Build GRCh38
rs772545649 Chr.Y: 2976865-2976865 on Build GRCh38
rs762918674 Chr.Y: 2976735-2976737 on Build GRCh38
rs761719378 Chr.Y: 2976767-2976767 on Build GRCh38
rs917679818 Chr.Y: 2976541-2976541 on Build GRCh38
rs775252294 Chr.Y: 2976726-2976746 on Build GRCh38
rs745501204 Chr.Y: 2976644-2976644 on Build GRCh38
rs949024153 Chr.Y: 2976546-2976546 on Build GRCh38
rs763007327 Chr.Y: 2976671-2976671 on Build GRCh38
rs767205748 Chr.Y: 2976791-2976791 on Build GRCh38
rs972464634 Chr.Y: 2976573-2976573 on Build GRCh38
rs765311293 Chr.Y: 2976873-2976873 on Build GRCh38
rs745422187 Chr.Y: 2976442-2976442 on Build GRCh38
rs774126265 Chr.Y: 2976738-2976738 on Build GRCh38
rs769494123 Chr.Y: 2976654-2976654 on Build GRCh38
rs113686315 Chr.Y: 2976728-2976728 on Build GRCh38
rs776910426 Chr.Y: 2976841-2976841 on Build GRCh38
rs780944575 Chr.Y: 2976633-2976633 on Build GRCh38
rs775084099 Chr.Y: 2976662-2976662 on Build GRCh38
rs768599885 Chr.Y: 2976687-2976687 on Build GRCh38

View Full Product Details