Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 53805362-53805907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDH15
Entrez Gene ID
65217
Gene Name
protocadherin related 15
Gene Aliases
CDHR15, DFNB23, USH1F
UniGene
Hs.280209
Gene Chromosome Location
Chr.10: 53802771-55629182 on Build GRCh38
Transcripts
NM_001142771.1 NM_001142772.1 XM_017016572.1

SNP Detail

SNP ID Chromosome Location
rs576236721 Chr.10: 53805556-53805556 on Build GRCh38
rs752038610 Chr.10: 53805619-53805620 on Build GRCh38
rs139265171 Chr.10: 53805474-53805475 on Build GRCh38
rs762859242 Chr.10: 53805466-53805466 on Build GRCh38
rs944679527 Chr.10: 53805772-53805772 on Build GRCh38
rs547691243 Chr.10: 53805661-53805661 on Build GRCh38
rs188656860 Chr.10: 53805821-53805821 on Build GRCh38
rs971040856 Chr.10: 53805495-53805495 on Build GRCh38
rs560504011 Chr.10: 53805697-53805697 on Build GRCh38
rs562434578 Chr.10: 53805510-53805510 on Build GRCh38
rs552683305 Chr.10: 53805782-53805782 on Build GRCh38
rs749050863 Chr.10: 53805827-53805828 on Build GRCh38
rs936513265 Chr.10: 53805726-53805726 on Build GRCh38
rs900335634 Chr.10: 53805481-53805481 on Build GRCh38
rs1005113552 Chr.10: 53805804-53805804 on Build GRCh38
rs184227993 Chr.10: 53805875-53805875 on Build GRCh38
rs1019298987 Chr.10: 53805869-53805870 on Build GRCh38
rs538441794 Chr.10: 53805796-53805796 on Build GRCh38
rs919684898 Chr.10: 53805580-53805580 on Build GRCh38
rs866798028 Chr.10: 53805428-53805428 on Build GRCh38
rs1004905350 Chr.10: 53805577-53805577 on Build GRCh38
rs1053816095 Chr.10: 53805421-53805422 on Build GRCh38
rs911831494 Chr.10: 53805441-53805441 on Build GRCh38
rs1000981269 Chr.10: 53805416-53805416 on Build GRCh38
rs548458404 Chr.10: 53805797-53805797 on Build GRCh38
rs541615966 Chr.10: 53805603-53805603 on Build GRCh38
rs994956839 Chr.10: 53805483-53805483 on Build GRCh38
rs989217555 Chr.10: 53805698-53805698 on Build GRCh38
rs201049227 Chr.10: 53805571-53805572 on Build GRCh38
rs1042595708 Chr.10: 53805638-53805638 on Build GRCh38
rs1049162697 Chr.10: 53805499-53805499 on Build GRCh38
rs886464332 Chr.10: 53805544-53805544 on Build GRCh38
rs911513667 Chr.10: 53805771-53805771 on Build GRCh38
rs569208331 Chr.10: 53805788-53805788 on Build GRCh38
rs914973480 Chr.10: 53805722-53805722 on Build GRCh38
rs1015607007 Chr.10: 53805844-53805844 on Build GRCh38
rs1002151845 Chr.10: 53805680-53805680 on Build GRCh38
rs1053763019 Chr.10: 53805735-53805735 on Build GRCh38
rs953811270 Chr.10: 53805864-53805865 on Build GRCh38
rs1015119147 Chr.10: 53805544-53805545 on Build GRCh38
rs966838635 Chr.10: 53805744-53805744 on Build GRCh38
rs74134791 Chr.10: 53805746-53805746 on Build GRCh38
rs527549957 Chr.10: 53805659-53805659 on Build GRCh38
rs564143196 Chr.10: 53805478-53805478 on Build GRCh38
rs761516428 Chr.10: 53805675-53805676 on Build GRCh38
rs191782955 Chr.10: 53805867-53805867 on Build GRCh38
rs1026734299 Chr.10: 53805859-53805860 on Build GRCh38
rs757309234 Chr.10: 53805428-53805429 on Build GRCh38
rs974650362 Chr.10: 53805776-53805776 on Build GRCh38
rs923773491 Chr.10: 53805649-53805649 on Build GRCh38
rs539936037 Chr.10: 53805854-53805858 on Build GRCh38
rs868320345 Chr.10: 53805817-53805817 on Build GRCh38
rs902582147 Chr.10: 53805411-53805411 on Build GRCh38
rs914940811 Chr.10: 53805392-53805392 on Build GRCh38
rs576408631 Chr.10: 53805437-53805437 on Build GRCh38
rs1046413734 Chr.10: 53805747-53805747 on Build GRCh38
rs895235895 Chr.10: 53805743-53805743 on Build GRCh38
rs531640542 Chr.10: 53805814-53805814 on Build GRCh38
rs114133702 Chr.10: 53805604-53805604 on Build GRCh38
rs74134790 Chr.10: 53805467-53805467 on Build GRCh38
rs1034517231 Chr.10: 53805695-53805695 on Build GRCh38
rs762759024 Chr.10: 53805846-53805847 on Build GRCh38

View Full Product Details