Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 58388579-58389077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFAM
Entrez Gene ID
7019
Gene Name
transcription factor A, mitochondrial
Gene Aliases
MTTF1, MTTFA, TCF6, TCF6L1, TCF6L2, TCF6L3
UniGene
Hs.594250
Gene Chromosome Location
Chr.10: 58385143-58399230 on Build GRCh38
Transcripts
NM_001270782.1 NM_003201.2 NR_073073.1 XM_011540120.2 XM_011540121.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6392776 c.431C>A p.T144K Chr.10: 58388809-58388809 on Build GRCh38
1348365 c.432delA p.E148fs*2 Chr.10: 58388810-58388810 on Build GRCh38
919152 c.375G>A p.Q125Q Chr.10: 58388753-58388753 on Build GRCh38
5807214 c.373C>T p.Q125* Chr.10: 58388751-58388751 on Build GRCh38
7326869 c.375_376insA p.T126fs*20 Chr.10: 58388809-58388810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2306604 Chr.10: 58388932-58388932 on Build GRCh38
rs745401132 Chr.10: 58388856-58388856 on Build GRCh38
rs140798898 Chr.10: 58389033-58389033 on Build GRCh38
rs1003979240 Chr.10: 58388998-58388998 on Build GRCh38
rs77418790 Chr.10: 58388704-58388704 on Build GRCh38
rs778549899 Chr.10: 58388712-58388712 on Build GRCh38
rs139514719 Chr.10: 58388693-58388693 on Build GRCh38
rs1013377990 Chr.10: 58389045-58389045 on Build GRCh38
rs757235180 Chr.10: 58388839-58388839 on Build GRCh38
rs755878506 Chr.10: 58388830-58388830 on Build GRCh38
rs16912188 Chr.10: 58388991-58388991 on Build GRCh38
rs147614456 Chr.10: 58389004-58389004 on Build GRCh38
rs373540254 Chr.10: 58389034-58389034 on Build GRCh38
rs377345031 Chr.10: 58388650-58388650 on Build GRCh38
rs372548468 Chr.10: 58388834-58388834 on Build GRCh38
rs373282082 Chr.10: 58388634-58388634 on Build GRCh38
rs557054190 Chr.10: 58388963-58388963 on Build GRCh38
rs767917663 Chr.10: 58388656-58388656 on Build GRCh38
rs1042083942 Chr.10: 58388886-58388886 on Build GRCh38
rs1055269607 Chr.10: 58389013-58389013 on Build GRCh38
rs753977902 Chr.10: 58388835-58388849 on Build GRCh38
rs1040250397 Chr.10: 58388607-58388607 on Build GRCh38
rs149704158 Chr.10: 58388700-58388700 on Build GRCh38
rs370074581 Chr.10: 58388623-58388623 on Build GRCh38
rs778603286 Chr.10: 58388855-58388855 on Build GRCh38
rs544132101 Chr.10: 58388810-58388810 on Build GRCh38
rs1025137755 Chr.10: 58389052-58389052 on Build GRCh38
rs775241728 Chr.10: 58388754-58388754 on Build GRCh38
rs756171504 Chr.10: 58388809-58388810 on Build GRCh38
rs755302480 Chr.10: 58388838-58388844 on Build GRCh38
rs753774417 Chr.10: 58388835-58388835 on Build GRCh38
rs946144276 Chr.10: 58388883-58388883 on Build GRCh38
rs771816601 Chr.10: 58388745-58388745 on Build GRCh38
rs997855966 Chr.10: 58388632-58388632 on Build GRCh38
rs756994866 Chr.10: 58388899-58388899 on Build GRCh38
rs1051539037 Chr.10: 58388950-58388950 on Build GRCh38
rs534877353 Chr.10: 58388783-58388783 on Build GRCh38
rs140210748 Chr.10: 58388809-58388809 on Build GRCh38
rs921363377 Chr.10: 58388918-58388918 on Build GRCh38
rs575228011 Chr.10: 58388986-58388986 on Build GRCh38
rs145518555 Chr.10: 58388744-58388744 on Build GRCh38
rs758607553 Chr.10: 58388692-58388692 on Build GRCh38
rs780284142 Chr.10: 58388810-58388811 on Build GRCh38
rs770411113 Chr.10: 58388709-58388709 on Build GRCh38
rs537283161 Chr.10: 58388691-58388691 on Build GRCh38
rs34084383 Chr.10: 58388655-58388656 on Build GRCh38
rs779865582 Chr.10: 58388870-58388870 on Build GRCh38
rs1016628795 Chr.10: 58388672-58388672 on Build GRCh38
rs762377043 Chr.10: 58388794-58388794 on Build GRCh38
rs749196414 Chr.10: 58388705-58388705 on Build GRCh38
rs772620246 Chr.10: 58388627-58388627 on Build GRCh38
rs777009100 Chr.10: 58388787-58388787 on Build GRCh38
rs375965328 Chr.10: 58388620-58388620 on Build GRCh38
rs867338263 Chr.10: 58389031-58389031 on Build GRCh38
rs570779709 Chr.10: 58388703-58388703 on Build GRCh38
rs760968829 Chr.10: 58388779-58388779 on Build GRCh38
rs760728446 Chr.10: 58388647-58388647 on Build GRCh38
rs151097728 Chr.10: 58388676-58388676 on Build GRCh38
rs765360369 Chr.10: 58388798-58388798 on Build GRCh38
rs370616747 Chr.10: 58388662-58388662 on Build GRCh38
rs936208947 Chr.10: 58388900-58388900 on Build GRCh38
rs751758908 Chr.10: 58388822-58388822 on Build GRCh38
rs868175464 Chr.10: 58388736-58388736 on Build GRCh38
rs539878663 Chr.10: 58388616-58388616 on Build GRCh38
rs764539268 Chr.10: 58388655-58388655 on Build GRCh38
rs959086371 Chr.10: 58388674-58388674 on Build GRCh38
rs553009666 Chr.10: 58388807-58388807 on Build GRCh38
rs78912196 Chr.10: 58388810-58388810 on Build GRCh38
rs764977211 Chr.10: 58388905-58388905 on Build GRCh38
rs974527181 Chr.10: 58388910-58388911 on Build GRCh38
rs960522124 Chr.10: 58388714-58388714 on Build GRCh38

View Full Product Details