Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 67098132-67098676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNA3
Entrez Gene ID
29119
Gene Name
catenin alpha 3
Gene Aliases
ARVD13, VR22
UniGene
Hs.21375
Gene Chromosome Location
Chr.10: 65912518-67696217 on Build GRCh38
Transcripts
NM_001127384.2 NM_013266.3 XM_017016151.1 XM_017016152.1 XM_017016153.1 XM_017016154.1 XM_017016155.1 XM_017016156.1 NM_178011.4 NR_111909.1

Gene Symbol
LRRTM3
Entrez Gene ID
347731
Gene Name
leucine rich repeat transmembrane neuronal 3
Gene Aliases
-
UniGene
Hs.652155
Gene Chromosome Location
Chr.10: 66926034-67101551 on Build GRCh38
Transcripts
NM_001127384.2 NM_013266.3 XM_017016151.1 XM_017016152.1 XM_017016153.1 XM_017016154.1 XM_017016155.1 XM_017016156.1 NM_178011.4 NR_111909.1

SNP Detail

SNP ID Chromosome Location
rs961397797 Chr.10: 67098354-67098354 on Build GRCh38
rs897367611 Chr.10: 67098636-67098636 on Build GRCh38
rs186527454 Chr.10: 67098524-67098524 on Build GRCh38
rs755182388 Chr.10: 67098418-67098418 on Build GRCh38
rs988409492 Chr.10: 67098558-67098558 on Build GRCh38
rs906026144 Chr.10: 67098619-67098619 on Build GRCh38
rs917017630 Chr.10: 67098584-67098584 on Build GRCh38
rs997197392 Chr.10: 67098581-67098581 on Build GRCh38
rs978518509 Chr.10: 67098395-67098395 on Build GRCh38
rs778875851 Chr.10: 67098501-67098501 on Build GRCh38
rs752754627 Chr.10: 67098333-67098333 on Build GRCh38
rs951050299 Chr.10: 67098250-67098250 on Build GRCh38
rs893012210 Chr.10: 67098480-67098481 on Build GRCh38
rs1038620458 Chr.10: 67098634-67098634 on Build GRCh38
rs895537189 Chr.10: 67098321-67098321 on Build GRCh38
rs1054342283 Chr.10: 67098465-67098465 on Build GRCh38
rs987321071 Chr.10: 67098427-67098427 on Build GRCh38
rs949391918 Chr.10: 67098376-67098376 on Build GRCh38
rs549419069 Chr.10: 67098580-67098580 on Build GRCh38
rs1044568921 Chr.10: 67098598-67098598 on Build GRCh38
rs1017305270 Chr.10: 67098191-67098191 on Build GRCh38
rs1029759362 Chr.10: 67098358-67098358 on Build GRCh38
rs1043145458 Chr.10: 67098198-67098198 on Build GRCh38
rs756272715 Chr.10: 67098506-67098506 on Build GRCh38
rs965021673 Chr.10: 67098454-67098454 on Build GRCh38
rs1006885324 Chr.10: 67098600-67098600 on Build GRCh38
rs934582044 Chr.10: 67098464-67098465 on Build GRCh38
rs529495108 Chr.10: 67098322-67098322 on Build GRCh38
rs1044521480 Chr.10: 67098323-67098323 on Build GRCh38
rs569572462 Chr.10: 67098585-67098585 on Build GRCh38
rs1012403976 Chr.10: 67098652-67098652 on Build GRCh38
rs1007267270 Chr.10: 67098177-67098177 on Build GRCh38
rs879248599 Chr.10: 67098527-67098527 on Build GRCh38
rs955413623 Chr.10: 67098363-67098363 on Build GRCh38
rs886069419 Chr.10: 67098589-67098589 on Build GRCh38
rs560959831 Chr.10: 67098481-67098484 on Build GRCh38
rs540200181 Chr.10: 67098168-67098168 on Build GRCh38
rs749262153 Chr.10: 67098355-67098355 on Build GRCh38
rs1052806988 Chr.10: 67098307-67098307 on Build GRCh38
rs943281423 Chr.10: 67098169-67098169 on Build GRCh38
rs1017382193 Chr.10: 67098345-67098345 on Build GRCh38
rs1005786331 Chr.10: 67098334-67098334 on Build GRCh38
rs978699144 Chr.10: 67098462-67098462 on Build GRCh38
rs925634505 Chr.10: 67098218-67098218 on Build GRCh38
rs1017435939 Chr.10: 67098435-67098435 on Build GRCh38
rs925825080 Chr.10: 67098512-67098512 on Build GRCh38
rs901550428 Chr.10: 67098197-67098197 on Build GRCh38
rs997125360 Chr.10: 67098245-67098245 on Build GRCh38
rs560133600 Chr.10: 67098268-67098268 on Build GRCh38
rs181916582 Chr.10: 67098336-67098336 on Build GRCh38
rs190579045 Chr.10: 67098621-67098621 on Build GRCh38

View Full Product Details