Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31576427-31576920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNF
Entrez Gene ID
7124
Gene Name
tumor necrosis factor
Gene Aliases
DIF, TNF-alpha, TNFA, TNFSF2, TNLG1F
UniGene
Hs.241570
Gene Chromosome Location
Chr.6: 31575567-31578336 on Build GRCh38
Transcripts
NM_000594.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7000215 c.195G>T p.R65S Chr.6: 31576542-31576542 on Build GRCh38
3783975 c.245G>A p.R82Q Chr.6: 31576779-31576779 on Build GRCh38
4885958 c.217C>G p.L73V Chr.6: 31576564-31576564 on Build GRCh38
4586599 c.251C>T p.P84L Chr.6: 31576785-31576785 on Build GRCh38
3777486 c.231C>T p.V77V Chr.6: 31576578-31576578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768947455 Chr.6: 31576523-31576523 on Build GRCh38
rs1800619 Chr.6: 31576729-31576729 on Build GRCh38
rs772270241 Chr.6: 31576779-31576779 on Build GRCh38
rs565005379 Chr.6: 31576840-31576840 on Build GRCh38
rs577329144 Chr.6: 31576455-31576455 on Build GRCh38
rs3730322 Chr.6: 31576641-31576641 on Build GRCh38
rs888924952 Chr.6: 31576877-31576877 on Build GRCh38
rs916067458 Chr.6: 31576680-31576680 on Build GRCh38
rs775365890 Chr.6: 31576579-31576579 on Build GRCh38
rs759609722 Chr.6: 31576552-31576552 on Build GRCh38
rs369783162 Chr.6: 31576772-31576772 on Build GRCh38
rs892418402 Chr.6: 31576789-31576789 on Build GRCh38
rs769643681 Chr.6: 31576572-31576572 on Build GRCh38
rs569914957 Chr.6: 31576446-31576446 on Build GRCh38
rs755342875 Chr.6: 31576746-31576746 on Build GRCh38
rs746433040 Chr.6: 31576504-31576504 on Build GRCh38
rs772331441 Chr.6: 31576539-31576539 on Build GRCh38
rs867719858 Chr.6: 31576651-31576651 on Build GRCh38
rs373391738 Chr.6: 31576605-31576605 on Build GRCh38
rs780350382 Chr.6: 31576731-31576731 on Build GRCh38
rs768383794 Chr.6: 31576855-31576855 on Build GRCh38
rs984990176 Chr.6: 31576663-31576663 on Build GRCh38
rs559043897 Chr.6: 31576454-31576454 on Build GRCh38
rs1023506640 Chr.6: 31576463-31576463 on Build GRCh38
rs967964242 Chr.6: 31576521-31576521 on Build GRCh38
rs756466639 Chr.6: 31576506-31576506 on Build GRCh38
rs761373748 Chr.6: 31576624-31576624 on Build GRCh38
rs776692566 Chr.6: 31576543-31576543 on Build GRCh38
rs766958740 Chr.6: 31576866-31576866 on Build GRCh38
rs750865758 Chr.6: 31576725-31576725 on Build GRCh38
rs56340803 Chr.6: 31576632-31576632 on Build GRCh38
rs1049796997 Chr.6: 31576722-31576722 on Build GRCh38
rs1800620 Chr.6: 31576814-31576814 on Build GRCh38
rs774257243 Chr.6: 31576530-31576530 on Build GRCh38
rs956294956 Chr.6: 31576565-31576565 on Build GRCh38
rs897338372 Chr.6: 31576782-31576782 on Build GRCh38
rs186267383 Chr.6: 31576631-31576631 on Build GRCh38
rs745738344 Chr.6: 31576786-31576786 on Build GRCh38
rs3093663 Chr.6: 31576448-31576448 on Build GRCh38
rs541437699 Chr.6: 31576459-31576459 on Build GRCh38
rs967910752 Chr.6: 31576883-31576883 on Build GRCh38
rs762461783 Chr.6: 31576585-31576585 on Build GRCh38
rs1056210947 Chr.6: 31576828-31576828 on Build GRCh38
rs972350853 Chr.6: 31576567-31576567 on Build GRCh38
rs1000808024 Chr.6: 31576897-31576897 on Build GRCh38
rs941620360 Chr.6: 31576751-31576751 on Build GRCh38
rs756628184 Chr.6: 31576730-31576730 on Build GRCh38
rs780604770 Chr.6: 31576609-31576610 on Build GRCh38
rs35131721 Chr.6: 31576538-31576538 on Build GRCh38
rs763838774 Chr.6: 31576590-31576590 on Build GRCh38
rs4645843 Chr.6: 31576785-31576785 on Build GRCh38
rs947804964 Chr.6: 31576710-31576710 on Build GRCh38
rs780403381 Chr.6: 31576507-31576507 on Build GRCh38
rs751100224 Chr.6: 31576492-31576492 on Build GRCh38
rs754263939 Chr.6: 31576732-31576732 on Build GRCh38
rs745792555 Chr.6: 31576748-31576748 on Build GRCh38
rs4645844 Chr.6: 31576875-31576875 on Build GRCh38
rs3093664 Chr.6: 31576865-31576865 on Build GRCh38
rs757664880 Chr.6: 31576500-31576500 on Build GRCh38
rs751210763 Chr.6: 31576620-31576620 on Build GRCh38
rs775106258 Chr.6: 31576822-31576822 on Build GRCh38
rs769637264 Chr.6: 31576822-31576823 on Build GRCh38
rs768258025 Chr.6: 31576619-31576619 on Build GRCh38
rs779177870 Chr.6: 31576759-31576759 on Build GRCh38
rs55741465 Chr.6: 31576647-31576647 on Build GRCh38
rs749414660 Chr.6: 31576522-31576522 on Build GRCh38
rs563535896 Chr.6: 31576677-31576677 on Build GRCh38
rs764984097 Chr.6: 31576864-31576864 on Build GRCh38
rs781640653 Chr.6: 31576503-31576503 on Build GRCh38
rs190788828 Chr.6: 31576794-31576794 on Build GRCh38

View Full Product Details