Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31577430-31577931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNF
Entrez Gene ID
7124
Gene Name
tumor necrosis factor
Gene Aliases
DIF, TNF-alpha, TNFA, TNFSF2, TNLG1F
UniGene
Hs.241570
Gene Chromosome Location
Chr.6: 31575567-31578336 on Build GRCh38
Transcripts
NM_000594.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4734843 c.634A>T p.I212F Chr.6: 31577469-31577469 on Build GRCh38
7339816 c.609G>T p.E203D Chr.6: 31577444-31577444 on Build GRCh38
3624582 c.690C>T p.I230I Chr.6: 31577525-31577525 on Build GRCh38
5935212 c.655G>A p.D219N Chr.6: 31577490-31577490 on Build GRCh38
5038047 c.601C>T p.Q201* Chr.6: 31577436-31577436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190947828 Chr.6: 31577541-31577541 on Build GRCh38
rs777492465 Chr.6: 31577548-31577548 on Build GRCh38
rs760929534 Chr.6: 31577870-31577870 on Build GRCh38
rs1043627155 Chr.6: 31577622-31577622 on Build GRCh38
rs750502134 Chr.6: 31577459-31577459 on Build GRCh38
rs918829488 Chr.6: 31577730-31577730 on Build GRCh38
rs767411749 Chr.6: 31577725-31577725 on Build GRCh38
rs760959992 Chr.6: 31577619-31577619 on Build GRCh38
rs766515527 Chr.6: 31577542-31577542 on Build GRCh38
rs557263022 Chr.6: 31577566-31577566 on Build GRCh38
rs774879769 Chr.6: 31577498-31577498 on Build GRCh38
rs769320071 Chr.6: 31577597-31577597 on Build GRCh38
rs756986997 Chr.6: 31577697-31577697 on Build GRCh38
rs763621932 Chr.6: 31577485-31577485 on Build GRCh38
rs892370128 Chr.6: 31577884-31577884 on Build GRCh38
rs10947218 Chr.6: 31577854-31577854 on Build GRCh38
rs752568182 Chr.6: 31577544-31577544 on Build GRCh38
rs1052552748 Chr.6: 31577879-31577879 on Build GRCh38
rs763152978 Chr.6: 31577679-31577679 on Build GRCh38
rs778803751 Chr.6: 31577463-31577463 on Build GRCh38
rs956203592 Chr.6: 31577895-31577895 on Build GRCh38
rs766384108 Chr.6: 31577639-31577639 on Build GRCh38
rs981945133 Chr.6: 31577617-31577617 on Build GRCh38
rs1006268320 Chr.6: 31577847-31577847 on Build GRCh38
rs770509340 Chr.6: 31577476-31577476 on Build GRCh38
rs769422678 Chr.6: 31577589-31577589 on Build GRCh38
rs748145803 Chr.6: 31577465-31577465 on Build GRCh38
rs753952568 Chr.6: 31577708-31577708 on Build GRCh38
rs745590537 Chr.6: 31577581-31577581 on Build GRCh38
rs887782506 Chr.6: 31577798-31577798 on Build GRCh38
rs780684380 Chr.6: 31577578-31577578 on Build GRCh38
rs186486397 Chr.6: 31577511-31577511 on Build GRCh38
rs762056461 Chr.6: 31577504-31577504 on Build GRCh38
rs763985399 Chr.6: 31577665-31577665 on Build GRCh38
rs751562173 Chr.6: 31577683-31577683 on Build GRCh38
rs780711335 Chr.6: 31577480-31577480 on Build GRCh38
rs1044801984 Chr.6: 31577792-31577792 on Build GRCh38
rs1030144612 Chr.6: 31577905-31577905 on Build GRCh38
rs918104667 Chr.6: 31577763-31577763 on Build GRCh38
rs541176107 Chr.6: 31577886-31577886 on Build GRCh38
rs987884598 Chr.6: 31577603-31577603 on Build GRCh38
rs775041173 Chr.6: 31577600-31577600 on Build GRCh38
rs777545571 Chr.6: 31577467-31577467 on Build GRCh38
rs985558695 Chr.6: 31577594-31577594 on Build GRCh38
rs749764028 Chr.6: 31577596-31577596 on Build GRCh38
rs575712826 Chr.6: 31577593-31577593 on Build GRCh38
rs370893734 Chr.6: 31577481-31577481 on Build GRCh38
rs750709123 Chr.6: 31577540-31577540 on Build GRCh38
rs936347053 Chr.6: 31577855-31577855 on Build GRCh38
rs962747745 Chr.6: 31577681-31577681 on Build GRCh38
rs767604552 Chr.6: 31577513-31577513 on Build GRCh38
rs3093665 Chr.6: 31577614-31577614 on Build GRCh38
rs756260984 Chr.6: 31577462-31577462 on Build GRCh38
rs769084971 Chr.6: 31577489-31577489 on Build GRCh38
rs762064944 Chr.6: 31577607-31577607 on Build GRCh38
rs4645845 Chr.6: 31577771-31577771 on Build GRCh38
rs943289567 Chr.6: 31577642-31577642 on Build GRCh38
rs141307820 Chr.6: 31577475-31577475 on Build GRCh38
rs753933440 Chr.6: 31577654-31577654 on Build GRCh38
rs376368223 Chr.6: 31577455-31577455 on Build GRCh38
rs758342189 Chr.6: 31577545-31577545 on Build GRCh38
rs762834115 Chr.6: 31577655-31577655 on Build GRCh38
rs757409861 Chr.6: 31577761-31577761 on Build GRCh38
rs535723784 Chr.6: 31577555-31577555 on Build GRCh38
rs758189183 Chr.6: 31577466-31577466 on Build GRCh38
rs372252268 Chr.6: 31577565-31577565 on Build GRCh38
rs930252458 Chr.6: 31577748-31577748 on Build GRCh38
rs901656209 Chr.6: 31577710-31577710 on Build GRCh38
rs997855018 Chr.6: 31577892-31577892 on Build GRCh38
rs772549106 Chr.6: 31577609-31577609 on Build GRCh38

View Full Product Details