Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 78138824-78139328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF9B
Entrez Gene ID
51616
Gene Name
TATA-box binding protein associated factor 9b
Gene Aliases
DN-7, DN7, TAF9L, TAFII31L, TFIID-31
UniGene
Hs.592248
Gene Chromosome Location
Chr.X: 78129748-78139682 on Build GRCh38
Transcripts
NM_015975.4

SNP Detail

SNP ID Chromosome Location
rs1040867137 Chr.X: 78138950-78138950 on Build GRCh38
rs782287314 Chr.X: 78138902-78138902 on Build GRCh38
rs782365505 Chr.X: 78139176-78139176 on Build GRCh38
rs367634846 Chr.X: 78139182-78139182 on Build GRCh38
rs952336365 Chr.X: 78139280-78139280 on Build GRCh38
rs782334180 Chr.X: 78138877-78138877 on Build GRCh38
rs931989047 Chr.X: 78139021-78139021 on Build GRCh38
rs940829444 Chr.X: 78138946-78138946 on Build GRCh38
rs782234816 Chr.X: 78139090-78139090 on Build GRCh38
rs781909952 Chr.X: 78138877-78138878 on Build GRCh38
rs782228385 Chr.X: 78138989-78138989 on Build GRCh38
rs923353056 Chr.X: 78138991-78138991 on Build GRCh38
rs1001852250 Chr.X: 78139205-78139205 on Build GRCh38
rs782473640 Chr.X: 78138953-78138953 on Build GRCh38
rs782620091 Chr.X: 78138885-78138885 on Build GRCh38
rs781946132 Chr.X: 78139114-78139114 on Build GRCh38
rs143257195 Chr.X: 78138870-78138870 on Build GRCh38
rs1052319059 Chr.X: 78139097-78139097 on Build GRCh38
rs371088100 Chr.X: 78138957-78138957 on Build GRCh38
rs781964968 Chr.X: 78138882-78138882 on Build GRCh38
rs190185473 Chr.X: 78139035-78139035 on Build GRCh38
rs782462790 Chr.X: 78139015-78139015 on Build GRCh38
rs782702243 Chr.X: 78138927-78138927 on Build GRCh38
rs781806254 Chr.X: 78138930-78138930 on Build GRCh38
rs1026599684 Chr.X: 78139278-78139278 on Build GRCh38
rs781893699 Chr.X: 78138903-78138903 on Build GRCh38
rs892299095 Chr.X: 78139102-78139102 on Build GRCh38
rs782422204 Chr.X: 78138858-78138858 on Build GRCh38
rs782136603 Chr.X: 78139236-78139236 on Build GRCh38
rs782060151 Chr.X: 78138874-78138874 on Build GRCh38
rs1015142229 Chr.X: 78139299-78139299 on Build GRCh38
rs1050400177 Chr.X: 78139026-78139026 on Build GRCh38
rs45563641 Chr.X: 78139139-78139139 on Build GRCh38
rs936565634 Chr.X: 78139096-78139096 on Build GRCh38
rs1013389544 Chr.X: 78139113-78139113 on Build GRCh38
rs181604070 Chr.X: 78139095-78139095 on Build GRCh38
rs952283074 Chr.X: 78138910-78138910 on Build GRCh38
rs985448476 Chr.X: 78139293-78139293 on Build GRCh38
rs782117490 Chr.X: 78138884-78138884 on Build GRCh38
rs927570872 Chr.X: 78138893-78138893 on Build GRCh38
rs781961981 Chr.X: 78139198-78139198 on Build GRCh38
rs35062750 Chr.X: 78139262-78139263 on Build GRCh38
rs985009458 Chr.X: 78138915-78138915 on Build GRCh38
rs782173722 Chr.X: 78138909-78138909 on Build GRCh38

View Full Product Details