Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 149478895-149479453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDS
Entrez Gene ID
3423
Gene Name
iduronate 2-sulfatase
Gene Aliases
MPS2, SIDS
UniGene
Hs.460960
Gene Chromosome Location
Chr.X: 149476990-149505354 on Build GRCh38
Transcripts
NM_000202.7 NM_001166550.3

SNP Detail

SNP ID Chromosome Location
rs1010444845 Chr.X: 149479105-149479105 on Build GRCh38
rs1036822317 Chr.X: 149479042-149479042 on Build GRCh38
rs1010475871 Chr.X: 149479397-149479397 on Build GRCh38
rs1141636 Chr.X: 149479050-149479050 on Build GRCh38
rs1000863873 Chr.X: 149479090-149479090 on Build GRCh38
rs782717636 Chr.X: 149479312-149479312 on Build GRCh38
rs188442928 Chr.X: 149479111-149479111 on Build GRCh38
rs909713677 Chr.X: 149479008-149479008 on Build GRCh38
rs1033774284 Chr.X: 149479174-149479174 on Build GRCh38
rs892040967 Chr.X: 149479104-149479104 on Build GRCh38
rs922422108 Chr.X: 149479307-149479307 on Build GRCh38
rs909837485 Chr.X: 149479244-149479244 on Build GRCh38
rs5936291 Chr.X: 149479080-149479080 on Build GRCh38
rs5936292 Chr.X: 149479167-149479167 on Build GRCh38
rs180697990 Chr.X: 149479359-149479359 on Build GRCh38
rs984024424 Chr.X: 149478949-149478949 on Build GRCh38
rs781872764 Chr.X: 149479431-149479431 on Build GRCh38
rs782192065 Chr.X: 149479306-149479306 on Build GRCh38
rs200754199 Chr.X: 149479178-149479178 on Build GRCh38
rs201612383 Chr.X: 149479130-149479130 on Build GRCh38
rs782729444 Chr.X: 149479378-149479378 on Build GRCh38
rs782357554 Chr.X: 149479079-149479079 on Build GRCh38
rs201426579 Chr.X: 149479207-149479207 on Build GRCh38
rs201800308 Chr.X: 149479074-149479074 on Build GRCh38
rs934064366 Chr.X: 149479077-149479077 on Build GRCh38
rs781799353 Chr.X: 149479317-149479317 on Build GRCh38
rs939829586 Chr.X: 149479041-149479041 on Build GRCh38
rs1033227946 Chr.X: 149479097-149479097 on Build GRCh38
rs5936188 Chr.X: 149479235-149479235 on Build GRCh38
rs970490966 Chr.X: 149479141-149479141 on Build GRCh38
rs934095465 Chr.X: 149479311-149479311 on Build GRCh38
rs1054856974 Chr.X: 149479367-149479367 on Build GRCh38
rs199533893 Chr.X: 149479138-149479138 on Build GRCh38
rs35415606 Chr.X: 149479302-149479303 on Build GRCh38
rs961268000 Chr.X: 149479262-149479262 on Build GRCh38
rs911216508 Chr.X: 149479326-149479326 on Build GRCh38

View Full Product Details